EcoRI nucleotide recognition sequence
Figure color legend
Name derivation: Escherichia coli RY13
Recognition and Cutting site: G^AATT C
C TTAA^G
Resulting DNA Ends: 5′ protruding ends
Prototype: EcoRI
Isoschizomers: None
Sensitivity to methylation:  CpG impaired if overlapped
Neoschizomers None
Anza RE generating compatible ends Anza™ 31 MunI, Anza™ 111 XapI
References