MfeI nucleotide recognition sequence
Figure color legend
Name derivation: Mycoplasma fermentans
Recognition and Cutting site: C^AATT G
G TTAA^C
Resulting DNA Ends: 5′ protruding ends
Prototype: MfeI
Isoschizomers: MunI
Sensitivity to methylation:  None
Neoschizomers None
Anza RE generating compatible ends Anza™ 11 EcoRI, Anza™ 111 XapI
References