Thermo Scientific KingFisher instruments use magnetic bead technology to automate nucleic acid and protein purification. Our automated solution reduces user variability and hands-on time for a variety of applications and throughput levels, delivering a versatile system that can adapt to your changing research needs while maintaining consistent results.
Find the right KingFisher instrument for you
  KingFisher Duo Prime KingFisher Flex KingFisher Presto
Size Compact Benchtop Benchtop Integrate with liquid handler handler
Throughput Low–Med throughput
6/12/24 samples per run/load
Med–High throughput
24/96 samples per run
High throughput
96 samples per run
Volume range 30-5000 µl 20-5000 µl 50-5000 µl
  Request demo
Learn more
Request demo
Learn more
Request demo
Learn more
  Compare all instruments
How KingFisher technology works
The Thermo Scientific KingFisher Purification System utilizes permanent magnetic rods and disposable tip combs. When the magnetic rod—sheathed inside the tip comb—is lowered into the solution, magnetic beads collect at the bottom of the tip comb. The tip comb can then be positioned in a different row or plate, and the beads are then released by moving the magnetic rods out of the tip comb. The tip comb also facilitates the mixing of reagents with the beads by moving up and down.
Watch the video for a detail view of the workflow
Find the right KingFisher instrument for you
  KingFisher Duo Prime KingFisher Flex KingFisher Presto
Size Compact Benchtop Benchtop Integrate with liquid handler handler
Throughput Low–Med throughput
6/12/24 samples per run/load
Med–High throughput
24/96 samples per run
High throughput
96 samples per run
Volume range 30-5000 µl 20-5000 µl 50-5000 µl
  Request demo
Learn more
Request demo
Learn more
Request demo
Learn more
  Compare all instruments
How KingFisher technology works
The Thermo Scientific KingFisher Purification System utilizes permanent magnetic rods and disposable tip combs. When the magnetic rod—sheathed inside the tip comb—is lowered into the solution, magnetic beads collect at the bottom of the tip comb. The tip comb can then be positioned in a different row or plate, and the beads are then released by moving the magnetic rods out of the tip comb. The tip comb also facilitates the mixing of reagents with the beads by moving up and down.
Watch the video for a detail view of the workflow
Optimize your KingFisher instruments with kits & reagents, protocols, and plastics
Nucleic Acids Immunoprecipitation Protein
  • Consistent, high-quality nucleic acid recovery
  • Wide range of sample types
  • Fast, automation-ready protocols with minimal hands-on time
  • Low background—little to no nonspecific binding, and no preclearing and high signal to noise ratio
  • Highly reproducible
  • Fast, automation-ready protocols
  • Antibody savings
  • Highly sensitive
  • Variety of assays and procedures based on affinity purification
  • Rapid processing of samples
  • Compatible with many downstream applications
  • Fast, automation-ready protocols
Order now
Learn more
Order now
Learn more
Order now
Learn more
Download automated protocols Design or customize your own protocols
Nucleic Acid Protocols IP/Protein Protocols BindIT Software
  • Protocols designed for nucleic acid purification procedures and reagent kits
  • Protocols for Immunoprecipitation, Protein A, G, Sheep Anti-Mouse, Sheep Anti-Rabbit, Streptavidin
  • Create and store protocols in a PC database
  • Compatible with all KingFisher instruments
  • Easy to use
  • Generate reports
Search protocols Search protocols
Learn more
Learn more
KingFisher Duo Prime KingFisher Flex KingFisher Presto
  • 96 Deep-Well plate
  • 24 Deep- Well plate
  • 96 KF plate
  • Elution strip
  • Cap for elution strip
  • 12-Tip comb
  • 6-Tip comb
  • Combi pack for 96 samples
  • 96 Deep-Well plate
  • 24 Deep- Well plate
  • 96 KF plate
  • CapMat for 96DW plate
  • 96-Tip comb for DW magnets
  • 96-Tip comb for KF magnets
  • 96-Tip comb for PCR magnets
  • 24 Deep-Well Tip comb and plate
  • 96 Deep-Well plate
  • 24 Deep- Well plate
  • 96 KF plate
  • 96-Tip comb for DW magnets
  • 24 Deep-Well Tip comb and plate
Order now Order now Order now
Nucleic Acids Immunoprecipitation Protein
  • Consistent, high-quality nucleic acid recovery
  • Wide range of sample types
  • Fast, automation-ready protocols with minimal hands-on time
  • Low background—little to no nonspecific binding, and no preclearing and high signal to noise ratio
  • Highly reproducible
  • Fast, automation-ready protocols
  • Antibody savings
  • Highly sensitive
  • Variety of assays and procedures based on affinity purification
  • Rapid processing of samples
  • Compatible with many downstream applications
  • Fast, automation-ready protocols
Order now
Learn more
Order now
Learn more
Order now
Learn more
Download automated protocols Design or customize your own protocols
Nucleic Acid Protocols IP/Protein Protocols BindIT Software
  • Protocols designed for nucleic acid purification procedures and reagent kits
  • Protocols for Immunoprecipitation, Protein A, G, Sheep Anti-Mouse, Sheep Anti-Rabbit, Streptavidin
  • Create and store protocols in a PC database
  • Compatible with all KingFisher instruments
  • Easy to use
  • Generate reports
Search protocols Search protocols
Learn more
Learn more
KingFisher Duo Prime KingFisher Flex KingFisher Presto
  • 96 Deep-Well plate
  • 24 Deep- Well plate
  • 96 KF plate
  • Elution strip
  • Cap for elution strip
  • 12-Tip comb
  • 6-Tip comb
  • Combi pack for 96 samples
  • 96 Deep-Well plate
  • 24 Deep- Well plate
  • 96 KF plate
  • CapMat for 96DW plate
  • 96-Tip comb for DW magnets
  • 96-Tip comb for KF magnets
  • 96-Tip comb for PCR magnets
  • 24 Deep-Well Tip comb and plate
  • 96 Deep-Well plate
  • 24 Deep- Well plate
  • 96 KF plate
  • 96-Tip comb for DW magnets
  • 24 Deep-Well Tip comb and plate
Order now Order now Order now
KingFisher Videos