Straight-from-culture automation
Innovative, straight-from-culture automation
The BenchPro® 2100 Plasmid Purification System is an innovative, automation that provides a completely walk-away solution for high quality purification of your large-scale plasmid DNA. Going straight from culture to purified plasmid, it simplifies the traditional, 22-step process of manual plasmid purification down to four easy set-up steps.
  1. Grow 125 mL of bacterial culture in LB medium with appropriate antibiotic
  2. Assemble the Reagent Tray and Cell Liner on the Waste Tray, and add your culture to the Cell Liner
  3. Load the BenchPro® 2100 Plasmid Purification Card into one of the instrument slots
  4. Run the purification protocol
With a less than 5-minute set-up time, the BenchPro® 2100  delivers large scale, transfection-grade plasmid DNA for all of your downstream application needs.
Unparalleled yield & purity
The BenchPro® 2100 Plasmid Processing Station is our instrument that provides a single, automated program capable of processing up to 1 mg of plasmid DNA from only 125 mL of E. Coli culture for one or two maxipreps at a time. The BenchPro® 2100 Plasmid Purification Card is a disposable, fluidics card that contains a novel anion-exchange membrane, guaranteeing the same, trusted, high-quality plasmid DNA as our manual PureLink™ HiPure kits provide for applications such as transfection of mammalian cells, automated and manual sequencing, PCR amplification, cloning, transformation, and in vitro transcription. The BenchPro® 2100 system is able to reproducibly purify a range of plasmid sizes all the way from from 2.7 to 20 kb from (Figure 1). In addition, the BenchPro® 2100 system outperforms the major competitor’s plasmid maxiprep kits when it comes to yield (Figure 2).
Figure 1. Average DNA yields of plasmids purified using the BenchPro ® 2100 Plasmid Purification System from 125 mL of overnight E. coli culture grown in LB media. (O.D., 2.0–2.4)
[Click image to enlarge]
Figure 2. Average DNA yields from 125 mL of E. coli culture (O.D., 2.0–2.4) of 1 Kb Plus (19 kb) and pcDNA3.1 (6.2 Kb) plasmids purified using the BenchPro ® 2100 Plasmid Purification system and Qiagen® HiSpeed Plasmid Maxi Kit.
[Click image to enlarge]
Consistency & reliability, while eliminating hands-on work & human errors
The BenchPro® 2100 Plasmid Purification System is designed to provide consistency and reliability by using prefilled and sealed reagents. All required buffers in appropriate volumes are supplied in a sealed Reagent Tray while a disposable Cell Liner with lid is included for addition of the culture. The BenchPro® 2100 Plasmid Purification System accurately performs all steps of the purification process to deliver consistent yield (Figure 3) but eliminates the tedious and repetitive liquid-handling steps associated with the manual process of plasmid purification. During the automated purification protocol, the appropriate amounts of reagents are pumped into the card, and incubation steps are performed per the protocol. Using a series of capture and purification membranes, E. coli cells are harvested and lysed, and the lysate is clarified. By eliminating hands on steps, which promote human errors, the BenchPro® 2100 Plasmid Purification System enhances experimental reproducibility and data consistency run after run.
Figure 3. DNA samples (1 µg of pcDNA 3.1; 6.2 kb) purified using the BenchPro ® 2100Plasmid Purification System and Qiagen® HiSpeed Plasmid Maxi Kit loaded onto a 0.8% agarose, ethidium bromide E-gel (20 min run at 70 V). Outside lanes contain 5 µg of 1 kb plus DNA ladder.
[Click image to enlarge]