Society for Laboratory Automation and Screening (SLAS) (2016) San Diego, CA, USA
by J. Braun, N. Roark, V. Blackston, C. Revankar, D. Piper, J. Yang, and J. Chesnut; Thermo Fisher Scientific, Carlsbad, CA, USA - 01/23/2016
Abstract
The CRISPR/Cas9 genome editing system is a powerful tool for genetic screens to identify potential genes that are involved in various biological processes in mammalian cells. As of today, all the screening reports using CRISPR libraries have been performed only in a pooled format. We have developed a high-throughput production process to construct arrayed lentiviral CRISPR libraries with 4 sequence-verified distinct gRNA constructs per gene per well in a 96-well format. The gRNAs are designed to target primarily 5’ coding exons using our proprietary gRNA design tool, attempting to maximize gene-knockout and minimize off-target effects. This arrayed format allows for the much more controlled delivery of gRNA per well, and eliminates the time-consuming deconvolution steps post-screening. The pre-made lentiviral library particles, when co-infected with lentiviral particles expressing Cas9, or alternatively in Cas9 stable expression cell lines, enable high-throughput loss-of-function screens using positive or negative selection strategies in a wide range of mammalian cell types, including primary and non-dividing cells. This new reverse genetic screening tool will enhance our ability for target discovery and validation in health and disease research and provide important information between genetic architecture and phenotypes.
Introduction
Functional characterization studies of the human genome have delivered a tremendous amount of information by using genome based loss-of-function screening in diverse models (1-2). RNA interference (RNAi) has been used as the predominant method for loss-of-function of genome screening, but it is limited by variable efficiency, frequent incompleteness of protein depletion and confounding off-target effects. The CRISPR (Clustered Regularly Interspaced Short Palindromic Repeats)/Cas9 nuclease mediated genome engineering system enables researchers to modify genomic DNA with more precise targeting while minimizing off-target effects (3-5). Certain types of mammalian cells are difficult to transfect using lipid reagents or electroporation.
To circumvent these difficulties, lentiviral vectors are commonly used as another delivery method as they can be easily titrated to manipulate transgene copy number and are stably maintained by integration into the genomic DNA during subsequent cell replication (6). Here we present the high-throughput production process to construct the arrayed lentiviral CRISPR library in a 96-well format. Furthermore, we describe the design and optimization of lentiviral CRISPR library for application in loss-of-function screenings. First, gRNAs are designed to primarily 5’ coding exons of a target gene using our CRISPR gRNA design tool in order to maximize knock-out efficiency and to minimize off-target effects. Four gRNAs per gene are cloned in high-throughput 96-well format followed by lentiviral particles production with a minimum titer of 106 TU/ml, which is calculated by various titration methods; p24 assay, GFP expression of control lentiviral particles, and antibiotic selection for resistant cells.
The subsequent lentiviral CRISPR library is amenable to screening workflows in a variety of methods. Here we show the functional validation of lentiviral CRISPR library particles with genomic cleavage efficiency and treatment with 6-Thioguanine (6-TG), which is one of anti-cancer chemotherapy drugs known as an antimetabolite. The genes that are involved in these pathways or processes could be phenotypically identified in a genetic screen for 6-TG resistance. Moreover, a human kinase loss-of-function screen using our proprietary CellSensor NF-κB-bla ME180 cell line, which is based on the ratiometric blue/green reporter assay, easily enables identification of genomic targets associated with the NF-κB pathway.
Methods
Lentiviral CRISPR Library Expression Constructs. The lentiviral CRISPR library expression constructs is generated by in-house high-throughput automation procedures followed by QC using in-house sequencing processes. 
Packaging of Lentiviral CRISPR Library Particles. Lentiviral particles are generated by in-house high-throughput automation procedures. The viral titer is measured based on p24 assay, GFP expression and antibiotic selection for QC.
Results
Lentiviral CRISPR vectors for the arrayed library
Lentiviral CRISPR Vectors for the Arrayed Library
  • Human codon-optimized S.pyogenes Cas9 protein
  • Blasticidin resistance linked to Cas9 through self cleavage 2A peptide
  • Specific sgRNA from u6 promoter
  • Four targets for each gene
  • Puromycin resistance from EF-1a promoter
  • Targeting 5' exons
  • Filteres off-target sites
Figure 1. Lentiviral CRISPR vectors for the arrayed library generation. The arrayed library screening approach provides controlled delivery of each gRNA per well, eliminating a time-consuming deconvolution step.
QC methods for lentivirus production
Lentiviral particles QC methods Target titer
Cas9 particles Antibiotic selection,
GCD
>107 TU/ml
HPRT1 control particles GFP%, GCD >108 TU/ml
Scramble control particles GFP%, GCD >108 TU/ml
Arrayed library GFP% of controls, GCD, antibiotic for selected plates >106 TU/ml
Pooled library Antibody selection, NGS >108 TU/ml
Figure 2. QC procedures for lentiviral particles production. p24 assay can be used to monitor the purification and biochemical behavior of lentiviral particles. Alternatively, HPRT1 and Scramble gRNA with GFP-Puromycin fusion cassette lentiviral particles are available to measure titer by GFP expression. Antibiotic selection is a common method for viral titration. Genome editing efficiency can be measured by GeneArt Genomic Cleavage Detection kit for functional validation.
High gene targeting efficiency by lentiviral transduction
High Gene Targeting Efficiency by Lentiviral Transduction
Figure 3. Lentiviral transduction to validate the gene targeting efficiency by co-infection with Cas9 lentiviral particles or alternatively in Cas9 stable expression cell line. High genome editing efficiency was observed in both single infection and co-infection as measured by GeneArt Genomic Cleavage Detection (GCD) kit.
Screening workflow of arrayed lentiviral CRISPR library particles
Screening Workflow of Arrayed Lentiviral CRISPR Library Particles
Figure 4. The standard screening process using the arrayed lentiviral CRISPR library includes assay development and optimization, primary screening, and hit identification and validation. Thermo Fisher Scientific provides products to support the entire screening processes.
QC controls for lentivirus production
QC Controls for Lentivirus Production
Figure 5. Functional validation of lentiviral CRISPR-HPRT1 or Scramble (SCR) with GFP-Puromycin fusion cassette particles in Cas9 stable expression cell line (A) Cas9 stable expression HT1080 cells are verified by western blot and the functional efficiency is validated by GCD assay with infection of lentiviral CRISPR-HPRT1 control particles. (B) HPRT1 control lentiviral particles infected cells were treated with 6-TG to confirm the functional effect of gene knock-out. (C) The final output was measured by Crystal Violet staining and PrestoBlue Cell Viability assay.
Lentiviral CRISPR particles for CellSensor NF-kB-bla ME180 cell line
Lentiviral CRISPR particles for CellSensor NF-kB-bla ME180 cell line
Figure 6. A subset of kinases screened in CellSensor NF-kB ME180 cells with a ratiometric reporter assay. After stimulation with TNFa, the ratio of blue/green fluorescence increased in unedited cells. Cells infected with lentiviral particles carrying gRNA that effectively disrupted the NF-κB pathway remained green with low ratio of blue/green fluorescence.
Workflow solutions for genome editing
Workflow Needs and Solutions for Genome Editing
Figure 7. Thermo Fisher Scientific workflow solutions for genome editing. We offer a collection of optimized and validated end-to-end solutions for your entire genome editing workflow, aimed at optimizing conditions and validating the functional efficiency of genome editing.
Conclusions
  • Invitrogen GeneArt arrayed lentiviral CRISPR libraries are powerful high-throughput loss-of-function screening tools for target identification.
  • The gRNA libraries targeting various human gene sets, such as kinases and GPCR are available as pre-made, ready to use lentiviral particles that are arrayed in 96-well plate format.
  • Lentiviral CRISPR library contains 4 sequence-verified distinct gRNA constructs per gene per well.
  • The average titer of lentiviral CRISPR library particles is >106 TU/ml.
  • Invitrogen CellSensor cell lines, combing with lentiviral CRISPR library particles offer simple high-throughput workflow for target screenings.
References
  1. Berns K et al., Nature. 428, 431–437 (2004) A large-scale RNAi screen in human cells identifies new components of the p53 pathway.
  2. Boutros M et al., Heidelberg Fly Array Consortium, Science 303, 832–835 (2004) Genome-wide RNAi analysis of growth and viability in Drosophila cells.
  3. Cong L et al., Science. 339, 819–823 (2013) Multiplex genome engineering using CRISPR/Cas systems.
  4. Mali P et al., Science. 339, 823–826 (2013) RNA-guided human genome engineering via Cas9.
  5. Shalem O et al., Science. 343 (6166): 84-87 (2014) Genome-Scale CRISPR-Cas9 Knockout Screening in Human Cells.
  6. Kabadi AM et al., Nucleic Acids Res. Oct 29;42(19):e147 (2014) Multiplex CRISPR/Cas9-based genome engineering from a single lentiviral vector.