Thermo Scientific variable oxygen control CO2 incubators are uniquely designed to optimize growth of mammalian cells in an environment that closely mimics in vivo conditions. Inside the body, oxygen concentrations range from 1% to 14%, rather than the 20–21% found in the atmosphere. In many cases cells cultured in low oxygen (hypoxia) are designed to grow faster, live longer, and show lower stress.
Since 1979 our variable oxygen control (or “tri-gas”) incubators have been creating hypoxic conditions to help cells grow, so they generate data which is more physiologically relevant.
Contact us
Five reasons for tri-gas CO2 incubators
Why choose a CO2 incubator with variable oxygen control?
1. Increased biological relevance
2. Greater cell number and longer life
3. Reduced differentiation and stress responses
4. Better simulation of tumor microenvironments
5. Variable oxygen control provides optimal growth
"We use the Heracell tri-gas incubator because the integrity of the cell is better, they develop better, and they are healthier."    – Neurobiology researcher, clinical research institute
Choose your tri-gas CO2 incubator
  Forma Series 2 Water Jacketed Forma Series 3 Water Jacketed
  Forma series 2 co2 incubator Forma series 3 co2 incubator
Description Proven benchmark for reliability Smart simplicity with proven reliability
Capacity 184L (6.5 cu. ft.) 184L (6.5 cu. ft.)
Chamber type Water jacketed Water jacketed
Door swing  Left hinged  Left hinged
Interior configuration choices Tri-gas › Tri-gas ›
Sterilization N/A N/A
HEPA filtration ISO Class 5 in 5 min ISO Class 5 in 5 min
Airflow technology Fan assisted Fan assisted
CO2 sensors TC or IR TC or IR
O2 control Optional 1-21% Optional 1-21%
Humidity Removable water pan Removable water pan
User interface Enviro-Scan iCAN touchscreen
  Heracell 150i Heracell VIOS 160i Forma Steri-cycle i160
  Heracell 150i co2 incubator Heracell vios 160i co2 incubator Forma steri-cycle i250 co2 incubator
Description Combines optimal culturing conditions with simplicity and reliability Advanced incubator design for high value cells Advanced incubator design for high value cells
Capacity 150L (5.3 cu. ft.)  165L (5.8 cu. ft.) 165L (5.8 cu. ft.)
Chamber type Direct heat Direct heat Direct heat
Door swing  Left hinged  Left hinged  Left hinged
Interior configuration choices Tri-gas › Tri-gas › Tri-gas ›
Sterilization ContraCon (90°C) Steri-Run (180°C) Steri-Run (180°C)
HEPA filtration N/A ISO Class 5 in 5 min ISO Class 5 in 5 min
Airflow technology Fan assisted THRIVE THRIVE
CO2 sensors TC or IR TC180 or IR180Si TC180 or IR180Si
O2 control Optional 1-21%
or 5-90%
Optional 1-21%
or 5-90%
Optional 1-21%
or 5-90%
Humidity Integrated water reservoir Covered integrated reservoir with easy draining Covered integrated reservoir with easy draining
User interface iCAN touchscreen iCAN touchscreen iCAN touchscreen
  Heracell 240i
Description Offers more capacity with optimal culturing conditions
Capacity 240L (8.4 cu. ft.)
Chamber type Direct heat
Door swing Left hinged
Interior configuration choices Tri-gas ›
Sterilization ContraCon (90°C)
HEPA filtration N/A
Airflow technology Fan assisted
CO2 sensors TC or IR
O2 control Optional 1-21%
or 5-90%
Humidity Integrated water reservoir
User interface iCAN touchscreen
  Forma Series 2 Water Jacketed Forma Series 3 Water Jacketed
  Forma series 2 co2 incubator Forma series 3 co2 incubator
Description Proven benchmark for reliability Smart simplicity with proven reliability
Capacity 184L (6.5 cu. ft.) 184L (6.5 cu. ft.)
Chamber type Water jacketed Water jacketed
Door swing  Left hinged  Left hinged
Interior configuration choices Tri-gas › Tri-gas ›
Sterilization N/A N/A
HEPA filtration ISO Class 5 in 5 min ISO Class 5 in 5 min
Airflow technology Fan assisted Fan assisted
CO2 sensors TC or IR TC or IR
O2 control Optional 1-21% Optional 1-21%
Humidity Removable water pan Removable water pan
User interface Enviro-Scan iCAN touchscreen
  Heracell 150i Heracell VIOS 160i Forma Steri-cycle i160
  Heracell 150i co2 incubator Heracell vios 160i co2 incubator Forma steri-cycle i250 co2 incubator
Description Combines optimal culturing conditions with simplicity and reliability Advanced incubator design for high value cells Advanced incubator design for high value cells
Capacity 150L (5.3 cu. ft.)  165L (5.8 cu. ft.) 165L (5.8 cu. ft.)
Chamber type Direct heat Direct heat Direct heat
Door swing  Left hinged  Left hinged  Left hinged
Interior configuration choices Tri-gas › Tri-gas › Tri-gas ›
Sterilization ContraCon (90°C) Steri-Run (180°C) Steri-Run (180°C)
HEPA filtration N/A ISO Class 5 in 5 min ISO Class 5 in 5 min
Airflow technology Fan assisted THRIVE THRIVE
CO2 sensors TC or IR TC180 or IR180Si TC180 or IR180Si
O2 control Optional 1-21%
or 5-90%
Optional 1-21%
or 5-90%
Optional 1-21%
or 5-90%
Humidity Integrated water reservoir Covered integrated reservoir with easy draining Covered integrated reservoir with easy draining
User interface iCAN touchscreen iCAN touchscreen iCAN touchscreen
  Heracell 240i
Description Offers more capacity with optimal culturing conditions
Capacity 240L (8.4 cu. ft.)
Chamber type Direct heat
Door swing Left hinged
Interior configuration choices Tri-gas ›
Sterilization ContraCon (90°C)
HEPA filtration N/A
Airflow technology Fan assisted
CO2 sensors TC or IR
O2 control Optional 1-21%
or 5-90%
Humidity Integrated water reservoir
User interface iCAN touchscreen
Five reasons to enhance your cell culture using a CO2 incubator
with variable oxygen control
Our tri-gas CO2 incubators are uniquely designed to optimize growth of mammalian cells in an environment that closely mimics in vivo conditions. Maximize your productivity by surrounding your important cells in an environment you can trust.
 The 5 Reasons for Variable O2 Control Brochure ›
Top
Key features to help you achieve your next discovery
Based on 60 years of engineering and design breakthroughs, our expertise is represented in key features that span our portfolio. Together these enhancements provide optimal cell growth, valuable contamination control technologies, and advanced design for highly critical applications, plus simplicity which allows you to spend more time pursuing your goals.
On-demand high temperature sterilization cycles for easy cleaning.
  • Our designs are independently tested to meet U.S. and EU pharmacopeia standards
  • Automatically radiates heat uniformly to all interior surfaces and quickly returns to selected operating conditions
  • Simplifies cleaning without the need for autoclaving and eliminates variability in manual cleaning
  • Independent third-party tests prove elimination of biological contaminants including fungi, spores, mycoplasma, and other bacteria
  • Avoids physical constraints of UV germicidal lamps and ongoing costs, handling and storage of potentially toxic germicides
  Thermo Scientific ContraCon Sterilization Cycle 140°C Sterilization Cycle Thermo Scientific Steri-Run Sterilization Cycle
Temperature type 90°C moist heat cycle 140°C dry heat cycle 180°C dry heat cycle
Description Push button simplicity without removal of sensors and components. Overnight sterilization cycle with extended time at 140°C to achieve sterilization. The new standard, an overnight cycle that reaches 180°C on all chamber surfaces.
Duration of sterilization cycle 24–25 hours  14 hours 12 hours
Sterility assurance level (SAL) 6 log 12 log 12 log
Independent validation
  Learn more:
 ContraCon Eliminates Contamination ›
Learn more:
 Effective Heat Sterilization in CO2 Incubators ›
Learn more:
 Steri-Run Sterilization Cycle Proves Total Sterilization ›
Thermo Scientific CO2 incubators feature fan-assisted airflow circulation for fast recovery and tight uniformity. Our airflow patterns are specifically designed for homogeneous distribution of critical environmental conditions (temperature, gas exchange and humidity). The in-chamber fan gently distributes the conditioned, humidified air throughout the chamber, ensuring all cells—regardless of location—experience the same conditions without risk of desiccation.1
  • Temperature uniformity specifications are validated according to the DIN 12880 standard, which requires 27 individual measurements at specific locations in the chamber.
  • Sensors and probes are positioned directly in the incubator chamber to ensure they measure the exact environment experienced by your cells.
  • A secondary temperature probe acts as an alarm to the primary sensor, providing peace of mind for you and extra security for your cells.
See THRIVE active airflow in action
Thermo Scientific THRIVE active airflow, included on our Heracell VIOS 160i and 250i and Forma Steri-Cycle i160 and i250 CO2 incubators, provides total recovery of critical growth parameters in 10 minutes or less following a 30-second door opening.1
  • The variable speed fan precisely controls the airflow to provide leading edge recovery.
  • The air stream first travels over the direct-heated covered, integrated water reservoir, which results in 50% faster humidity recovery than a standard water pan design.
  • The in-line HEPA filter cleans the airstream of all sizes of particles and microorganisms, protecting cultures from contamination.
  • The air then passes over the in-chamber sensors, which react and condition the air before gently circulating it among the cultures.
1 Based on internal standards for 30-second door opening, recovery calculated to 98% of starting value for temperature and CO2 , and 95% of starting value of humidity.
Because the goal is to mimic in vivo conditions, all incubator sensors are positioned directly in the incubation chamber to ensure they measure the exact environment experienced by your cells. This design requires more robust sensor construction but means the sensors are designed to react immediately to correct temperature and atmosphere variations as they occur. We offer two different CO2 sensor technologies.
Thermal conductivity (TC) CO2 sensor
  • Our most commonly requested CO2 sensor due to its reliability and longevity of operation
  • Measures electrical resistance according to thermal conductivity of gas passing over the sensor
  • Low maintenance, stable, and long life because it has no consuming element such as an incandescent bulb
  • High temperature resistance—does not need to be removed during sterilization cycles
  • NEW TC180 CO2 sensor incorporates humidity compensation for enhanced stability and recovery (available in the Heracell VIOS and Forma Steri-Cycle CO2 incubators)
Infrared (IR) CO2 sensor
  • Ideal for sensitive applications and remote monitoring or when frequent access to incubator is required
  • Determines the CO2 concentration based on its IR light absorption at the characteristic wavelength of 4.3 µm
  • Operates independent of humidity and temperature
  • NEW IR180Si replaces the traditional incandescent IR light source with silicone MEMS emitter technology for improved stability and service life (available in the Heracell VIOS and Forma Steri-Cycle CO2 incubators)
Added culturing flexibility with variable oxygen control (tri-gas)
Increase biological relevance by simulating physiological conditions, resulting in cell behaviors that are more predictive of an in vivo environment.
  • Choice of 1–21% and 5–90% O2 ranges
  • Simulate hypoxic (1–21%) environments for primary cell, stem cell, and embryo research applications
  • Achieve better simulation of tumor microenvironments with low oxygen culturing
  • Simulate hyperoxic (5–90%) conditions for research in lung, retina, and other sensitive tissues
Featured resource
Five reasons to enhance your cell culture using a Thermo Scientific CO2 incubator with variable oxygen control
Learn more
A selection of humidity reservoir designs are available to enhance performance and workflows. Our unique integrated solutions are directly heated to facilitate high relative humidity (rH) levels and faster recovery. An external humidity reservoir simplifies maintenance by allowing water refill without disrupting culturing. Full active rH parameter control is ideal for applications requiring flexibility and precise monitoring of humidity levels.
  Water pan Integrated water reservoir Covered integrated water reservoir External humidity reservoir
Description Traditional removable humidity pan Large directly heated panless reservoir provides high rH levels and fast recovery Protected, directly heated integrated water pan provides high rH levels with leading recovery and easy filling and draining Active humidity control with an external reservoir for easy replenishment of water
Volume 3L 3L (4.5L for large capacity models) 3L 3L
Water level monitoring   Yes, low level alarm Yes, visual monitoring and low level alarm Yes, visual monitoring and low level alarm
Popular models Forma Direct Heat and Steri-Cycle; Forma Series 2 and Series 3 Water Jacketed; Midi 40
Heracell 150i and 240i
Heracell VIOS 160i and 250i; Forma Steri-Cycle i160 and i250
Forma Steri-Cult; BBD 6220
iCAN touchscreen interface
The intelligent Thermo Scientific iCAN touchscreen interface is designed to simplify your interactions with the CO2 incubator. The bright LED screen provides at-a-glance monitoring even from across the lab. Enjoy complete data visibility and performance trend graphing, in multiple languages. Store data and usage logs onboard or download.
  • Door mounted position for easy access
  • Configurable settings and options
  • Audio and visual alarms
  • On-screen menu prompts
  • Error and usage logs
  • Data logging and performance trend graphing
  • Multiple language selection
Enviro-Scan interface
The Thermo Scientific Enviro-Scan LED membrane display lets you easily monitor temperature, CO2, and rH (optional) operating parameters via the bright display. The alphanumeric message center provides process and alarm status.
  • Configurable settings and options
  • Audio and visual alarms
  • On-screen menu prompts
CO2 incubator selector tool
With so many choices, which Thermo Scientific CO2 incubator should you choose? Based on your cell culture applications, needs, or preferences, our selector can help you decide on the best incubator for your lab.
Go to selector tool
Related product categories