compact benchtop centrifuges
Our small benchtop centrifuges were engineered to fit in your lab space and designed to anticipate and address your lab's most pressing needs. This line of centrifuges accommodates multi‐laboratory settings, with the flexibility to adapt to evolving clinical and research applications. A floor model is available for laboratories where bench space is limited.
Browse our complete portfolio or let one of our experts help you make a selection.
Contact us
Optimize bench space
while achieving capacity
compact benchtop centrifuge optimizes space
Process up to 8 x 50 mL conical tubes at over 12,000 x g, 24 x 5/7 mL blood tubes, microplates and a wide range of microtubes at speeds up to 30,279 x g, all in one versatile benchtop centrifuge.
Say yes to a more ergonomic design
ergonomic compact benchtop centrifuge
Thermo Scientific innovation brings Auto-Lock Rotor Exchange, a secure push-button rotor installation, plus ClickSeal Biocontainment Lids, for glove-friendly, one-handed operation.
Accommodate more with
14 rotor options
compact benchtop centrifuge microplate rotor
Support applications from clinical chemistry, clinical microbiology, hematology, immunology, to cellular biology, genomics/molecular biology, proteomics, and biochemistry, plus clinical and pharmaceutical studies.
Quick links
What kind of compact centrifuge fits your needs best?
Our industry-leading portfolio of compact centrifuges includes multiple models that come in ventilated, refrigerated, and floor-standing units in trusted brands and voltages necessary to meet the needs of our customers around the world. Choose your favorite brand below.
Ventilated
Refrigerated
Floor-standing
(portable)
  ventilated compact benchtop centrifuge refrigerated compact benchtop centrifuge floor-standing compact benchtop centrifuge
Request a quote
  A compact platform with multiple choices of rotor, including both fixed angle and swinging bucket. Outstanding capacity up to 24 x 5/6mL blood tubes. Smart and simple interface with four customized program memory. High-speed microliter performance at up to 30,279 x g enables quicker spins and improved separations. Designed for a wide range of applications including clinical diagnostic, molecular biology and cell culture. Ergonomically designed floor-standing model enhances placement flexibility in the lab. An ideal choice of centrifuge to improve the workflow efficiency in the cell culture lab.
Max capacity 6 x 50mL (fixed angle)
4 x 145mL (swinging bucket)
6 x 50mL (fixed angle)
4 x 145mL (swinging bucket)
6 x 50mL (fixed angle)
4 x 145mL (swinging bucket)
Max speed 16,000 rpm (fixed angle)
4,500 rpm (swinging bucket)
17,850 rpm (fixed angle)
4,500 rpm (swinging bucket)
17,850 rpm (fixed angle)
4,500 rpm (swinging bucket)
Max RCF 24,328 x g (fixed angle)
3,260 x g (swinging bucket)
30,279 x g (fixed angle)
3,260 x g (swinging bucket)
30,279 x g (fixed angle)
3,260 x g (swinging bucket)
Programs 4 direct program buttons 3 direct program buttons, 96 additional programs accessible via folder
3 direct program buttons,
96 additional programs accessible via folder
Dimensions
(H x W x D)
Lid closed:
12.2 x 14.6 x 18.9 in.
(31 x 37 x 48cm)
Lid open:
26.8 x 14.6 x 18.9 in.
(67 x 37 x 48cm)
Lid closed:
12.6 x 18.1 x 26.4 in.
(32 x 46 x 67cm)
Lid open:
27.5 x 18.1 x 26.4 in.
(70 x 46 x 67cm)
Lid closed:
30.7 to 32.7 (adjustable) x 18.3 x 20.5 in.
(79 to 84 x 47 x 52cm)
Lid open:
30.7 to 32.7 (adjustable) x 18.3 x 20.5 in.
(117 to 122 x 47 x 52cm)
Weight 77 lbs. (35 kg) 156 lbs. (71kg)
191 lbs. (87kg)
Display type LCD LCD LCD
Ventilated
Refrigerated
  ventilated compact benchtop centrifuge refrigerated compact benchtop centrifuge
Request a quote
  A compact platform with multiple choices of rotor, including both fixed angle and swinging bucket. Outstanding capacity up to 24 x 5/6mL blood tubes. Smart and simple interface with four customized program memory. High-speed microliter performance at up to 30,279 x g enables quicker spins and improved separations. Designed for a wide range of applications including clinical diagnostic, molecular biology and cell culture.
Max capacity 6 x 50mL (fixed angle)
4 x 145mL (swinging bucket)
6 x 50mL (fixed angle)
4 x 145mL (swinging bucket)
Max speed 16,000 rpm (fixed angle)
4,500 rpm (swinging bucket)
17,850 rpm (fixed angle)
4,500 rpm (swinging bucket)
Max RCF 24,328 x g (fixed angle)
3,260 x g (swinging bucket)
30,279 x g (fixed angle)
3,260 x g (swinging bucket)
Programs 4 direct program buttons 3 direct program buttons, 96 additional programs accessible via folder
Dimensions w/lid closed (H x W x D) 12.2 x 14.6 x 18.9 in.
(31 x 37 x 48cm)
12.6 x 18.1 x 26.4 in.
(32 x 46 x 67cm)
Weight 77 lbs. (35 kg) 156 lbs. (71kg)
Display type LCD LCD
Ventilated
Refrigerated
  ventilated compact benchtop centrifuge refrigerated compact benchtop centrifuge
Request a quote
  A compact platform with multiple choices of rotor, including both fixed angle and swinging bucket. Outstanding capacity up to 24 x 5/6mL blood tubes. Smart and simple interface with four customized program memory. High-speed microliter performance at up to 30,279 x g enables quicker spins and improved separations. Designed for wide range of applications including clinical diagnostic, molecular biology and cell culture.
Max capacity 6 x 50mL (fixed angle)
4 x 145mL (swinging bucket)
6 x 50mL (fixed angle)
4 x 145mL (swinging bucket)
Max speed 16,000 rpm (fixed angle)
4,500 rpm (swinging bucket)
17,850 rpm (fixed angle)
4,500 rpm (swinging bucket)
Max RCF 24,328 x g (fixed angle)
3,260 x g (swinging bucket)
30,279 x g (fixed angle)
3,260 x g (swinging bucket)
Programs 4 direct program buttons 3 direct program buttons, 96 additional programs accessible via folder
Dimensions w/lid closed (HWD) 12.2 x 14.6 x 18.9 in.
(31 x 37 x 48cm)
12.6 x 18.1 x 26.4 in.
(32 x 46 x 67cm)
Weight 77 lbs. (35 kg) 156 lbs. (71kg)
Display type LCD LCD
Ventilated
Refrigerated
Floor-standing
(portable)
  ventilated compact benchtop centrifuge refrigerated compact benchtop centrifuge floor-standing compact benchtop centrifuge
Request a quote
  A compact platform with multiple choices of rotor, including both fixed angle and swinging bucket. Outstanding capacity up to 24 x 5/6mL blood tubes. Smart and simple interface with four customized program memory. High-speed microliter performance at up to 30,279 x g enables quicker spins and improved separations. Designed for a wide range of applications including clinical diagnostic, molecular biology and cell culture. Ergonomically designed floor-standing model enhances placement flexibility in the lab. An ideal choice of centrifuge to improve the workflow efficiency in the cell culture lab.
Max capacity 6 x 50mL (fixed angle)
4 x 145mL (swinging bucket)
6 x 50mL (fixed angle)
4 x 145mL (swinging bucket)
6 x 50mL (fixed angle)
4 x 145mL (swinging bucket)
Max speed 16,000 rpm (fixed angle)
4,500 rpm (swinging bucket)
17,850 rpm (fixed angle)
4,500 rpm (swinging bucket)
17,850 rpm (fixed angle)
4,500 rpm (swinging bucket)
Max RCF 24,328 x g (fixed angle)
3,260 x g (swinging bucket)
30,279 x g (fixed angle)
3,260 x g (swinging bucket)
30,279 x g (fixed angle)
3,260 x g (swinging bucket)
Programs 4 direct program buttons 3 direct program buttons, 96 additional programs accessible via folder
3 direct program buttons,
96 additional programs accessible via folder
Dimensions
(H x W x D)
Lid closed:
12.2 x 14.6 x 18.9 in.
(31 x 37 x 48cm)
Lid open:
26.8 x 14.6 x 18.9 in.
(67 x 37 x 48cm)
Lid closed:
12.6 x 18.1 x 26.4 in.
(32 x 46 x 67cm)
Lid open:
27.5 x 18.1 x 26.4 in.
(70 x 46 x 67cm)
Lid closed:
30.7 to 32.7 (adjustable) x 18.3 x 20.5 in.
(79 to 84 x 47 x 52cm)
Lid open:
30.7 to 32.7 (adjustable) x 18.3 x 20.5 in.
(117 to 122 x 47 x 52cm)
Weight 77 lbs. (35 kg) 156 lbs. (71kg)
191 lbs. (87kg)
Display type LCD LCD LCD
Ventilated
Refrigerated
  ventilated compact benchtop centrifuge refrigerated compact benchtop centrifuge
Request a quote
  A compact platform with multiple choices of rotor, including both fixed angle and swinging bucket. Outstanding capacity up to 24 x 5/6mL blood tubes. Smart and simple interface with four customized program memory. High-speed microliter performance at up to 30,279 x g enables quicker spins and improved separations. Designed for a wide range of applications including clinical diagnostic, molecular biology and cell culture.
Max capacity 6 x 50mL (fixed angle)
4 x 145mL (swinging bucket)
6 x 50mL (fixed angle)
4 x 145mL (swinging bucket)
Max speed 16,000 rpm (fixed angle)
4,500 rpm (swinging bucket)
17,850 rpm (fixed angle)
4,500 rpm (swinging bucket)
Max RCF 24,328 x g (fixed angle)
3,260 x g (swinging bucket)
30,279 x g (fixed angle)
3,260 x g (swinging bucket)
Programs 4 direct program buttons 3 direct program buttons, 96 additional programs accessible via folder
Dimensions w/lid closed (H x W x D) 12.2 x 14.6 x 18.9 in.
(31 x 37 x 48cm)
12.6 x 18.1 x 26.4 in.
(32 x 46 x 67cm)
Weight 77 lbs. (35 kg) 156 lbs. (71kg)
Display type LCD LCD
Ventilated
Refrigerated
  ventilated compact benchtop centrifuge refrigerated compact benchtop centrifuge
Request a quote
  A compact platform with multiple choices of rotor, including both fixed angle and swinging bucket. Outstanding capacity up to 24 x 5/6mL blood tubes. Smart and simple interface with four customized program memory. High-speed microliter performance at up to 30,279 x g enables quicker spins and improved separations. Designed for wide range of applications including clinical diagnostic, molecular biology and cell culture.
Max capacity 6 x 50mL (fixed angle)
4 x 145mL (swinging bucket)
6 x 50mL (fixed angle)
4 x 145mL (swinging bucket)
Max speed 16,000 rpm (fixed angle)
4,500 rpm (swinging bucket)
17,850 rpm (fixed angle)
4,500 rpm (swinging bucket)
Max RCF 24,328 x g (fixed angle)
3,260 x g (swinging bucket)
30,279 x g (fixed angle)
3,260 x g (swinging bucket)
Programs 4 direct program buttons 3 direct program buttons, 96 additional programs accessible via folder
Dimensions w/lid closed (HWD) 12.2 x 14.6 x 18.9 in.
(31 x 37 x 48cm)
12.6 x 18.1 x 26.4 in.
(32 x 46 x 67cm)
Weight 77 lbs. (35 kg) 156 lbs. (71kg)
Display type LCD LCD
Need something even smaller?
We also have mini centrifuges and microcentrifuges.
Top
Rotors for compact centrifuges
Our extensive rotor portfolio provides application flexibility to support multiple lab settings, multiple users, and versatility as the needs of your laboratory evolve.
Swinging bucket rotors
compact benchtop centrifuge swinging bucket rotor
Fixed angle rotors
ompact benchtop centrifuge fixed angle rotors
Top
Sorvall ST8 compact centrifuge brochure
The Sorvall ST8 compact centrifuge series fits into your space and your applications with its compact footprint, ergonomic design and multiple rotor options that adapt to evolving clinical and research needs. It stands out for its outstanding reliability, consistency, and safety.
  • Simple solutions
  • Flexible processing
  • Reliable quality
  • Safe operation
Open brochure
Top
Take a virtual tour of our benchtop centrifuges
Take a self-guided tour and learn how our centrifuges and rotors are designed around you and your applications.
They fit in to: They stand out for:
  • your space
  • your processes and protocols
  • the evolving needs of your
    future in the lab
  • innovative design and technologies
  • solid reliability
  • consistent results
While in the tour, take a look at our convenient packages to find just the right centrifuge, rotor, and adapter combination you need to get started.
Take a tour
Top
Benchtop centrifuges videos
Top
Related products
Microcentrifuges
Floor models
Centrifuge rotors
Centrifuge ware
Resources
Support
  • Unity Lab Services
    Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.