Ideal for research or controlled environments, our high performance benchtop centrifuges meet customers' needs for application flexibility and high speeds. They come in medium 1.6L and large 4L sizes with floor/portable options, and they offer a high performing drive with an advanced operating interface designed for maximum protocol versatility, complementing the numerous rotor configurations available.
Get unparalleled processing performance, maximum versatility, and exceptional energy efficiency with our high performance benchtop centrifuges.
Innovative Thermo Scientific rotor technologies
One-handed sample protection, biocontainment certified*
Glove-friendly, one-handed open/close capability of our ClickSeal biocontainment lids provide simple operation for all laboratory users, eliminating multi-turn screw caps and complicated high pressure clips.
Secure, push-button rotor exchange in as little as 3 seconds
Trouble-free rotor installation and removal with Auto-Lock rotor exchange provides full chamber access for cleaning convenience, quick switch between applications, and the flexibility to evolve with the needs of your lab. No tools required. No complicated rotor changing procedures.
Outstanding durability, ergonomics and performance
Lightweight carbon fiber and innovative construction allow unique rotor designs not possible with metal. Plus, their corrosion- and fatigue-resistance secures the rotor’s structural integrity for outstanding durability, and it's backed by a 15-year limited warranty.**
Quick links
Which high performance benchtop centrifuge fits your needs best?
Easy to use with intuitive controls and a design for easy sample loading, unloading, and cleaning.
Choose 1.6 liter capacity models ›
  • Max capacity 4 x 400mL
  • Max speed 15,200rpm
  • Max RCF 25,830 x g
  • 14 optional rotors
Choose 4 liter capacity models › 
  • Max capacity 4 x 1000mL
  • Max speed 15,200rpm
  • Max RCF 25,830 x g
  • 17 optional rotors
Learn more
Speed and flexibility for reliable separations—all in a compact footprint, featuring intelligent design that eliminates imbalances for maximum productivity and results.
  • Max capacity 4 x 180mL
  • Max speed 23,300rpm
  • Max RCF 50,377 x g
  •  9 optional rotors
1.6L (4 x 400mL) benchtop centrifuges
Ventilated
Refrigerated
  ventilated high performance benchtop centrifuge refrigerated high performance benchtop centrifuge
Request a quote
  Ventilated centrifuges are great for samples that are not temperature sensitive. Remember that ventilated centrifuges may not be at the laboratory's ambient room temperature, since spinning rotors create heat. Any samples that can be negatively impacted by an increase in temperature would benefit. When spinning large capacity rotors at higher speeds, there is more heat; refrigeration can maintain optimal temperatures.
Max capacity   6 x 100mL (fixed angle)
4 x 400mL (swinging bucket)
6 x 100mL (fixed angle)
4 x 400mL (swinging bucket)
Max speed 15,200rpm (fixed angle)
6,300rpm (swinging bucket)
15,200rpm (fixed angle)
6,300rpm (swinging bucket)
Max RCF 25,830 x g (fixed angle)
7,188 x g (swinging bucket)
25,830 x g (fixed angle)
7,188 x g (swinging bucket)
Programs 99, 5 with direct program buttons 99, 5 with direct program buttons
Dimensions (H x W x D) Lid closed:
14.2 x 17.3 x 23.8 in.
(36 x 44 x 60.5 cm)
Lid opened:
26 x 17.3 x 23.8 in.
(66 x 44 x 60.5 cm)
Lid closed:
14.2 x 24.5 x 23.8 in.
(36 x 62.3 x 60.5 cm)
Lid opened:
26 x 24.5 x 23.8 in.
(66 x 62.3 x 60.5 cm)
Weight 126.8 lbs.
57.5kg
201.7 lbs.
91.5kg
Display type LCD LCD
Ventilated
Refrigerated
  ventilated high performance benchtop centrifuge refrigerated high performance benchtop centrifuge
Request a quote
  Ventilated centrifuges are great for samples that are not temperature sensitive. Remember that ventilated centrifuges may not be at the laboratory's ambient room temperature, since spinning rotors create heat. Any samples that can be negatively impacted by an increase in temperature would benefit. When spinning large capacity rotors at higher speeds, there is more heat; refrigeration can maintain optimal temperatures.
Max capacity   6 x 100mL (fixed angle)
4 x 400mL (swinging bucket)
6 x 100mL (fixed angle)
4 x 400mL (swinging bucket)
Max speed 15,200rpm (fixed angle)
6,300rpm (swinging bucket)
15,200rpm (fixed angle)
6,300rpm (swinging bucket)
Max RCF 25,830 x g (fixed angle)
7,188 x g (swinging bucket)
25,830 x g (fixed angle)
7,188 x g (swinging bucket)
Programs 99, 5 with direct program buttons 99, 5 with direct program buttons
Dimensions (H x W x D) Lid closed:
14.2 x 17.3 x 23.8 in.
(36 x 44 x 60.5 cm)
Lid opened:
26 x 17.3 x 23.8 in.
(66 x 44 x 60.5 cm)
Lid closed:
14.2 x 24.5 x 23.8 in.
(36 x 62.3 x 60.5 cm)
Lid opened:
26 x 24.5 x 23.8 in.
(66 x 62.3 x 60.5 cm)
Weight 126.8 lbs.
57.5kg
201.7 lbs.
91.5kg
Display type LCD LCD
Ventilated
Refrigerated
  ventilated high performance benchtop centrifuge refrigerated high performance benchtop centrifuge
Request a quote
  Ventilated centrifuges are great for samples that are not temperature sensitive. Remember that ventilated centrifuges may not be at the laboratory's ambient room temperature, since spinning rotors create heat. Any samples that can be negatively impacted by an increase in temperature would benefit. When spinning large capacity rotors at higher speeds, there is more heat; refrigeration can maintain optimal temperatures.
Max capacity   6 x 100mL (fixed angle)
4 x 400mL (swinging bucket)
6 x 100mL (fixed angle)
4 x 400mL (swinging bucket)
Max speed 15,200rpm (fixed angle)
6,300rpm (swinging bucket)
15,200rpm (fixed angle)
6,300rpm (swinging bucket)
Max RCF 25,830 x g (fixed angle)
7,188 x g (swinging bucket)
25,830 x g (fixed angle)
7,188 x g (swinging bucket)
Programs 99, 5 with direct program buttons 99, 5 with direct program buttons
Dimensions (H x W x D) Lid closed:
14.2 x 17.3 x 23.8 in.
(36 x 44 x 60.5 cm)
Lid opened:
26 x 17.3 x 23.8 in.
(66 x 44 x 60.5 cm)
Lid closed:
14.2 x 24.5 x 23.8 in.
(36 x 62.3 x 60.5 cm)
Lid opened:
26 x 24.5 x 23.8 in.
(66 x 62.3 x 60.5 cm)
Weight 126.8 lbs.
57.5kg
201.7 lbs.
91.5kg
Display type LCD LCD
Ventilated
Refrigerated
  ventilated high performance benchtop centrifuge refrigerated high performance benchtop centrifuge
Request a quote
  Ventilated centrifuges are great for samples that are not temperature sensitive. Remember that ventilated centrifuges may not be at the laboratory's ambient room temperature, since spinning rotors create heat. Any samples that can be negatively impacted by an increase in temperature would benefit. When spinning large capacity rotors at higher speeds, there is more heat; refrigeration can maintain optimal temperatures.
Max capacity   6 x 100mL (fixed angle)
4 x 400mL (swinging bucket)
6 x 100mL (fixed angle)
4 x 400mL (swinging bucket)
Max speed 15,200rpm (fixed angle)
6,300rpm (swinging bucket)
15,200rpm (fixed angle)
6,300rpm (swinging bucket)
Max RCF 25,830 x g (fixed angle)
7,188 x g (swinging bucket)
25,830 x g (fixed angle)
7,188 x g (swinging bucket)
Programs 99, 5 with direct program buttons 99, 5 with direct program buttons
Dimensions (H x W x D) Lid closed:
14.2 x 17.3 x 23.8 in.
(36 x 44 x 60.5 cm)
Lid opened:
26 x 17.3 x 23.8 in.
(66 x 44 x 60.5 cm)
Lid closed:
14.2 x 24.5 x 23.8 in.
(36 x 62.3 x 60.5 cm)
Lid opened:
26 x 24.5 x 23.8 in.
(66 x 62.3 x 60.5 cm)
Weight 126.8 lbs.
57.5kg
201.7 lbs.
91.5kg
Display type LCD LCD
Top
4L (4 x 1000mL) benchtop centrifuges
Ventilated
Refrigerated
Floor standing (portable)
Floor standing (portable) refrigerated
  ventilated high performance benchtop centrifuge refrigerated high performance benchtop centrifuge portable high performance benchtop centrifuge floor standing high performance benchtop centrifuge
Request a quote
  Ventilated centrifuges are great for samples that are not temperature sensitive. Remember that ventilated centrifuges may not be at the laboratory's ambient room temperature, since spinning rotors create heat.
Any samples that can be negatively impacted by an increase in temperature would benefit. When spinning large capacity rotors at higher speeds, there is more heat; refrigeration can maintain optimal temperatures.
Along with our ventilated benchtop models, floor models offer an option to save bench space. Our high performance, refrigerated floor models offer optimal flexibility in both your experimental applications as well as your laboratory workflow saving precious bench space.
Max capacity   6 x 250mL (fixed angle)
4 x 1000mL (swinging bucket)
6 x 250mL (fixed angle)
4 x 1000mL (swinging bucket)
6 x 250mL (fixed angle)
4 x 1000mL (swinging bucket)
6 x 250mL (fixed angle)
4 x 1000mL (swinging bucket)
Max speed 15,200rpm (fixed angle)
6,300rpm (swinging bucket)
15,200rpm (fixed angle)
6,300rpm (swinging bucket)
15,200rpm (fixed angle)
6,300rpm (swinging bucket)
15,200rpm (fixed angle)
6,300rpm (swinging bucket)
Max RCF 25,830 x g (fixed angle)
7,164 x g (swinging bucket)
25,830 x g (fixed angle)
7,164 x g (swinging bucket)
25,830 x g (fixed angle)
7,164 x g (swinging bucket)
25,830 x g (fixed angle)
7,164 x g (swinging bucket)
Programs 99, 5 direct program buttons 99, 5 direct program buttons 99, 5 direct program buttons 99, 5 direct program buttons
Dimensions  (H x W x D) Lid closed:
14.2 x 22.2 x 26.4 in.
(36 x 56.5 x 67 cm)
Lid opened:
30.3 x 22.2 x 26.4 in.
(77 x 56.5 x 67 cm)
Lid closed:
14.2 x 24.5 x 23.8 in.
(36 x 62.3 x 60.5 cm)
Lid opened:
30.3 x 29.3 x 26.4 in.
(77 x 74.5 x 67 cm)
Lid closed:
33.2 x 22.2 x 26.4 in.
(84 x 56.5 x 67 cm)
Lid opened:
49.2 x 22.2 x 26.4 in.
(125 x 56.5 x 67 cm)
Lid closed:
33.2 x 22.2 x 26.4 in.
(84 x 56.5 x 67 cm)
Lid opened:
49.2 x 22.2 x 26.4 in.
(125 x 56.5 x 67 cm)
Weight 189.6 lbs.
86kg
255.7 lbs.
116kg
314.2 lbs.
142.5kg
336.2 lbs.
152.5kg
Display type LCD LCD LCD LCD
Ventilated
Refrigerated
Floor standing (portable)
Floor standing (portable) refrigerated
  ventilated high performance benchtop centrifuge refrigerated high performance benchtop centrifuge portable high performance benchtop centrifuge floor standing high performance benchtop centrifuge
Request a quote
  Ventilated centrifuges are great for samples that are not temperature sensitive. Remember that ventilated centrifuges may not be at the laboratory's ambient room temperature, since spinning rotors create heat.
Any samples that can be negatively impacted by an increase in temperature would benefit. When spinning large capacity rotors at higher speeds, there is more heat; refrigeration can maintain optimal temperatures.
Along with our ventilated benchtop models, floor models offer an option to save bench space. Our high performance, refrigerated floor models offer optimal flexibility in both your experimental applications as well as your laboratory workflow saving precious bench space.
Max capacity   6 x 250mL (fixed angle)
4 x 1000mL (swinging bucket)
6 x 250mL (fixed angle)
4 x 1000mL (swinging bucket)
6 x 250mL (fixed angle)
4 x 1000mL (swinging bucket)
6 x 250mL (fixed angle)
4 x 1000mL (swinging bucket)
Max speed 15,200rpm (fixed angle)
6,300rpm (swinging bucket)
15,200rpm (fixed angle)
6,300rpm (swinging bucket)
15,200rpm (fixed angle)
6,300rpm (swinging bucket)
15,200rpm (fixed angle)
6,300rpm (swinging bucket)
Max RCF 25,830 x g (fixed angle)
7,164 x g (swinging bucket)
25,830 x g (fixed angle)
7,164 x g (swinging bucket)
25,830 x g (fixed angle)
7,164 x g (swinging bucket)
25,830 x g (fixed angle)
7,164 x g (swinging bucket)
Programs 99, 5 direct program buttons 99, 5 direct program buttons 99, 5 direct program buttons 99, 5 direct program buttons
Dimensions  (H x W x D) Lid closed:
14.2 x 22.2 x 26.4 in.
(36 x 56.5 x 67 cm)
Lid opened:
30.3 x 22.2 x 26.4 in.
(77 x 56.5 x 67 cm)
Lid closed:
14.2 x 24.5 x 23.8 in.
(36 x 62.3 x 60.5 cm)
Lid opened:
30.3 x 29.3 x 26.4 in.
(77 x 74.5 x 67 cm)
Lid closed:
33.2 x 22.2 x 26.4 in.
(84 x 56.5 x 67 cm)
Lid opened:
49.2 x 22.2 x 26.4 in.
(125 x 56.5 x 67 cm)
Lid closed:
33.2 x 22.2 x 26.4 in.
(84 x 56.5 x 67 cm)
Lid opened:
49.2 x 22.2 x 26.4 in.
(125 x 56.5 x 67 cm)
Weight 189.6 lbs.
86kg
255.7 lbs.
116kg
314.2 lbs.
142.5kg
336.2 lbs.
152.5kg
Display type LCD LCD LCD LCD
Ventilated
Refrigerated
Floor standing (portable)
Floor standing (portable) refrigerated
  ventilated high performance benchtop centrifuge refrigerated high performance benchtop centrifuge portable high performance benchtop centrifuge floor standing high performance benchtop centrifuge
Request a quote
  Ventilated centrifuges are great for samples that are not temperature sensitive. Remember that ventilated centrifuges may not be at the laboratory's ambient room temperature, since spinning rotors create heat.
Any samples that can be negatively impacted by an increase in temperature would benefit. When spinning large capacity rotors at higher speeds, there is more heat; refrigeration can maintain optimal temperatures.
Along with our ventilated benchtop models, floor models offer an option to save bench space. Our high performance, refrigerated floor models offer optimal flexibility in both your experimental applications as well as your laboratory workflow saving precious bench space.
Max capacity   6 x 250mL (fixed angle)
4 x 1000mL (swinging bucket)
6 x 250mL (fixed angle)
4 x 1000mL (swinging bucket)
6 x 250mL (fixed angle)
4 x 1000mL (swinging bucket)
6 x 250mL (fixed angle)
4 x 1000mL (swinging bucket)
Max speed 15,200rpm (fixed angle)
6,300rpm (swinging bucket)
15,200rpm (fixed angle)
6,300rpm (swinging bucket)
15,200rpm (fixed angle)
6,300rpm (swinging bucket)
15,200rpm (fixed angle)
6,300rpm (swinging bucket)
Max RCF 25,830 x g (fixed angle)
7,164 x g (swinging bucket)
25,830 x g (fixed angle)
7,164 x g (swinging bucket)
25,830 x g (fixed angle)
7,164 x g (swinging bucket)
25,830 x g (fixed angle)
7,164 x g (swinging bucket)
Programs 99, 5 direct program buttons 99, 5 direct program buttons 99, 5 direct program buttons 99, 5 direct program buttons
Dimensions  (H x W x D) Lid closed:
14.2 x 22.2 x 26.4 in.
(36 x 56.5 x 67 cm)
Lid opened:
30.3 x 22.2 x 26.4 in.
(77 x 56.5 x 67 cm)
Lid closed:
14.2 x 24.5 x 23.8 in.
(36 x 62.3 x 60.5 cm)
Lid opened:
30.3 x 29.3 x 26.4 in.
(77 x 74.5 x 67 cm)
Lid closed:
33.2 x 22.2 x 26.4 in.
(84 x 56.5 x 67 cm)
Lid opened:
49.2 x 22.2 x 26.4 in.
(125 x 56.5 x 67 cm)
Lid closed:
33.2 x 22.2 x 26.4 in.
(84 x 56.5 x 67 cm)
Lid opened:
49.2 x 22.2 x 26.4 in.
(125 x 56.5 x 67 cm)
Weight 189.6 lbs.
86kg
255.7 lbs.
116kg
314.2 lbs.
142.5kg
336.2 lbs.
152.5kg
Display type LCD LCD LCD LCD
Ventilated
Refrigerated
Floor standing (portable)
Floor standing (portable) refrigerated
  ventilated high performance benchtop centrifuge refrigerated high performance benchtop centrifuge portable high performance benchtop centrifuge floor standing high performance benchtop centrifuge
Request a quote
  Ventilated centrifuges are great for samples that are not temperature sensitive. Remember that ventilated centrifuges may not be at the laboratory's ambient room temperature, since spinning rotors create heat.
Any samples that can be negatively impacted by an increase in temperature would benefit. When spinning large capacity rotors at higher speeds, there is more heat; refrigeration can maintain optimal temperatures.
Along with our ventilated benchtop models, floor models offer an option to save bench space. Our high performance, refrigerated floor models offer optimal flexibility in both your experimental applications as well as your laboratory workflow saving precious bench space.
Max capacity   6 x 250mL (fixed angle)
4 x 1000mL (swinging bucket)
6 x 250mL (fixed angle)
4 x 1000mL (swinging bucket)
6 x 250mL (fixed angle)
4 x 1000mL (swinging bucket)
6 x 250mL (fixed angle)
4 x 1000mL (swinging bucket)
Max speed 15,200rpm (fixed angle)
6,300rpm (swinging bucket)
15,200rpm (fixed angle)
6,300rpm (swinging bucket)
15,200rpm (fixed angle)
6,300rpm (swinging bucket)
15,200rpm (fixed angle)
6,300rpm (swinging bucket)
Max RCF 25,830 x g (fixed angle)
7,164 x g (swinging bucket)
25,830 x g (fixed angle)
7,164 x g (swinging bucket)
25,830 x g (fixed angle)
7,164 x g (swinging bucket)
25,830 x g (fixed angle)
7,164 x g (swinging bucket)
Programs 99, 5 direct program buttons 99, 5 direct program buttons 99, 5 direct program buttons 99, 5 direct program buttons
Dimensions  (H x W x D) Lid closed:
14.2 x 22.2 x 26.4 in.
(36 x 56.5 x 67 cm)
Lid opened:
30.3 x 22.2 x 26.4 in.
(77 x 56.5 x 67 cm)
Lid closed:
14.2 x 24.5 x 23.8 in.
(36 x 62.3 x 60.5 cm)
Lid opened:
30.3 x 29.3 x 26.4 in.
(77 x 74.5 x 67 cm)
Lid closed:
33.2 x 22.2 x 26.4 in.
(84 x 56.5 x 67 cm)
Lid opened:
49.2 x 22.2 x 26.4 in.
(125 x 56.5 x 67 cm)
Lid closed:
33.2 x 22.2 x 26.4 in.
(84 x 56.5 x 67 cm)
Lid opened:
49.2 x 22.2 x 26.4 in.
(125 x 56.5 x 67 cm)
Weight 189.6 lbs.
86kg
255.7 lbs.
116kg
314.2 lbs.
142.5kg
336.2 lbs.
152.5kg
Display type LCD LCD LCD LCD
*Biocontainment certification by Public Health England Porton Down, UK.
**Subject to Thermo Fisher Scientific’s standard limited warranty and country and dealer participation.
Top
Rotors for high performance benchtop centrifuges
Our extensive rotor portfolio provides application flexibility to support multiple lab settings, multiple users, and versatility as the needs of your laboratory evolve.
Swinging bucket rotors
Rotors for 1.6L capacity centrifuges
Rotors for 4L capacity centrifuges
Rotors for 1.6L and 4L capacity centrifuges
Fixed angle rotors
Rotors for 1.6L capacity centrifuges
Rotors for 4L capacity centrifuges
Rotors for 1.6L and 4L capacity centrifuges
Top
Fiberlite carbon fiber rotors brochure
Thermo Scientific Fiberlite rotors accelerate productivity while offering unequaled durability. They will maximize your centrifuge performance with versatility, speed and a robust corrosion-free design. Backed by a 15-year warranty, the most comprehensive warranty coverage available.*  
  • Improved ergonomics for added safety
  • Lightweight design for faster acceleration and deceleration
  • Corrosion resistance for easier care and maintenance
  • Fatigue-resistant design for longer life
Open brochure
* Subject to Thermo Fisher Scientific’s standard limited warranty. Ask your sales representative for details or review our general terms and conditions of sale on our Terms of Use page.
Top
Take a virtual tour of our benchtop centrifuges
Take a self-guided tour and learn how our centrifuges and rotors are designed around you and your applications.
They fit in to: They stand out for:
  • your space
  • your processes and protocols
  • the evolving needs of your
    future in the lab
  • innovative design and technologies
  • solid reliability
  • consistent results
While in the tour, take a look at our convenient packages to find just the right centrifuge, rotor, and adapter combination you need to get started.
Take a tour
Top
Benchtop centrifuges videos
Top
Related products for high performance benchtop centrifuges
Microcentrifuges
Floor models
Centrifuge rotors
Centrifuge ware
Resources
Support
  • Unity Lab Services
    Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.