Microcentrifuge rotor ClickSeal biocontainment lid
Speed and versatility for your microcentrifuge tube applications
Choose between our standard rotor (spinning 24 tubes per run) for basic microtube processing to mini-preps and spin-columns, and the flexibility of 6 additional rotor choices to accommodate 0.5mL to 5mL microtubes, PCR tubes and hematocrit capillaries. These high performance engineered polymer rotors are designed for outstanding chemical resistance to minimize corrosion from aggressive reagents.
Learn about how an engineered polymer rotor impacts the operation and maintenance of a microcentrifuge. View the Smart Note ›
Get more information
Thermo Scientific rotors for your microcentrifuge tubes
Rotor Rotor capacity Max tube dimensions Max speed (rpm)
& RCF (xg)
Catalog #
24 x 1.5/2.0mL rotor with ClickSeal biocontainment lid1
Standard rotor with exceptional versatility; 24 tubes per run from basic microtube processing to mini-preps and spin columns and additional flexibility with available adapters.
24 x 1.5/2.0
11 x 50 14,800 rpm
21,100 xg
75003424
0.5/0.6mL µL tube adapters (24) 24 x 0.5/0.6 7 x 32 14,800 rpm
21,100 xg
76003252
0.25/0.4mL µL tube adapters (24) 24 x 0.25/0.4 6 x 24 14,800 rpm
21,100 xg
76003251
0.2mL PCR tube adapters (24) 24 x 0.2 6 x 24
14,800 rpm
21,100 xg
76003250
Dual Row 18 x 2.0/0.5mL rotor with screw-on lid
Accommodate two different size tubes in one rotor without using adapters.
18 x 0.5/2.0 11 x 50
14,800 rpm
20,800 xg
75003418
36 x 0.5mL rotor with screw-on lid
Increase throughput for smaller microtubes.
36 x 0.5 7 x 32
14,800 rpm
19,300 xg
75003436
10 x 5mL rotor with ClickSeal biocontainment lid
Increase your nucleic acid sample separation.
10 x 5.0 17 x 62 14,800 rpm
20,326 xg
75003465
PCR 8 x 8 rotor with screw-on lid
Centrifuge up to 8 PCR strips at one time.
64 x 0.2 6 x 24 14,800 rpm
17,100 xg
75003489
PCR 4 x 8 rotor with ClickSeal biocontainment lid1
Process up to 4 PCR strips with certified1 sample protection.
32 x 0.2 6 x 24 14,800 rpm
16,200 xg
75003440
Hematocrit rotor
Achieve fast capillary separations with included linear scale to read results.
24 capillaries NA 14,800 rpm
16,800 xg
75003473
1Biocontainment certification by Public Health England, Porton Down, UK on all rotors with ClickSeal™ biocontainment lids, except for the 10 x 5mL rotor.
Featured microcentrifuge video
Microcentrifuges that fit in and stand out
Accelerate your micro-volume protocols with high speeds (up to 21,100 xg), rotor versatility and outstanding safety with certified ClickSeal biocontainment.
Related microcentrifuge rotor categories
Use our compact, safe and easy-to-use microcentrifuges to support your micro-volume protocols. They combine power with versatility and convenience, and can accelerate your sample preparation processes with RCF up to 17,000 xg
or 21,100 xg.
Microcentrifuge tubes
Choose from a broad range of microcentrifuge tubes, designed to meet the most rigorous requirements of centrifugation and daily laboratory use. They offer pure protection and security during sample preparation, transfer and storage.
Improve downstream results with optimized cell lysis and protein extraction reagents, organelle isolation and subcellular fractionation kits, protease and phosphatase inhibitors, dialysis devices, desalting columns, protein concentrators, and ion exchange and affinity purification resins and beads.
How does an engineered polymer rotor impact the operation and maintenance of a microcentrifuge?
Microcentrifuge rotor smart note
Microcentrifuges are critical to both life science research and clinical diagnostic applications, and the rotor selection can impact the safety and productivity of these high-use instruments. Lightweight and corrosion-resistant polypropylene rotors, available for Thermo Scientific microcentrifuges, are designed to enhance safety and productivity.
Open Smart Note
Resources
Support
Unity Lab Services 
Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.