microcentrifuges
Compact, easy-to-use microcentrifuges
Accelerate your micro-volume separations with our compact, safe, and easy-to-use microcentrifuges, offering power and versatility for routine laboratory work as well as high-end applications. Available in two speed options with both refrigerated and ventilated models, including ready-to-use configurations that can be ordered online today, find the reliability, flexibility and convenience you need to maximize your lab's productivity.
Browse our portfolio below or contact us and one of our experts will help you choose a mini or microcentrifuge best for your application.
Open brochure
Contact us
Quick links
Mini or micro?
Mini centrifuges
Microcentrifuges
mini centrifuge microcentrifuge
Increase efficiency and user comfort in your lab with mySPIN 12 and 6 mini centrifuges.
  • 2,000–9,800 xg max RCF
  • 6,300–12,500 xg max speed
  • Each instrument comes with 2 rotors (6 and 16 for mySPIN 6 and 12 and 32 for mySPIN 12)
  • 6–12 1.5/2.0mL tubes with available 0.2/0.5mL adapters
Choose a mini centrifuge
Perform complex or simple protocols with microcentrifuges and an extensive selection of optional rotors.
  • 17,000–21,100 xg  max RCF
  • 13,300–14,800 xg  max speed
  • 7 available rotors for 0.2–5mL tube volumes, PCR tube strips, and capillary tubes
Choose a microcentrifuge
Which microcentrifuge is right for you?
Find the ideal blend of capabilities to support your micro-volume protocols—from basic microtube processing to nucleic acid mini-preps and spin-columns to hematocrit capillaries—in a small footprint with:
  • Intuitive controls, easy-to-read displays, and fast one-click centrifuge lid closure
  • Fast acceleration and deceleration, and outstanding corrosion resistance with lightweight engineered polymer rotors
  • Wide variety of rotors support 0.2, 0.5, 1.5, 2.0mL microtubes, PCR strip tubes, 5mL tubes, and hematocrit
    capillary tubes
  • Added sample security with Thermo Scientific™ ClickSeal™ biocontainment lids (selected rotors only)
Thermo Scientific 21,100 x g microcentrifuges are ideal for high-end research applications, reducing spin times by up to 20% when compared with the 17,000 x g models. Available in both ventilated and refrigerated models plus a choice of 7 rotors to find the ideal fit for your application.
Ventilated
Refrigerated
  microcentrifuge microcentrifuge
  21 21R
Max RCF 21,100 x g 21,100 x g
Max speed 14,800 rpm 14,800 rpm
Included rotor 24 x 1.5/2.0mL rotor (additional rotors available)
Max noise level 56 dBA 50 dBA
Time set range 1 min to 99 min; 1 min increments + HOLD mode
Temperature range Ambient Set from –9°C to +40°C in 1°C increments
Certifications            • CSA certified
                • CE marked        
     • UL listed
             • IVD compliant
Technical standards IEC 61010-1, IEC 61010-2-020 (refer to manual for more details)
Dimensions
(H x W x D)
8.9 x 9.6 x 13.8in. (22.5 x 24.3 x 35.2cm) 12.9 x 11.6 x 17.5in. (33.0 x 29.5 x 44.5cm)
Product name
Thermo Scientific 17,000 x g microcentrifuges are excellent for routine laboratory work, and both ventilated and refrigerated models are available, in addition to a choice of 7 rotors to find the ideal fit for your application.
Ventilated
Refrigerated
  microcentrifuge microcentrifuge
  17 17R
Max RCF 17,000 x g 17,000 x g
Max speed 13,300 rpm 13,300 rpm
Included rotor 24 x 1.5/2.0mL rotor (additional rotors available)
Max noise level 56 dBA 50 dBA
Time set range 1 min to 99 min; 1 min increments + HOLD mode
Temperature range Ambient Set from –9°C to +40°C in 1°C increments
Certifications            • CSA certified
                • CE marked        
             • UL listed        
             • IVD Compliant
Technical standards IEC 61010-1, IEC 61010-2-020 (refer to manual for more details)
Dimensions
(H x W x D)
8.9 x 9.6 x 13.8in. (22.5 x 24.3 x 35.2cm) 12.9 x 11.6 x 17.5in. (33.0 x 29.5 x 44.5cm)
Product name
Thermo Scientific 21,100 x g microcentrifuges are ideal for high-end research applications, reducing spin times by up to 20% when compared with the 17,000 x g models. Available in both ventilated and refrigerated models plus a choice of 7 rotors to find the ideal fit for your application.
Ventilated
Refrigerated
  microcentrifuge microcentrifuge
  21 21R
Max RCF 21,100 x g 21,100 x g
Max speed 14,800 rpm 14,800 rpm
Included rotor 24 x 1.5/2.0mL rotor (additional rotors available)
Max noise level 56 dBA 50 dBA
Time set range 1 min to 99 min; 1 min increments + HOLD mode
Temperature range Ambient Set from –9°C to +40°C in 1°C increments
Certifications            • CSA certified
                • CE marked        
     • UL listed
             • IVD compliant
Technical standards IEC 61010-1, IEC 61010-2-020 (refer to manual for more details)
Dimensions
(H x W x D)
8.9 x 9.6 x 13.8in. (22.5 x 24.3 x 35.2cm) 12.9 x 11.6 x 17.5in. (33.0 x 29.5 x 44.5cm)
Product name
Thermo Scientific 17,000 x g microcentrifuges are excellent for routine laboratory work, and both ventilated and refrigerated models are available, in addition to a choice of 7 rotors to find the ideal fit for your application.
Ventilated
Refrigerated
  microcentrifuge microcentrifuge
  17 17R
Max RCF 17,000 x g 17,000 x g
Max speed 13,300 rpm 13,300 rpm
Included rotor 24 x 1.5/2.0mL rotor (additional rotors available)
Max noise level 56 dBA 50 dBA
Time set range 1 min to 99 min; 1 min increments + HOLD mode
Temperature range Ambient Set from –9°C to +40°C in 1°C increments
Certifications            • CSA certified
                • CE marked        
             • UL listed        
             • IVD Compliant
Technical standards IEC 61010-1, IEC 61010-2-020 (refer to manual for more details)
Dimensions
(H x W x D)
8.9 x 9.6 x 13.8in. (22.5 x 24.3 x 35.2cm) 12.9 x 11.6 x 17.5in. (33.0 x 29.5 x 44.5cm)
Product name
Featured microcentrifuge video
Microcentrifuges that fit in and stand out
Accelerate your micro-volume protocols with high speeds (up to 21,100 xg), rotor versatility and outstanding safety with certified ClickSeal biocontainment.
Microcentrifuge Rotors
Benchtop Centrifuges
Centrifuge ware
Protein Purification and Isolation
Resources
Support
  • Unity Lab Services 
    Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.