MicroPlateReadersandWasher
Expand functionality beyond the standard unit
Our reliable and easy to use Thermo Scientific™ Wellwash™ microplate strip washers provide secure washing performance for routine and research ELISA applications. Wellwash washers feature a large color display with graphical user interface, local language versions, and non-pressurized bottles for optimal ease-of-use, convenience, and safety.
Request a quote ›
Which Wellwash product is best for your needs?
  photo of multidrop combi
Wellwash
photo of multidrop combi nl
Wellwash Versa
Wash head 1x8, 1x12 two 1x8, two 1x12, 2x8 cell wash head
Wash bottles 1 bottle (2 liter) 3 bottles (two 4 liter and one 2 liter)
Plate formats 96 96, 384
Cell washing No Yes
Robot compatibility No Yes