ART pipette tips
Tips that fit any pipette in your lab
Thermo Scientific™ ART™ tips offer a complete selection of barrier, non-filter and specialty tips so it’s easier than ever to find a tip to fit virtually any pipette or application in your lab.
View good laboratory practices ›Take the ART challenge ›
Select the ART tips that are right for you
ART Barrier Tips
Prevent aerosol contamination with self-sealing barrier tips.
ART Barrier Tips
ART Non-Filter Tips
Proven ART quality tips ranging from 10µL to 10mL.
ART Non-Filter Tips
ART Specialty Tips
Available in wide bore, gel loading and extended length tips.
ART Specialty Tips
Need help finding a specific tip or packaging type? Contact a representative ›
ART barrier vs. traditional filter tips
ART barrier tips differ from traditional filter tips. ART self-sealing barrier tips are the only tips that seal completely to prevent sample from touching the pipette tip cone. When the barrier seals it serves as a warning to the user of potential pipette contamination.
Researchers have made ART self-sealing barrier tips their first choice in sample protection. ART barrier tips provide 100% security against aerosol and liquid contamination.
The ART Barrier prevents cross contamination by blocking the passage of aerosols, liquids, radioactive isotopes, and biologic materials to the pipette or to subsequent samples in an experiment.
Test them for yourself and take the ART challenge.
See it to believe it
Maximize sample recovery
Standard vs ART tips comparison
The recovery
Low retention pipette tips enhances sample recovery significantly compared to standard pipette tips. Inert hydrophobic polymer technology reduces sample loss due to adhesion within the tip.
The result
  • Less liquid retention to improve liquid handling
  • Improve volume accuracy to increase reliability
  • Precise sample delivery to save expensive reagents
Test them for yourself and take the ART challenge.
More, reliable tips that produce less waste and save space
Its an original. The ART Barrier and Non-Filtered Reload System provides an environmentally sustainable solution while maintaining stringent quality and sterility standards. With offset wells that allow the tips to stack, this advanced system reduces waste by up to 54%, compared to hinge racks and takes up to 55% less bench top and storage space.
Discover greener alternatives with Art Reload Pipette Tip Towers!
Learn more ›
ART Gel Loading Tips
Gel loading tips
Microtip design fits between the glass plates that are 1 or 1.5mm thick to access wells, allowing easy loading of wells. plus the added security of the ART barrier protects your pipette from contamination.
Volume
Compatibility
20µL
Universal
20µL Gilson Pipetman
Related products
Worry-free pipetting is here with the only tip that clips onto your pipette for a complete seal on every channel. Select Thermo Scientific™ F1-ClipTip™ Manual or Thermo Scientific™ E1-ClipTip™ Electronic and Equalizer pipettes, which feature adjustable tip spacing models.
Delivering years of unmatched productivity and ergonomics across thousands of labs and applications.
Electronic and manual multichannel pipettes with optional adjustable tip spacing and Thermo Scientific™
ClipTip™ technology.
Designed, developed and manufactured to perfectly fit Thermo Scientific pipettes. A pipette’s performance is directly related to the quality of the pipette tip for optimal liquid
handling results.