Limited-time savings* that puts money back into your research
  AmpliTaq Gold® 360 AccuPrime™ Pfx
Amplifies wide target range XXXX XXXX
Amplicon size <5 kb <15 kb
Fidelity vs. Taq 1x 26x
Yield XXXX XXXX
Step 1: Choose a PCR DNA polymerase
Product Size Cat. No.
AmpliTaq Gold® 360 Master Mix
1 mL 4398876
5 mL 4398881
50 mL 4398886
10 x 5 mL 4398901
AccuPrime™ Pfx SuperMix
200 reactions 12344040
Step 2: Choose any plastics product
Step 3: Enter promotion code XXXX to save 40% when you buy both
*This promotion is open to customers in the US (excluding Puerto Rico) and Canada who purchase either AmpliTaq Gold® Master Mix and any plastics product or AccuPrime™ Pfx SuperMix and any plastics product in a single purchase order.  Discount will apply to qualifying orders received by Life Technologies no later than June 30, 2014, or until promotional supplies are depleted, whichever comes first. Customer can use the discount only once. Discount applies to the list price in effect at the time the order is received by Life Technologies. Cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
In your research, you don’t have time to rescue failed PCR strategies. Save time and effort we can't know what results they need with Life Technologies™ PCR master mixes and supermixes. The benefits are clear.
Hot-start PCR for higher specificity and yield
When choosing a polymerase for PCR, we recommend starting with AmpliTaq Gold® 360 or AccuPrime™ Pfx DNA polymerase. AmpliTaq Gold® 360 polymerase can be used on a wide range of templates (e.g., routine, AT- and GC-rich) (Figure 1), and AccuPrime™ Pfx polymerase provides the benefit of maximum fidelity and specificity (Figure 2). Both offer robust and reliable amplification and are available in easy-to-use, premixed master mix formats.
Figure 1. AmpliTaq Gold® 360 Master Mix amplifies a broader range of targets. Panel A shows products amplified with AmpliTaq Gold® 360 Master Mix, while panel B shows the same products amplified using the Competitor X master mix. PCR was performed using 1 ng of template DNA per reaction, using cycling conditions according to each manufacturer’s recommendations. All enzyme concentrations were standardized at 0.025 U/µL. Annealing temperatures were uniform across the selection targets. Amplicons ranged from 300 to 1,400 bp in length, with an average length of 553 bp. Each reaction was performed in duplicate. Amplicons are labeled as follows: A = high AT; E = easy amplification; D = primer-dimer; H = homopolymer; SQ = sequencing challenge; L = long; G = high GC. The high-GC amplicon (G) reactions included 5 µL of 360 GC Enhancer.
Figure 2. AccuPrime™ Pfx DNA Polymerase delivers higher fidelity and specificity than other proofreading enzymes. AccuPrime™ Pfx DNA Polymerase was tested alongside three other proofreading enzymes. Each reaction was prepared according to the enzyme manufacturer’s recommendations in 50 μL reaction volumes containing 1X buffer. 20% of each reaction was analyzed on a 0.8% agarose gel. 1 = c-myc (822 bp); 2 = p53 (2,380 bp); 3 = Hbg (3.6 kb); 4 = rhod (6.1 kb); 5 = rhod (6.8 kb).
Automatic hot-start PCR for room temperature setup and master mix convenience
No need for on-ice setup or the addition of multiple components: both AmpliTaq Gold® 360 and AccuPrime™ Pfx provide automatic hot-start PCR, which allows you to set up reactions at room temperature without the problem of extension at misprimed sites. With the master mix and supermix formats, the addition of primers and template is all that is required for robust amplification.
Hot-start PCR for higher specificity and yield
When choosing a polymerase for PCR, we recommend starting with AmpliTaq Gold® 360 or AccuPrime™ Pfx DNA polymerase. AmpliTaq Gold® 360 polymerase can be used on a wide range of templates (e.g., routine, AT- and GC-rich) (Figure 1), and AccuPrime™ Pfx polymerase provides the benefit of maximum fidelity and specificity (Figure 2). Both offer robust and reliable amplification and are available in easy-to-use, premixed master mix formats.
Figure 1. AmpliTaq Gold® 360 Master Mix amplifies a broader range of targets. Panel A shows products amplified with AmpliTaq Gold® 360 Master Mix, while panel B shows the same products amplified using the Competitor X master mix. PCR was performed using 1 ng of template DNA per reaction, using cycling conditions according to each manufacturer’s recommendations. All enzyme concentrations were standardized at 0.025 U/µL. Annealing temperatures were uniform across the selection targets. Amplicons ranged from 300 to 1,400 bp in length, with an average length of 553 bp. Each reaction was performed in duplicate. Amplicons are labeled as follows: A = high AT; E = easy amplification; D = primer-dimer; H = homopolymer; SQ = sequencing challenge; L = long; G = high GC. The high-GC amplicon (G) reactions included 5 µL of 360 GC Enhancer.
Figure 2. AccuPrime™ Pfx DNA Polymerase delivers higher fidelity and specificity than other proofreading enzymes. AccuPrime™ Pfx DNA Polymerase was tested alongside three other proofreading enzymes. Each reaction was prepared according to the enzyme manufacturer’s recommendations in 50 μL reaction volumes containing 1X buffer. 20% of each reaction was analyzed on a 0.8% agarose gel. 1 = c-myc (822 bp); 2 = p53 (2,380 bp); 3 = Hbg (3.6 kb); 4 = rhod (6.1 kb); 5 = rhod (6.8 kb).
Automatic hot-start PCR for room temperature setup and master mix convenience
No need for on-ice setup or the addition of multiple components: both AmpliTaq Gold® 360 and AccuPrime™ Pfx provide automatic hot-start PCR, which allows you to set up reactions at room temperature without the problem of extension at misprimed sites. With the master mix and supermix formats, the addition of primers and template is all that is required for robust amplification.