PCR simplified with universal annealing
Invitrogen Platinum II Taq Hot-Start DNA Polymerase is designed to get you to your research destination, faster. A universal primer annealing feature reduces optimization steps and allows for co-cycling of all assays. A unique combination of innovative buffer, high-performance Taq DNA polymerase, and superior hot-start technology enables exceptional PCR results, even in the toughest applications.
Highlights
  • Innovative universal primer annealing buffer—reduces tedious optimization steps and helps save time by enabling co-cycling of all assays
  • Engineered Taq DNA polymerase—allows fast cycling and successful amplification even in the presence of inhibitors
  • Platinum hot-start technology—offers superior specificity, sensitivity, and yields; allows for room temperature reaction setup
  • Optional green buffer—helps reduce pipetting errors with direct gel loading
On this page
Engineered for better PCR
Click the buttons below and discover the benefits of Platinum II Taq Hot-Start DNA Polymerase.
Universal annealing
Fast and robust
interactive pcr
Superior specificity
Direct gel loading
Platinum II Taq Hot-Start DNA Polymerase advantages
PCR assays using conventional PCR reagents require specific protocols for amplification of each DNA fragment due to varying primer annealing temperatures and different durations of the extension step. Therefore, individual assays cannot be amplified in the same PCR run. With Platinum II Taq Hot-Start DNA Polymerase, different PCR assays can be cycled in parallel using the same protocol with universal primer annealing temperature and the extension step selected for the longest fragment to be amplified. Moreover, Platinum II Taq Hot-Start DNA Polymerase is a “fast” DNA polymerase; thus, the combination of this next generation DNA polymerase and the universal protocol permits fast cycling of all assays in as little as 30 minutes.
Platinum2TaqHot-StartDNA-Polymerase-cycle-together
Figure 1. Time saving enabled by assay co-cycling. PCR assays using conventional PCR reagents require specific protocols for amplification of each DNA fragment because of the different primer annealing temperatures and extension steps. Therefore, with traditional PCR reagents, multiple targets often cannot be amplified together in the same PCR run. With Platinum II Taq Hot-Start DNA Polymerase, different PCR assays can be cycled together using one protocol with a universal primer annealing temperature and the extension step selected for the longest fragment to be amplified. Moreover, Platinum II Taq Hot-Start DNA Polymerase is a fast DNA polymerase, delivering PCR results in as little as 30 minutes.
Time saving enabled by assay co-cycling
Figure 2. Platinum II Taq Hot-Start DNA Polymerase enables cycling of shorter and longer amplicons together. 132 bp, 251 bp, 1,005 bp, and 3.9 kb fragments were amplified from 50 ng of human genomic DNA in 50 μL reactions using Platinum II Taq Hot-Start DNA Polymerase or other hot-start DNA polymerases: (A) NEB OneTaq Hot Start DNA Polymerase, (B) Qiagen Fast Cycling PCR Kit, (C) Roche FastStart Taq DNA Polymerase. The same protocol was used for all four targets with the annealing and extension settings indicated. The size marker is Thermo Scientific ZipRuler Express DNA Ladder 2.
Platinum II Taq Hot-Start DNA Polymerase is an engineered enzyme with increased DNA synthesis rate. Therefore, with Platinum II Taq Hot-Start DNA Polymerase PCR results are generally more than 2 times faster than other hot-start Taq DNA polymerases.
Fast cycling reduces PCR run time
Figure 3. Fast cycling reduces PCR run time. Amplification of a 529 bp fragment from 50 ng of human genomic DNA in 50 μL reactions for 35 cycles was carried out using Platinum II Taq Hot-Start DNA Polymerase and hot-start DNA polymerases from other suppliers: (A) Sigma-Aldrich KAPA2G Fast HotStart PCR Kit, (B) NEB OneTaq Hot Start DNA Polymerase, (C) Promega GoTaq G2 DNA Polymerase, (D) Toyobo Quick Taq HS DyeMix, (E) Roche FastStart Taq DNA Polymerase, and (F) Sigma-Aldrich JumpStart Taq DNA Polymerase. Cycling times for each polymerase are shown in purple, while ramping times on the ProFlex PCR System (6°C/sec peak block ramp rate) are shown in red. PCR product analysis in 1% TAE agarose gels is presented below the graph. The size marker is the ZipRuler Express DNA Ladder 2.
The high sensitivity of Platinum II Taq Hot-Start DNA Polymerase enables successful amplification of specific product in experiments where there is a limited amount of starting material or the target DNA is in low concentration in the sample. 
High sensitivity and reliable amplification from low amounts of input DNA
Figure 4. High sensitivity and reliable amplification from low amounts of input DNA. Amplification of a 529 bp fragment from 0 (no template control); 0.016; 0.08; 0.4; 2; 10; 50; 250 ng of human genomic DNA were amplified in 50 μL PCR reactions using Platinum II Taq Hot-Start DNA Polymerase or competitor DNA polymerases (A—KAPA2G Fast HotStart, B—NEB OneTaq Hot Start, C—Promega GoTaq G2, D—Sigma JumpStart Taq, and E—Takara Taq HS Perfect Mix). The estimated copy number is ~5 copies per 0.016 ng of human genomic DNA. The molecular weight marker is ZipRuler Express DNA Ladder 2.
Platinum II Taq Hot-Start DNA Polymerase was engineered for resistance to inhibitors and helps enable successful amplification with samples of suboptimal purity.
Resistance to inhibitors
Figure 5. Resistance to inhibitors. Amplification of a 1 kb fragment from human genomic DNA using Platinum II Taq Hot-Start DNA Polymerase or competitor DNA polymerases (A—KAPA 2G Robust HotStart, B—NEB OneTaq Hot Start, C—Promega GoTaq G2, and D—Takara Taq Hot Start Version) in reaction mixtures containing: 1—humic acid (up to final concentration of 1.3 µg/mL), 2—hemin (up to final concentration of 6 µM), 3—xylan (up to final concentration of 0.26 mg/mL), or 4—no inhibitor control. The molecular weight marker is ZipRuler Express DNA Ladder 2.
Amplification of DNA extracted from FFPE tissue samples
Figure 6. Amplification of DNA extracted from FFPE tissue samples. Amplification of a 527 bp fragment from varying amounts of DNA extracted from mouse FFPE tissue samples using Platinum II Taq Hot-Start DNA polymerase. RecoverAll Total Nucleic Acid Isolation Kit for FFPE was used for DNA extraction. NTC: no template control. PC: positive control from 1 ng of purified mouse genomic DNA. The molecular weight marker is ZipRuler Express DNA Ladder 2.
The formulation of Platinum II Taq Hot-Start DNA Polymerase and 2X Master Mixes allows for amplification of versatile range of targets, from AT-rich to GC-rich. A separate vial of Platinum GC Enhancer is provided for specific amplification and improved yields of targets with high-GC content.
Robust amplification of AT-rich and GC-rich targets
Figure 7. Robust amplification of AT-rich and GC-rich targets. Various DNA fragments of increasing GC content (indicated above the corresponding lanes) were amplified from 100 ng of human genomic DNA in 50 µL PCR reactions. Platinum GC Enhancer was used for targets with >65% GC. The molecular weight marker is ZipRuler Express DNA Ladder 2.
What is the difference between Platinum II Taq Hot-Start DNA Polymerase and Platinum Taq DNA Polymerase?
Platinum Taq DNA Polymerase has been trusted by researchers for over two decades and has been used in several thousand publications. Platinum II Taq Hot-Start DNA Polymerase is the next generation hot-start DNA polymerase, newly engineered for rapid, robust performance. We recommend that investigators starting new projects use Platinum II Taq DNA Polymerase so that they can benefit from its superior performance, summarized in the table below.
Comparison of technical specifications between Platinum II Taq Hot-Start and Platinum Taq DNA Polymerases
  Platinum II Taq Hot-Start DNA Polymerase Platinum Taq DNA Polymerase
Technical specifications
Universal annealing protocol Yes No
Speed 15 sec/kb 1 min/kb
Flexible extension stepa Yes No
Inhibitor resistance Yes No
Target length Up to 5 kb Up to 5 kb
Hot-start modification Antibody-mediated Antibody-mediated
Fidelity versus Taq DNA Polymerase 1x 1x
Blunt or 3’A end 3’A 3’A
Benchtop stability of assembled PCR reactions 24 h 24 h
GC-rich templates Yes Yes
Controlled low level of residual human and bacterial DNAd Yes No
Formats
Master mix Colorless/greenb Colorless/greenb
Stand-alone enzyme Colorless/greenc Colorless/greenb
aThe extension step can be extended up to 60 sec/kb without the effect on specificity bDirect gel loading with green buffer options. cGreen buffer available as separate item for use with stand-alone enzyme dDuring manufacturing of Platinum II Taq Hot- Start DNA Polymerase, strict measures are taken to control and verify that no more than one copy of residual bacterial genomic DNA is present per unit of the polymerase.
Ordering information
New! Platinum II Taq Hot-Start DNA Polymerase
Resources
Product manuals
Application notes
PCR education
Learn about the history of PCR, setup considerations, importance of DNA polymerase choice, common methods and applications, and troubleshooting.
Molecular Biology Webinar Series
View our webinar series to learn more about molecular biology topics such as reverse transcription, PCR and cloning.
Support
PCR and Synthesis Support Center
Find tips, troubleshooting help, and resources for your end-point PCR and cDNA applications.
Product Support Documentation
Search for manuals, protocols, Material Safety Data Sheets, product literature and certificates by catalog number or product name.
Contact Us
Email or call our Technical Application Scientists for additional questions regarding PCR enzymes and master mixes.