Life Technologies’ has been providing instruments, reagents, and solutions for plant-science research for over two decades.
Fragment Analysis Solutions for Agriculture Biotechnology
Life Technologies has developed CE solutions for high-throughput genotyping, genomic profiling, chimerism, and copy number analysis to enable sequence-to-function analysis for today’s plant biotechnology researchers.
View products
Sequencing Solutions for Plant Sciences
Life Technologies sequencing solutions enable key applications in plant research such as de novo sequencing, genotyping and gene expression analysis.
View all
AFLP®
Amplified fragment length polymorphism (AFLP®) is a technique originally developed for plant-breeding applications, but is now employed universally as a method for fingerprinting DNA from any source. Read the AFLP® application note on plant mapping.
View products
Plant Nucleic Acid Purification
Life Technologies offers an expansive portfolio of kits, reagents, and devices for the isolation, purification, analysis, and quantitation of plant genomic DNA, total RNA, mRNA, and microRNA or transcriptome RNA.
View products
Custom Gene Expression Assays
TaqMan® Gene Expression Assays
We offer hundreds of inventoried, made-to-order, plant-specific TaqMan® Gene Expression Assays that can be customized to your specifications.
MicroRNA and non-coding RNA TaqMan® Assays
Inventoried TaqMan® MicroRNA Assays are available for several species, including Arabidopsis. These assays are aligned with the Sanger miRBase Registry. If you can’t find the assay you need from our stocked list, you can design your own Assay.
Genotyping
TaqMan® genotyping assays are optimized for end-point fluorescence detection and are useful in a variety of applications in agriculture and plant biotechnology, including linkage mapping and population genetics. For higher throughput, the TaqMan® SNP genotyping assays (stocked or custom) can be supplied as an Open Array® Genotyping plate, which enables one person to validate or screen up to 256 SNPs across >1,500 samples in one day, without the use of robotics.
Order SNP genotyping assays now
Plant Transformation
ElectroMAX™ LBA4404 Cells have been successfully used to transform a variety of plant species in plant biotechnology research, including tobacco and Arabidopsis, at high efficiency.
Algae Engineering Kits
New GeneArt® Algae Engineering Kits for Chlamydomonas reinhardtii and Synechococcus elongatus are the first commercially available genetic modification and expression systems for photosynthetic microalgae. These kits are designed for rapid scale-up and production and consistent, defined quality.
Learn more
GMO Testing
Our TaqMan® GMO Detection Kits enable you to detect the genetically modified organism (GMO) in seed, grain, and processed foods, and their ingredients.
Application note:Isolation, Detection, and Quantitation of Genetically Modified DNA in Food Materials
Browse Selected Products for Plant Research Workflows