proquantum-270x195
A powerful innovation in protein quantitation
Invitrogen ProQuantum immunoassays provides researchers with an easy-to-run, high-performance, target-specific protein detection assay with no proprietary instrument to purchase. Utilizing proximity ligation assay (PLA®) technology, ProQuantum offers the analyte specificity of antibody–antigen binding with the signal detection and amplification capabilities of qPCR to achieve a highly sensitive assay. ProQuantum can also detect lower levels of protein and uses lower sample consumption than traditional methods.
To hear the technical details from the R&D expert, watch the on-demand webinar or to see how easy the assay is to run, click on one of the red buttons below:
Watch webinar
To learn more about the technology and benefits of ProQuantum high-sensitivity immunoassays and how they can help you in your research, download the ProQuantum high-sensitivity immunoassays brochure by clicking the button below:
Download brochure
Learn more here or email us at invitrogenimmunoassays@thermofisher.com and we will connect you with a representative who can help.
Ordering ProQuantum immunoassays
NEW protein targets coming soon:
  • Human G-CSF
  • Mouse IFNγ
  • Human IL-4
  • Human IL-22
  • Human MIP-1β
  • Human VEGF
Available protein targets include:
ProQuantum immunoassays benefits at-a-glance:
  • High-sensitivity – detect low levels of proteins with greater sensitivity than traditional methods (Figure 1)
  • Broad dynamic range – ≥5 logarithmic units, minimizing sample dilutions to ensure they fall within the range
  • Small sample consumption – use 2–5 μL of sample (instead of 75 μL for triplicate wells with other methods)
  • Fast, easy workflow – typically 2 hours from sample to answer, no wash steps, and a single 1-hour incubation
  • No proprietary instrument to purchase – runs on any real-time PCR instrument
  • Includes intuitive cloud-based software – comprehensive data analysis and statistical group–wise comparison
Here is how it works
ProQuantum immunoassays utilize a matched pair of target-specific antibodies each conjugated to a DNA oligonucleotide. This platform leverages the sensitivity and large dynamic range of Applied Biosystems TaqMan real-time PCR technology. During antibody–analyte binding, the two DNA oligos are brought into close proximity, which then allows for ligation of the two strands and subsequent creation of template strand for amplification (Figure 2).
ProQuantum High-Sensitivity Human Assay standard curves
Figure 1. Standard curve for Invitrogen ProQuantum High-Sensitivity Human IFNγ, IL-1β, IL-8, and IL-6 immunoassay kits (red) shows larger dynamic range and improved separation at the low end of the curve for greater sensitivity than normal and ultrasensitive ELISAs (blue).
PLA by ProQuantum immunoassay
Figure 2. ProQuantum immunoassay schematic. (A) Antibody-conjugates binding to target (1-hr incubation). (B) Ligation and amplification of signal (in qPCR instrument).
Robust and intuitive Cloud-based analysis software
Use the software (Figure 3) to access customized plate and assay setup instructions as well as obtain results including robust statistical group–wise comparisons.
ProQuantum software
Figure 3. Intuitive software for easy and comprehensive data analysis.
Greener product and packaging
Thermo Fisher Scientific is committed to designing our products with the environment in mind—it’s part of how we enable our customers to make the world healthier, cleaner, and safer. This fact sheet provides the rationale behind the environmental claim that Invitrogen ProQuantum Immunoassay Kits generate less waste and have more sustainable packaging than traditional ELISA kits. To learn more download the green features fact sheet here.
q315-social-media-email
New target-specific assays are being developed on a monthly basis
Stay in the loop and sign up to receive our new product release announcements.
Sign up
Let us know what future targets you would like to see
We are continuously developing new ProQuantum immunoassays. You can contribute to our product development roadmap. Email us at invitrogenimmunoassays@thermofisher.com and let us know what protein targets you would like to see next.
Explore the entire immunoassay portfolio: thermofisher.com/immunoassays