NovaBright™ Chemiluminescent Beta-gal & Luciferase Reporter Gene Assays
NovaBright beta-galactosidase and firefly luciferase dual enzyme reporter gene chemiluminescent detection kit provides rapid and sensitive sequential detection of both reporter gene enzymes in the same sample. The use of two reporter genes, one as an experimental reporter (luciferase) and the other as a constitutively expressed transfection control reporter (b-galactosidase) is often necessary to accurately quantitate activity from experimental reporter constructs.
NovaBright β-galactosidase and Firefly Luciferase Dual Enzyme Reporter Gene Assays
NovaBright™ β-galactosidase and firefly luciferase dual enzyme reporter gene assays enable greater precision in normalization of transfection efficiency by using the same sample in a single experiment with a reference gene of your choice. Readout for both reporter genes is performed sequentially in less than 1 hour of assay time.
NovaBright™ β-galactosidase and Firefly Luciferase Dual Enzyme Reporter Gene Assays
Detection of Firefly Luciferase and ß-Galactosidase with NovaBright Dual Enzyme Reporter Gene System.
  • Normalization in the same sample for accurate quantitation of expression
  • Convenient dual detection by reading flash and glow luminescence
  • Sensitive over 7 orders of magnitude
NovaBright Beta-gal & Luciferase Workflow
NovaBright™ Beta-gal & Luciferase
More NovaBright Chemiluminescent Reporter Gene Assays
Resources
Learning
Support