We offer a variety of specialty reagents for western blotting to:
  • Enhance signal sensitivity and intensity
  • Enable detection with much less primary antibody
  • Selectively detect target proteins and avoid interference from denatured IP antibody fragments in western blots of immunoprecipitated proteins 
Popular products
This kit contains a membrane treatment reagent and a primary antibody diluent that increase both signal intensity and sensitivity 3- to 10-fold compared to detection performed without it.
This is a post-transfer nitrocellulose membrane-treatment reagent that enables western blot detection with much less primary antibody. By treating nitrocellulose membranes for a few minutes in this reagent immediately after transfer and before probing, you can use much less (more dilute) primary antibody solutions while obtaining the same level of detection
A horseradish peroxidase conjugate that is optimized for post-immunoprecipitation western blot detection of primary antibodies without interference from denatured IP antibody fragments and allows enhanced western blot detection of target proteins following IP assays (immunoprecipitation).
Resources
Learning resources
Support resources