DNA Sequencing
DNA Sequencing is the process of reading nucleotide bases in a DNA molecule. Unlock the genome and answer biology’s most challenging questions with our innovative and accessible sequencing solutions.
For over 25 years, our sequencers have contributed to significant scientific breakthroughs, including sequencing of the first human genome and the discovery of genes implicated in diseases like cystic fibrosis. Our ongoing innovation in sequencing technologies continues to drive new discoveries.
From whole genome sequencing to targeted sequencing of specific genomic regions, our sequencing portfolio supports a wide range of throughput and research application needs for DNA sequencing.
Which platform is right for you?
Featured sequencing products
Ion Torrent Next-Generation Sequencing
Sanger Sequencing
Sequencing Kits and Reagents
Sequencing individual genes, gene regions, or sets of genes is a common targeted sequencing approach used in cancer and inherited disease research to screen known or discover novel germline and/or somatic mutations in focused areas of the genome.
"From the beginning" or de novo sequencing is the method of building a reference genome. This is achieved by randomly fragmenting the target DNA, sequencing then assembling those individual fragments to build a draft or even finished genome.
Whole Genome Sequencing (WGS) entails sequencing the entire genome and comparing that to a reference genome in order to detect the full range of genetic variation such as SNPs, insertions, deletions, inversions, complex rearrangements, and copy number variation.
Accurate variant confirmation using orthogonal technology (independent chemistry) follows discovery of novel variants. We offer the gold standard and a fast approach for labs to validate their discoveries.
Targeted sequencing of the exome employs enrichment strategies that target coding exons. The exome encompasses approximately 1% of the genome, yet contains approximately 85% of disease-causing mutations. For genetic researchers trying to unravel the causes of over 6,800 rare diseases [1], exome sequencing enables the identification of single-nucleotide variants (SNVs), small insertions or deletions (indels), and rare de novo mutations that explain the heritability of complex diseases [2].
Sequencing the 16S rRNA gene is an accurate and reliable method for microbial identification.
References
  1. National Institutes of Health, Office of Rare Diseases Research.
  2. Marth GT, Yu F, Indap AR, et al. (2011) The functional spectrum of low-frequency coding variation. Genome Biol 12(9):R84.
One tube for SNPs, indels, and CNVs - Ion AmpliSeq targeted sequencing panels