The Chip is the Machine™
Ion semiconductor sequencing making high-quality next-generation sequencing accessible for all.
Have a sales rep contact me >
Ion PGM™ System Product Selection Guide
The Chip is the Machine™ and Ion Torrent™ directly benefits from four decades of exponential improvement in semiconductor technology. This enables our chips to scale in density for any research application—from small, targeted experiments to large genomes—all on a single platform. See how Ion Torrent Chips work.
With as little as 10 ng input DNA, the new Ion 314™, 316™, and 318™ Chips yield 99.99% consensus accuracy and up to a 50% increase in output over previous chips, in as little as 2 hours on the Ion PGM™ System.
For whole genome sequencing on the Ion Proton™ System, the Ion PI™ Chip enables the highest throughput with up to 10 Gb reads in 2–4 hours.
This unique combination of simplicity and easy scalability makes semiconductor sequencing an accessible next-generation sequencing solution for all researchers, from whole genome/transcriptome to small genome and small RNA sequencing applications.
Compare the data for Ion PGM™ and Ion PI™ Chips, and choose the chip that works best for your sequencing needs:
Ion PGM™ Chip
Run time
Output
200 bp read
400 bp read
200 bp read
400 bp read
Ion 314™ Chip v2
2.3 hr
3.7 hr
30–50 Mb
60–100 Mb
Ion 316™ Chip v2
3.0 hr
4.9 hr
300–500 Mb
600 Mb–1 Gb
Ion 318™ Chip v2
4.4 hr
7.3 hr
600 Mb–1 Gb
1.2–2 Gb
Ion Proton™ Chip
Run time
Output
200 bp read
200 bp read
Ion PI™ Chip
2–4 hr
Up to 10 Gb
Featured Categories
Publications
Read and download Ion Torrent publications.
Technology
Explore the technology that makes sequencing faster and more accessible.
Products & Services
Learn more and order Ion Torrent™ sequencers, chips, reagents, and accessories.
Community
The Ion Torrent™ community is written collaboratively by the people who use it. Join the conversation!