Quantitative-Gene-Expression
TaqMan Assays are the gold standard in quantitative genomic analysis, offering high specificity, reproducibility, sensitivity and unsurpassed content.  Life Technologies offers several options for utilizing Taqman® assays as a tool for characterizing pluripotent stem cells.
Request Information
Which TaqMan Array is Right for your Pluripotent Stem Cells?
  Order now Order now Order now Order now
  Comparison to a reference standard Fast or standard run options Streamline loading Broadest data set
  New! TaqMan hPSC Scorecard Panel
Learn more >
TaqMan Human Stem Cell Pluripotency Panel (Taqman array plate) TaqMan Human Stem Cell Pluripotency Panel (TaqMan array card) TaqMan Open Array Human Stem Cell Panel, QuantStudio 12K Flex
No. of assays 94 92 92 631
Plate Format 96 and 384-well 96-well fast or standard plate 384-well microfluidic card OpenArray® Plate
Source of gene list Academic, Individual inventor
Reference publication
ISCI
Reference publication
ISCI
Reference publication
ISCI
Instrument compatibility QuantStudio 12K Flex, ViiA7™, StepOnePlus QuantStudio 12K Flex, 7500, ViiA7, StepOnePlus QuantStudio 12K Flex, ViiA7 QuantStudio 12K Flex with OpenArray block
Pluripotency measure
Trilineage differentiation measure      
New! Stem Cell Services for characterization
Characterization services can be provided as a stand-alone service or added to any CellModel Services reprogramming, differentiation, or assay development project.