RPMI   GIBCO® RPMI Media 1640 has a wide range of applications for mammalian cells, including the culture of fresh human lymphocytes, fusion protocols, and growth of hybrid cells.
Cell Culture with Advanced Media
Growth comparisons of several cell lines grown in Advanced (Reduced Serum) Media (1–2% FBS) compared to classical media (5% FBS) show equivalent or superior cell growth and no change in morphology.
View our Advanced RPMI 1640 Media
RPMI Media
teaser-circle.par.4555.image.56.56.1
Improve cell viability and growth with GlutaMAX™ Media and Supplement
While results may vary depending on the cell line, using GlutaMAX™-I Supplement in place of L-glutamine in cell culture can improve cell viability and growth, potentially increasing productivity levels. Increase media stability, minimize toxic ammonia build-up, and maximize cell performance. Get the most from your cell cultures with GlutaMAX™ media and Supplement .
Learn more about GlutaMAX™ media and GlutaMAX™-I Supplements