ProbesOnline, June 2009
In This Issue
FEATURED NEW PRODUCTS
Optimize Cell Viability Assays for Multicolor Panels LIVE/DEAD® Fixable Dead Cell Stain Sampler Kit
Detect Apoptosis with Nuclear Chromatin StainingVybrant® DyeCycle™ Violet/SYTOX® AADvanced™ Apoptosis Kit
New Mouse Targets for Flow CytometryPrimary Anti-mouse Monoclonal Antibody Conjugates
New Products for Cell &Tissue Analysis
See all of this month's New Products for Cell & Tissue Analysis
NEW APPLICATIONS
DEPARTMENTS
Check out the latest issue of BioProbes
FEATURED NEW PRODUCTS
what it is
LIVE/DEAD® cell viability assays are used to differentially stain live and dead cells in a variety of mammalian cell types. These cells can then be fixed with formaldehyde for subsequent analysis by flow cytometry. The LIVE/DEAD® Fixable Dead Cell Stain Sampler Kit contains all 8 LIVE/DEAD® reactive dyes that are excited with laser lines ranging from UV to far red, making it easier to identify the optimal dead cell stain for use in your multicolor panels.
what it offers
  • Post-fixation analysis—staining pattern preserved
  • Accuracy—eliminates staining artifacts
  • Flexibility—broad dye selection
how it works
LIVE/DEAD® cell viability assays employ the reaction of a fluorescent reactive dye with amines on cellular proteins. Viable cells exhibit faint staining on the cell surface, whereas the dye penetrates the interior of dead cells, resulting in at least a 50-fold increase in fluorescence compared to live cells. Because the dye reacts covalently with proteins, the staining pattern is completely preserved following sample fixation with formaldehyde.
LIVE/DEAD® Fixable Violet Dye
Live-cell gating using LIVE/DEAD® Fixable Violet dye eliminates staining artifacts from analysis. In a comparison between live-cell gating using scatter (A), and live-cell gating using LIVE/DEAD® Fixable Violet dye (B), staining artifacts using scatter gating are illustrated. The significant number of dead cells in subsequent analysis using scatter (C) are noted, as compared to the use of the LIVE/DEAD® Fixable Violet dye (D) to eliminate dead cells. Reproduced with permission from Elsevier ((J Immunol Methods 313:199(2006)).
Product Quantity Cat. No.  
LIVE/DEAD® Fixable Dead Cell Stain Sampler Kit 1 kit L34960 Order Now
what it is
Apoptotic cells undergo many characteristic morphological and biochemical changes, including the compaction and fragmentation of nuclear chromatin, plasma membrane shrinkage, and loss of membrane asymmetry. The Vybrant® DyeCycle™ Violet/SYTOX® AADvanced™ Apoptosis kit employs the combination of two Molecular Probes® proprietary dyes for fluorescence analysis of both the compacted state of nuclear chromatin and changes in plasma membrane integrity. 
what it offers
  • Convenience—reliable alternative to annexin V assays
  • Flexibility—free up space on the popular 488 nm laser
  • Accuracy—combination of two reliable assays
how it works
Vybrant® DyeCycle™ Violet stain (excitation/emission maxima ~370/440 nm) is a violet-excited, cell-permeant dye that binds to double-stranded DNA. Condensed chromatin of apoptotic cells stained with Vybrant® DyeCycle™ Violet stain appears brighter than the chromatin of normal cells. SYTOX® AADvanced™ dead cell stain (excitation/emission maxima ~546/660 nm) is a red-fluorescent, cell-impermeant dye that can only penetrate cells with damaged plasma membranes. The combination of these two dyes makes it easy to clearly distinguish normal, apoptotic, and dead-cell populations by flow cytometry.
Apoptosis in Jurkat Cells
Detection of apoptosis in Jurkat cells.
Jurkat cells (human T cell leukemia) were untreated (A) or treated with 10 μM camptothecin to induce apoptosis (B) Cells were then mixed with the reagents in the Vybrant® DyeCycle™ Violet/SYTOX® AADvanced™ Apoptosis Kit and analyzed by flow cytometry using 405/488 nm dual excitation. Note that the camptothecin-treated cells have a higher percentage of apoptotic cells than the basal level of apoptosis observed in the control cells. A = apoptotic cells, V = viable cells, N = necrotic cells.
Product Quantity Cat. No.  
Vybrant® DyeCycle™ Violet/SYTOX® AADvanced™ Apoptosis Kit
1 kit A35135 Order Now
what they are
Invitrogen now offers a broad range of antibodies with mouse reactivity for a variety of flow cytometry research applications including murine studies of T, B, and NK cells, cell adhesion, cell signaling, inflammatory responses, and MHC class I molecules. These primary anti-mouse monoclonal antibodies are conjugated to fluorescein isothiocyanate (FITC) and phycoerythrin (PE), and are validated for flow cytometry.
what they offer
  • New specificities—immunology targets
  • Mouse reactivity—for murine studies  
how they work
These FITC- and PE- labeled antibodies are excited by 488 nm lasers, and are commonly used with the first (FL1) and second (FL2) channels on most flow cytometers. FITC has fluorescence emission at 525 nm, while PE has emission at 575 nm.
Mouse Spleen Cells
Detection of CD45.1+ mouse spleen cells. SJL cells were stained with mouse anti-mouse CD45.1-FITC, and small lymphocytes were gated and analyzed on a FACScan™ flow cytometer (BDIS, San Jose, CA).
Product Quantity Cat. No.  
CD103, hamster anti-mouse, FITC 500 µg/1 mL HMCD10301 Order Now
CD103, hamster anti-mouse, PE 100 µg/1 mL HMCD10304 Order Now
CD107a (LAMP-1), rat anti-mouse, FITC 500 µg/1 mL RMCD107A01 Order Now
CD107b (LAMP-2), rat anti-mouse, FITC 500 µg/1 mL RMCD107B01 Order Now
CD154, hamster anti-mouse, FITC 500 µg/1 mL HMCD15401 Order Now
CD154, hamster anti-mouse, PE 100 µg/1 mL HMCD15404 Order Now
NEW APPLICATIONS
The Countess™ Automated Cell Counter
The Countess™ Automated Cell Counter uses trypan blue staining combined with a sophisticated image analysis algorithm to provide accurate cell and viability counts in just 30 seconds. The algorithm also measures the average size of live, dead, and total cells to give you all the data you need from your cell cultures without using a hemocytometer.
Counting Blood Cells is Now Faster and Easier
The Countess™ Automated Cell Counter has also been used to accurately count blood cells including peripheral blood mononuclear cells (PBMCs), red blood cells (RBCs), and white blood cells (WBCs) from lysed whole blood and Ficoll preparations. Because the Countess™ instrument uses trypan blue staining, the viability of WBCs is easily determined, while platelets and noncellular debris are unstained and excluded from the count. It should be noted that the Countess™ instrument does not differentiate between WBC types and that viability information is not available for RBCs.
Countess Cell Counter    
Product Quantity Cat. No.  
Countess™ Automated Cell Counter each C10227 Order Now
Countess™ Automated Cell Counter Starter Kit
(includes 1 cell counter and 11 boxes of slides)
1 kit C10310 Order Now
Countess™ Automated Cell Counter Lab Starter Kit
(includes 1 cell counter and 101 boxes of slides)
1 kit C10311 Order Now
Countess™ Cell Counting Chamber Slides 50 slides (100 counts) C10228 Order Now
Countess™ Cell Counting Chamber Slides 500 slides (1,000 counts) C10312 Order Now
Countess™ Cell Counting Chamber Slides 1,250 slides (2,500 counts) C10313 Order Now
Countess™ Cell Counting Chamber Slides 2,500 slides (5,000 counts) C10314 Order Now
Countess™ Cell Counting Chamber Slides 5,000 slides (10,000 counts) C10315 Order Now
DEPARTMENTS
Buzzworthy
The FluxOR™ Assay Enables Analysis of Electric Pulse–induced Membrane Nanopores
Lipid nanopores can form a stable, ion channel–like conduction pathway in cell membrane.
Pakhomov, A.G. et al. (2009) Biochem Biophys Res Commun (manuscript accepted).
Can electrical pulses induce ion-selective membrane disruption?
It is widely known that electric pulses can cause the formation of pores in cell membranes; the phenomenon is the basis for electroporation, an established technique for introducing substances into cells. In their recent study, Pakhomov and colleagues describe the formation and properties of very small membrane pores—"nanopores"—following the application of nanosecond electric pulses (nsEPs).
Methods
Electrophysiological observations of nanopore behavior were performed through the use of a modified FluxOR™ assay, wherein the uptake of thallium ion (Tl+) by nsEP-treated cells was monitored via an increase in the Tl+-dependent fluorescence emission of the FluxOR™ probe.
Results
In response to applied nsEPs, both GH3 cells (characterized by the presence of multiple endogenous ion channel types) and CHO-K1 cells (which express relatively few endogenous channel types) exhibited an increase in whole-cell conductance and inward current rectification. Nanopores are so called because the group demonstrates them to be mostly impermeable to the fluorescent probe propidium iodide, setting an effective upper pore size limit of ~1 nm. nsEP-induced nanopores were stable for many minutes, and eventually dissipated either by spontaneous resealing or by the formation of larger, propidium-permeant pores.
The group suggests that nanopores could result through a number of mechanisms in addition to the application of nsEPs, and their relatively long lifetime merits their consideration as an alternative ion-selective pathway that could impact the interpretation of other studies of transmembrane ion flow. 
View bibliography reference
Product Quantity Cat. No.  
FluxOR™ Thallium Detection Kit *for 10 microplates* 1 kit F10016 pHrodo, SE
FluxOR™ Thallium Detection Kit *for 100 microplates* 1 kit F10017 pHrodo S. aureus BioParticles
Free Online Technical Webinars   Free online technical webinars
You are invited to join us for a series of biweekly technical webinars from the comfort of your desk. The webinars will initially focus on imaging-related applications, but we welcome your feedback for additional topics throughout the course of the year. Upcoming topics will be announced each month via email.
Presentations will last approximately 45 minutes, followed by 15 minutes for live Q&A.
Webinars Date Time
Approaches to Cell Proliferation and Cell Counting June 23, 2009 10:00 a.m. PT
Assessing Cell Health and Viability July 7, 2009 10:00 a.m. PT
Missed our previous webinars? Find our recorded webinars here!
Glial Fibrillary Acidic Protein (GFAP)   Midsagittal section of rat cerebellum stained to reveal IP3 receptors and glial fibrillary acidic protein (GFAP). Purkinje neurons (green) were stained using a rabbit primary antibody targeted to the IP3 receptor, and visualized with Qdot® 525 goat F(ab')2 anti-rabbit IgG conjugate. In astrocytes (red), GFAP was stained using a mouse primary antibody targeted to GFAP, and visualized with Qdot® 655 goat F(ab')2 anti-mouse IgG conjugate. Nuclei were stained blue with Hoechst 33342. Image submitted by Tom Deerinck, University of California, San Diego.
Product Quantity Cat. No.  
Qdot® 525 goat F(ab')2 anti-rabbit IgG conjugate 200 µL Q11441MP Order Now
Qdot® 655 goat F(ab')2 anti-mouse IgG conjugate 200 µL Q11021MP Order Now
Hoechst 33342 trihydrochloride, trihydrate *FluoroPure™ grade 100 mg H21492 Order Now
Antibody Conjugation Kits: Minimize Time, Maximize Yield
Using a direct antibody conjugate for fluorescence imaging or flow cytometry can eliminate the noise commonly associated with nonspecific secondary antibody binding. Directly labeled antibodies also allow you to use more than one same-species antibody in a single experiment. Invitrogen offers a wide variety of antibody conjugation kits for labeling µg to mg quantities of IgG antibody with bright and photostable Alexa Fluor® dyes or biotin.
Molecular Probes® antibody labeling kits have been carefully configured to minimize hands-on time while maximizing conjugate yield. Labeled antibodies are ready in as little as 2 hours and require only ~10–20 minutes of hands-on time. Which labeling kit to use depends entirely on the amount of antibody to be labeled. 
The kits use time-tested, amine-reactive labels to covalently attach a fluorophore or biotin to your antibody. The result is a strong carboxamide linkage between the label and the antibody, with exceptional stability—from months to years.
Invitrogen’s comprehensive selection of antibody labeling kits is based on the amount of antibody to be labeled.
Immunoassay Reagents   Build Your Own Immunoassay
Our new web resource makes it easy to develop your own immunoassay—a customizable alternative to using traditional ELISA kits. Find validated antibody pairs, ELISA-grade single antibodies, and proven detection reagents—from standard TMB chromogen to fluorescent Amplex® UltraRed—for your platform and desired sensitivity.
The Cell Resource Center
The Cell Resource Center is your portal to a wide range of cellular technologies including GIBCO® cells, media, and reagents, Molecular Probes® fluorescent reagents and detection technologies, Dynal® bead-based isolation systems, drug discovery products, and services to support your research needs.
  • Learn More about Cellular Technologies
While you're there, check out the new Technology Chalkboard!
Technology Chalkboard    
Ultra-High–Throughput Cell-based Assays
In collaboration with TTP LabTech, Invitrogen scientists have validated Molecular Probes® high-content screening (HCS) cell health and cytotoxicity kits on the ultra-high–throughput laser scanning Acumen® eX3 system, a high-content imaging platform for the rapid generation of multiplexed cell-based data. The combination of robust multiparametric HCS kits with rapid, whole-well data generation enabled by the Acumen® eX3 system offers a whole new level of throughput and content for a variety of cell-based imaging applications. 
  • Download posters from the Invitrogen/TTP LabTech Collaboration, and other posters relevant to high-content imaging and analysis.
Glial Fibrillary Acidic Protein (GFAP)   Midsagittal section of rat cerebellum stained to reveal IP3 receptors and glial fibrillary acidic protein (GFAP). Purkinje neurons (green) were stained using a rabbit primary antibody targeted to the IP3 receptor, and visualized with Qdot® 525 goat F(ab')2 anti-rabbit IgG conjugate. In astrocytes (red), GFAP was stained using a mouse primary antibody targeted to GFAP, and visualized with Qdot® 655 goat F(ab')2 anti-mouse IgG conjugate. Nuclei were stained blue with Hoechst 33342. Image submitted by Tom Deerinck, University of California, San Diego.
Product Quantity Cat. No.  
Qdot® 525 goat F(ab')2 anti-rabbit IgG conjugate 200 µL Q11441MP Order Now
Qdot® 655 goat F(ab')2 anti-mouse IgG conjugate 200 µL Q11021MP Order Now
Hoechst 33342 trihydrochloride, trihydrate *FluoroPure™ grade 100 mg H21492 Order Now
Antibody Conjugation Kits: Minimize Time, Maximize Yield
Using a direct antibody conjugate for fluorescence imaging or flow cytometry can eliminate the noise commonly associated with nonspecific secondary antibody binding. Directly labeled antibodies also allow you to use more than one same-species antibody in a single experiment. Invitrogen offers a wide variety of antibody conjugation kits for labeling µg to mg quantities of IgG antibody with bright and photostable Alexa Fluor® dyes or biotin.
Molecular Probes® antibody labeling kits have been carefully configured to minimize hands-on time while maximizing conjugate yield. Labeled antibodies are ready in as little as 2 hours and require only ~10–20 minutes of hands-on time. Which labeling kit to use depends entirely on the amount of antibody to be labeled. 
The kits use time-tested, amine-reactive labels to covalently attach a fluorophore or biotin to your antibody. The result is a strong carboxamide linkage between the label and the antibody, with exceptional stability—from months to years.
Invitrogen’s comprehensive selection of antibody labeling kits is based on the amount of antibody to be labeled.
Immunoassay Reagents   Build Your Own Immunoassay
Our new web resource makes it easy to develop your own immunoassay—a customizable alternative to using traditional ELISA kits. Find validated antibody pairs, ELISA-grade single antibodies, and proven detection reagents—from standard TMB chromogen to fluorescent Amplex® UltraRed—for your platform and desired sensitivity.
The Cell Resource Center
The Cell Resource Center is your portal to a wide range of cellular technologies including GIBCO® cells, media, and reagents, Molecular Probes® fluorescent reagents and detection technologies, Dynal® bead-based isolation systems, drug discovery products, and services to support your research needs.
  • Learn More about Cellular Technologies
While you're there, check out the new Technology Chalkboard!
Technology Chalkboard    
Ultra-High–Throughput Cell-based Assays
In collaboration with TTP LabTech, Invitrogen scientists have validated Molecular Probes® high-content screening (HCS) cell health and cytotoxicity kits on the ultra-high–throughput laser scanning Acumen® eX3 system, a high-content imaging platform for the rapid generation of multiplexed cell-based data. The combination of robust multiparametric HCS kits with rapid, whole-well data generation enabled by the Acumen® eX3 system offers a whole new level of throughput and content for a variety of cell-based imaging applications. 
  • Download posters from the Invitrogen/TTP LabTech Collaboration, and other posters relevant to high-content imaging and analysis.
Molecular Probes® The Handbook
©2009 Life Technologies Corporation. All rights reserved. The trademarks mentioned herein are the property of Life Technologies Corporation or their respective owners. Feel free to distribute ProbesOnline to friends and colleagues, but please keep this copyright statement intact.