Quicklinks
Related Product Information
Introduction - E-Gel® Electrophoresis System
SYBR Safe™ DNA Gel Stain
SYBR Safe™ DNA gel stain has been specifically developed for reduced mutagenicity, making it safer than ethidium bromide for staining DNA in agarose gels. The detection sensitivity of E-Gel® with SYBR Safe™ stain is similar to that of E-Gel® containing ethidium bromide. DNA bands stained with SYBR Safe™ DNA gel stain can be detected using a standard UV transilluminator, a visible-light transilluminator or a laser-based scanner.
Safety Features of SYBR Safe™
  • SYBR Safe™ DNA gel stain is not classified as hazardous waste under U.S. Federal regulations
  • SYBR Safe™ stain meets the requirements of the Clean Water Act and the National Pollutant Discharge Elimination System regulations (though E-Gel® with SYBR Safe™ generally does not generate liquid waste).
  • SYBR Safe™ DNA gel stain does not induce transformations in primary cultures of Syrian hamster embryo (SHE) cells. In contrast, ethidium bromide tests positive in the SHE cell assay, consistent with its known activity as a strong mutagen.
  • SYBR Safe™ stain does not cause mutations in mouse lymphoma cells at the TK locus, nor does it induce chromosomal aberrations in cultured human peripheral blood lymphocytes, with or without S9 metabolic activation.
  • Compared to ethidium bromide, SYBR Safe™ DNA gel stain causes fewer mutations in the standard Ames test. Weakly positive results occurred in only four out of seven Salmonella strains and only with activation by a mammalian S9 fraction.
  • A single oral administration of SYBR Safe™ DNA gel stain produces no signs of mortality or toxicity at a limit dose of 5000 mg/kg.
  • View studies documenting the safety of SYBR Safe™ in the SYBR Safe™ White Paper document.
Disposal of SYBR Safe™
SYBR Safe™ DNA gel stain is not classified as hazardous waste, but as disposal regulations vary, please contact your safety office or local municipality for appropriate SYBR Safe disposal in your community.
Spectrum of SYBR Safe™ Bound to nucleic acids, SYBR Safe™ stain has fluorescence excitation maxima at 280 and 502 nm, and an emission maximum at 530 nm (see figure below).
Normalized fluorescence excitation and emission spectra of SYBR Safe™ DNA gel stain, determined in the presence of DNA
Visualization of SYBR Safe™
Detect DNA bands stained with SYBR Safe™ DNA gel stain using a blue-light transilluminator, a standard UV transilluminator, or a laser-based scanner. For photographing gels, a special filter may be required.
Cloning Benefits of SYBR Safe™
Using the SYBR Safe™-blue-light method dramatically reduces DNA damage. As a result, cloning efficiency can improve ten- to thousand-fold.
TOP
E-Gel® Electrophoresis System
Introduction
The E-Gel® agarose gel electrophoresis system is a complete bufferless system for agarose gel electrophoresis of DNA samples.
The major components of the system are:
  • E-Gel® pre-cast agarose gels
  • E-Gel® PowerBase™ v.4. or Mother E-Base™
E-Gel® pre-cast agarose gels are self-contained gels that include electrodes packaged inside a dry, disposable, UV-transparent cassette. The E-Gel® agarose gels run in a specially designed device that is a base and power supply combined into one device. They either contain SYBR Safe™ DNA gel stain, ethidium bromide, or no DNA gel stain.
Advantages of E-Gel®
Using E-Gel® agarose gels for electrophoresis of DNA samples offer the following advantages:
  • Provides fast, safe, consistent, high-resolution electrophoresis
  • Eliminates the need to prepare agarose gels and buffers, and to stain gels
  • Compatible with most commercially available robotic systems for high-throughput agarose gel electrophoresis
  • Available in a variety of agarose percentages, well formats, and throughput to suit your applications
  • Offered with SYBR Safe™ DNA gel stain, a safer, more environmentally friendly alternative to ethidium bromide
Available DNA Gel Stains
E-Gel® agarose gels come in three formats for staining your DNA:
  • Regular E-Gel® agarose gels contain the standard DNA gel stain ethidium bromide
  • E-Gel® with SYBR Safe™ contains SYBR Safe™ DNA gel stain, which is not classified as hazardous waste under U.S. Federal regulations, and improves cloning efficiency when using blue light for imaging
  • Clear E-Gel® does not contain a DNA gel stain, and is intended for custom staining or for analyzing fragments coupled to a fluorescent label.
SYBR Safe™ DNA gel stain
offers the following advantages:
  • SYBR Safe™ DNA gel stain is not classified as hazardous waste under U.S. Federal regulations and meets the requirements of the Clean Water Act and the National Pollutant Discharge Elimination System regulations.
  • SYBR Safe™ DNA gel stain does not cause mutations, chromosomal aberrations, or transformations in appropriate mammalian test systems, in contrast to ethidium bromide which is a strong mutagen.
  • A single oral administration of SYBR Safe™ DNA gel stain produces no signs of mortality or toxicity at a limit dose of 5000 mg/kg.
  • Visualizing E-Gel® with SYBR Safe™ using blue light transilluminators dramatically reduces DNA damage that lowers cloning efficiency.
  • For details on SYBR Safe™ DNA gel stain 
Throughput Formats
Two types of E-Gel® agarose electrophoresis systems are available from Invitrogen based on your throughput requirements.
  • Low-Throughput E-Gel® Electrophoresis System
  • Medium/High-Throughput E-Gel® Electrophoresis System
Low-Throughput E-Gel Electrophoresis System
The Low-Throughput E-Gel® Electrophoresis System is designed for electrophoresis of 12-16 DNA samples per gel. The system consists of the following components:
  • E-Gel® with SYBR Safe™, E-Gel® single comb, double comb, and Clear E-Gel® pre-cast agarose gels
  • Gels are available in a variety of well formats and percentages. Choose an appropriate gel based on your application.
  • E-Gel® PowerBase™ v.4
  • The E-Gel® PowerBase™ v.4 is a base and a power supply in one device.
  • The E-Gel® PowerBase™ connects directly to an electrical outlet using the adaptor supplied with the base. (page 6). •E-Gel® Opener
  • The E-Gel® Opener is an easy-to-use device specifically designed to open any E-Gel® single comb, double comb, or Clear E-Gels®.
The Medium/High-Throughput E-Gel® Electrophoresis System
The Medium/High-Throughput E-Gel® Electrophoresis System is designed for electrophoresis of 48-96 DNA samples per gel. This system is compatible for use with multichannel pipettors or automated liquid handling systems.
The system consists of the following components:
E-Gel® 48 gels 
TOP
  • Each E-Gel® 48 gel contains 48 sample lanes and 4 marker lanes and is designed for medium-throughput agarose electrophoresis of nucleic acids.
E-Gel® 96 gels
  • Each E-Gel® 96 gel contains 96 sample lanes and 8 marker lanes in a patented, staggered-well format and is designed for high-throughput agarose electrophoresis of nucleic acids.
E-Base™ Electrophoresis Device
  • The E-Base™ is a base and a power supply all in one device and is an easy-to-use, pre-programmed device specifically designed for electrophoresis of E-Gel® 48 and 96 gels.
E-Holder™ Platform
  • The E-Holder™ Platform is designed to hold E-Gel® 96 gels during robotic loading. The E-Holder™ is used to load multiple gels on a robotic platform while other gels are running on the E-Base™.
E-Editor™ 2.02 Software
  • The E-Editor™ 2.02 software allows you to quickly reconfigure digital images of E-Gel 48 or 96 gel results for analysis and documentation. The E-Editor™ 2.02 software can be downloaded for free from our Web site at www.lifetechnologies.com/egels.
Choosing a Gel for Your Application
To obtain the best results for your application, it is important to choose the correct agarose percentage and well format.
The table below lists the various types of gel and resolution for each gel type.
Gel Type
No. Rows
No. Wells
Sample Volume
Run Length
Run Time
% Agarose
Resolution
E-Gel ® with SYBR Safe™
1
12
10-20 µl
5.8 cm
30 min.
1.2%
2%
100 bp-5 kb
100 bp-2 kb
E-Gel ® single comb
1
12
10-20 µl
5.8 cm
30 min.
0.8%
1.2%
2%
4%
800 bp-10 kb
100 bp-5 kb
100 bp-2 kb
20 bp-500 bp
E-Gel ® double comb
2*
16 sample
2 marker
10-20 µl
10 µl
2.9 cm
15 min.
0.8%
2%
1 kb-10 kb
100 bp-2 kb
Clear E-Gel ®
1
12
20 µl
5.8 cm
30 min.
1.2%
100 bp-5 kb
E-Gel ® 48 Gel
2*
48 sample
4 marker
10-15 µl
15 µl
3.2 cm
20 min.
1%
2%
4%
400 bp-10 kb
50 bp-3 kb
10 bp-400 bp
E-Gel ® 96 Gel
8*
96 sample
8 marker
10-20 µl
20 µl
1.6 cm
12 min.
1%
2%
1 kb-10 kb
100 bp-2 kb
*Wells compatible for loading with a multichannel pipettor.
TOP
Low-Throughput E-Gel® Electrophoresis System
System Components
The Low-Throughput E-Gel® Electrophoresis System consists of the following components:
  • E-Gel® with SYBR Safe™, E-Gel® single comb, double comb, and Clear E-Gel® pre-cast agarose gels 
  • E-Gel® PowerBase™ v.4
  • E-Gel® Opener
Note:   The E-Gel® Base previously available from Invitrogen can be used for electrophoresis of E-Gel® with SYBR Safe™, E-Gel® single comb, double comb, and Clear E-Gel® agarose gels.
Applications
E-Gel® agarose gels are suitable for analyzing:
  • PCR products
  • Restriction digests
  • RT-PCR reactions
 When to Use E-Gel® With SYBR Safe™
  • E-Gel® with SYBR Safe™ contains SYBR Safe™ DNA gel stain instead of ethidium bromide. Use E-Gel® with SYBR Safe™ for these reasons:
  • If you want to minimize your hazardous waste, since SYBR Safe™ DNA gel stain is not classified as such under U.S. Federal regulations.
  • If you want to protect yourself or your co-workers, because E-Gel® with SYBR Safe™ eliminates the use of the strong mutagen ethidium bromide and reduces UV exposure.
  • If you want to maximize cloning efficiency, since E-Gel® with SYBR Safe™ dramatically reduces DNA damage if using blue light transilluminators.  
When to Use E-Gel® With SYBR Safe®
Clear E-Gel® agarose gels do not contain ethidium bromide in the gels and are ideal for the following applications:
  • Post-staining gels with sensitive stains such as SYBR® Green I or II, or SYBR® Gold.
  • Analyze DNA fragments pre-labeled with fluorescein, Texas Red® or Alexa Fluor®
  • Perform downstream applications with DNA in which ethidium bromide may interfere
  • Determine accurate size and form of the DNA band (most DNA staining dyes intercalate into the DNA and change molecular weight and form of the DNA)
  • Stain only one lane of the gel and then excise a specific band or size range in the remaining gel for cloning purposes thereby avoiding UV exposure to the DNA resulting in a significant increase in the cloning efficiency.
E-Gel® Single Comb and Double Comb Gels
The E-Gel® single comb and double comb gels are bufferless gels containing electrodes embedded in the agarose matrix. Each gel contains an ion generating system (TAE buffer system), a pH balancing system, and ethidium bromide for DNA staining and is packaged inside an UV-transparent cassette.
To create a patented bufferless system, each E-Gel® single comb and double comb cassette contains two ion exchange matrices (IEMs) that are in contact with the gel and electrodes. The IEMs supply a continuous flow of ions through out the gel resulting in a sustained electric field required for running the gel (see figure below).
Clear E-Gel® agarose gels are single comb gels that do not contain any nucleic acid stain in the agarose.
E-Gel® PowerBase™
The E-Gel® PowerBase™ Version 4 (figure below) is an easy-to-use, automated device specifically designed to simplify electrophoresis of single comb or double comb E-Gel® agarose gels from Invitrogen. The E-Gel® PowerBase™ is a base and a power supply all in one device.
The operation of the E-Gel® PowerBase™ v. 4 is controlled by two buttons on top of the base. The left button is for a double comb run and right button is for a single comb run (see the label on the unit). To select different electrophoresis runs for the PowerBase™, do one of the following.
  • Press and release the button OR
  • Press and hold the button 
E-Gel® Base
The E-Gel® Base (see figure below) previously available from Invitrogen connects to a power supply and is used for electrophoresis of E-Gel® single comb, double comb, and Clear E-Gel® agarose gels.
E-Gel® Opener
The E-Gel® Opener is an easy-to-use device specifically designed to open any E-Gel® single comb, double comb, or Clear E-Gels® for excision of DNA fragments or for blotting.
TOP
Product Specifications
E-Gel® Single Comb and Double Comb Gel Specifications
The E-Gel ® cassette is 8 cm x 10 cm and 0.6 cm thick. The thickness of the E-Gel ® gel is 3 mm and the volume of the gel is 20 ml.
  • Single comb gel—Each well is 4.1 mm wide and the space between wells is 1 mm. The running distance is 5.8 cm. Each gel contains 12 lanes.
  • Double comb gel—The sample well is 4.6 mm wide and the marker well is 2.8 mm wide. The running distance from each comb is 2.9 cm. Each gel contains two rows of 8 sample wells and 2 marker wells (M). The wells of the double comb gel are compatible for loading with a multichannel pipettor.
E-Gel® 48 Gel Specifications
Each E-Gel ® 48 gel contains 48 sample wells and 4 marker wells (M).
Cassette Size: 13.5 cm (l) x 10.8 cm (w) x 0.67 cm (thick)
Gel Thickness: 3.7 mm
Gel Volume: 50 ml
Gel Percentage: 1%, 2%, and 4%
Well Depth: 3 mm
Dimensions of the Well: 3.6 mm (l) x 2.2 mm (w)
Running Distance (one well to the next): 32 mm
Space between Well Center: 4.5 mm
The wells of the E-Gel ® 48 gel are compatible for loading with a multichannel pipettor.
E-Gel® 96 Gel Specifications
Each E-Gel ® 96 gel contains 96 sample wells and 8 marker wells (M).
Cassette Size: 13.5 cm (l) x 10.8 cm (w) x 0.67 cm (thick)
Gel Thickness: 3.7 mm
Gel Volume: 50 ml
Gel Percentage: 1% & 2%
Well Depth: 3 mm
Well Opening: 3.8 mm x 1.8 mm
Well Bottom:
3.3 mm x 1.1 mm
Running Distance (one well to the next): 16 mm
Space between Wells: 9 mm
The wells of the E-Gel ® 96 cassette are compatible with a multichannel pipettor or 8, 12, or 96-tip robotic loading devices.
TOP
E-Base™ Specifications
The specifications for Mother E-Base™ and Daughter E-Base™ are listed below.
Dimensions: 14.6 cm x 15 cm x 5.3 cm
Weight:
 Mother E-Base™- 370 g
 Daughter E-Base™- 271 g
Safety: Double Insulation, UL listed, and CE certified
Temperature: Ambient 15°C to 40°C
Built-in Features: Digital time display (00-99 minutes), alarm, light LED
The SBS (Society for Biomolecules Screening) standard 96-well plate format of the E-Base™ fits on most robotic platforms allowing the loading and electrophoresis of gels on the E-Base™ directly on the robot.
E-Gel® PowerBase™ v.4 Specifications
The specifications for E-Gel ® PowerBase™ V.4 are listed below.
Dimensions: 12.5 cm x 13 cm x 13.5 cm
Weight:
1.19 lbs (540 g) with adaptor
Safety: UL listed and CE certified
Temperature: Ambient 15°C to 40°C
Built-in Features: alarm, light LED
E-Gel® PowerBase™ Adaptor Specifications
The E-Gel ® PowerBase™ v.4 is designed for use with an adaptor included with the PowerBase™. Use only UL Listed Class 2 Direct Plug-in Adaptor included with the PowerBase™. Input and Output supplied by the adaptor are shown in the table below.
12 V DC, 880 mA
Country Input Output
U.S. and Canada 110-120 V AC, 60 Hz12 V DC, 880 mA  
Europe 220-240 V AC, 50 Hz 12 V DC, 880 mA
E-Gel® Base Specifications
The specifications for E-Gel ® Base are listed below:
Dimensions: 12.5 x 13 x 3.5 cm
Weight:
3.18 oz. (90 g)
Temperature: Ambient 15C to 40°C 
TOP
Sample Preparation for E-Gel® with SYBR Safe™
Introduction
For optimal results, follow the guidelines for preparing your DNA sample as described in this section.
Materials Needed
  • DNA sample
  • Loading buffer (optional)
  • Molecular weight markers
Amount of DNA
Use 20-100 ng DNA per band for samples containing one unique band, or up to 500 ng per lane (E-Gel® 1.2% with SYBR Safe™) or 700 ng per lane (E-Gel® 2% with SYBR Safe™) of samples containing multiple bands. If you are unsure how much to use, test a range of concentrations to determine the optimal concentration for your particular sample. Excess DNA will cause poor resolution.
Loading Method
The DNA samples are loaded in E-Gel® agarose gels using the One-Step Loading or Two-Step Loading method.
The One-Step Loading method is routinely used to load E-Gel® agarose gels.
The Two-Step Loading method is used to load E-Gel® agarose gels, if the One-Step Loading method produces fuzzy or indistinct bands, or the gel was removed from its plastic pouch for more than 15 minutes.
Prepare loading buffer and sample volumes based on the loading method as described below.
Total Sample Volume
The recommended total sample volume for each gel type is listed in the table below.
Note:   For best results, keep all sample volumes uniform. If you do not have enough samples to load all wells of the gel, load an equal volume of deionized water into any empty wells.
Gel Type
One-Step Loading
Two-Step Loading
E-Gel ® with SYBR Safe
20 µl
10 µl
Loading Buffer
TOP
  • Use of loading buffer is optional.
  • You can load your samples directly into the wells, if no loading buffer is used. If you are using loading buffer, mix the required amount of DNA with the loading buffer. We recommend using a loading buffer with the following formulation:
  • For One-Step Loading method, use a loading buffer that yields a final concentration of 10 mM Tris-HCl, pH 7.5; 1 mM EDTA; 0.005% bromophenol blue; 0.005% xylene cyanol FF.
  • For Two-Step Loading method, use a loading buffer that yields a final concentration of 10% glycerol (or 6% Ficoll 400) 10 mM Tris-HCl, pH 7.5; 1 mM EDTA; 0.005% bromophenol blue; 0.005% xylene cyanol FF
  • If you are using 10X BlueJuice™ Gel Loading Buffer or TrackIt™ Loading Buffer available from Invitrogen, dilute this buffer 50- to 200-fold to obtain optimal results with E-Gel® agarose gels. For Two-Step loading with 50- to 200 fold diluted 10X BlueJuice™ Gel Loading Buffer or TrackIt™ Loading Buffer, 10% glycerol may be added.
High Salt Samples
Important: To obtain the best results, prepare your high salt samples as described below.
Samples containing > 20 mM NaCl or 5 mM EDTA (i.e. certain restriction enzyme and PCR buffers) will cause loss of resolution on E-Gel® agarose gels. Dilute samples 2- to 20-fold as described:
  • Restriction Digests—Digest 500 ng-1 µg of DNA in 20 µl final volume. Use 2 µl and dilute as described below.
  • PCR Samples—Use 1-5 µl of a 50 µl reaction and dilute as described below.
One-Step Loading
Two-Step Loading
Dilute samples with the loading buffer, deionized water, or TE to a final volume of 20 µl
Dilute samples with loading buffer, deionized water, or TE buffer to obtain a final sample volume of 10 µl (10% glycerol may be added)
Preparing Samples
Prepare your samples based on the loading method used as described below:
One-Step Loading
Two-Step Loading
Total Sample Volume
Add deionized water to the required amount of DNA to bring the total sample volume to 20 µl.
Mix the required amount of DNA with deionized water to obtain a total sample volume of 10 µl (10% glycerol may be added). When loading into the gel, first load 10 µl of deionized water directly into each well and then add 10 µl of sample.
Loading Buffer
Optional: Instead of water, you may use a loading buffer that yields a final concentration of 10 mM Tris-HCl, 1 mM EDTA, pH 7.5; 0.005% bromophenol blue; and 0.005% xylene cyanol FF.
To use 10X BlueJuice™TrackIt™ Loading Buffer, dilute this buffer 50- to 200-fold to obtain the optimal dye concentration.
Optional: Instead of water, you may use a loading buffer that yields a final concentration of 10 mM Tris-HCl, pH 8.1; 1 mM EDTA, pH 7.5; 0.005% bromophenol blue; 0.005% xylene cyanol FF (10% glycerol may be added).
To use 10X BlueJuice TrackIt™ Loading Buffer, dilute this buffer 50- to 200-fold to obtain the optimal dye concentration.
DNA Molecular Weight Markers
We recommend using the following DNA molecular weight markers for different types of E-Gel® agarose gels to obtain good resolution.
Note:   Supercoiled DNA molecular weight markers may produce a slightly fuzzy pattern when run on E-Gel® with SYBR Safe™ agarose gels. Consider using Clear E-Gel® agarose gels and post-staining with SYBR Safe™ DNA Gel Stain.
Product
Markers
Catalog no.
Amount Used
E-Gel ® 1.2% with SYBR Safe
TrackIt 100 bp DNA Ladder
10488-058
Load 500-700 ng markers in a volume of 20 µl.
TrackIt 1 Kb Plus DNA Ladder
10488-085
100 bp DNA Ladder
15628-019
1 Kb Plus DNA Ladder
10787-018
High DNA Mass Ladder
10496-016
E-Gel ® High Range DNA Marker
12352-019
E-Gel ® 2% with SYBR Safe
TrackIt 25 bp DNA Ladder
10488-022
TrackIt 50 bp DNA Ladder
10488-043
25 bp DNA Ladder
10597-011
50 bp DNA Ladder
10416-014
100 bp DNA Ladder
15628-019
Low DNA Mass Ladder
10068-013
E-Gel ® Low Range Quantitative DNA Marker
12373-031
TOP
Loading E-Gel® with SYBR Safe™ Gels
Introduction
After you have prepared your samples, you are ready to proceed with electrophoresis. Instructions are provided below to load and run E-Gel® with SYBR Safe™ (which come as single comb gels) using the E-Gel® PowerBase™ v.4.The total run times are 30–33 minutes.
Pre-running E-Gel® with SYBR Safe™
You must first pre-run the E-Gel® with SYBR Safe™ agarose gel for 2 minutes with the comb in place before loading your samples to ensure proper resolution of your DNA fragments.
Each E-Gel® cassette is supplied individually wrapped and ready for use. To set up and use an E-Gel® with SYBR Safe™ gel, follow the instructions below:
  1. Plug the E-Gel® PowerBase™ v.4 into an electrical outlet using the adaptor plug.
  2. Open the package containing the gel and insert the gel (with the comb in place) into the apparatus right edge first. Press firmly at the top and bottom to seat the gel in the base. You should hear a snap when it is in place. The Invitrogen logo should be located at the bottom of the base, close to the positive pole. See the diagram below. A steady, red light will illuminate when the E-Gel® gel is correctly inserted (Ready Mode).
  3. Press and hold either button until the red light turns to a flashing green light. This indicates that the 2-minute pre-run has started.
  4. At the end of the pre-run, current will automatically shut off. The flashing green light will change to a flashing red light and the PowerBase™ will beep rapidly.
  5. Press and release either button to stop the beeping (you will hear only one beep). The light will change from a flashing red light to a steady red light.
 Method of Loading Samples
The E-Gel® with SYBR Safe™ agarose gels are designed for loading samples manually using either the One-Step Loading method or the Two-Step Loading method. The One-Step Loading method is routinely used to load E-Gel® with SYBR Safe™ gels; use the Two-Step Loading method, if the One-Step Loading method produces fuzzy or indistinct bands, or the gel was removed from its plastic pouch for more than 15 minutes.
One-Step Loading Method
All wells in the gel must contain sample or water. Avoid introducing bubbles while loading, as bubbles will cause bands to distort.
  1. Remove the comb from the E-Gel® with SYBR Safe™ gel using both hands to lift the comb gently by rolling the comb slowly towards you. Be careful to pull the comb straight up from both sides. Do not bend the comb. Remove any excess fluid using a pipette.
  2. Load 20 µl of sample per sample well.
  3. Load 20 µl (500-700 ng) of the appropriate molecular weight markers.
  4. Load 20 µl of water into any remaining empty wells.
Alternate Method:
Two-Step Loading
All wells in the gel must contain sample or water. Avoid introducing bubbles while loading, as bubbles will cause bands to distort.
  1. Remove the comb from the E-Gel® with SYBR Safe™ gel using both hands to lift the comb gently by rolling the comb slowly towards you. Be careful to pull the comb straight up from both sides. Do not bend the comb. Remove any excess fluid using a pipette.
  2. Load 10 µl of deionized water into each well (including sample, molecular weight marker and empty wells). Do not premix with sample.
  3. Load 10 µl of sample containing per sample well (10% glycerol may be added).
  4. Load 10 µl (500-700 ng) of the appropriate molecular weight markers (10% glycerol may be added).
  5. Load 10 µl of water into any remaining empty wells (10% glycerol may be added).
TOP
Running E-Gel® with SYBR Safe™ Gels
Electrophoresis of E-Gel® with SYBR Safe™
  1. Choose the 30-minute run for E-Gel® with SYBR Safe™ gels. Press and release the 30-min button to start the electrophoresis run. The light will change to a steady green light.
  2.  
    Note:   The actual running time may vary between 30-33 minutes.
  3. Current through the E-Gel® gel automatically shuts off at the end of each run. The E-Gel® PowerBase™ v.4 signals the end of the run with a flashing red light and rapid beeping.
  4. Press and release either button to stop the beeping. The light will turn to a steady red light.
  5. At the end of the run, remove the gel cassette from the power unit and analyze your results as described in the next sections.
E-Gel® agarose gels can only be used once. Do not re-use them.
E-Gel® with SYBR Safe™ gels already contain the SYBR Safe™ DNA gel stain, and therefore do not have to be stained after the electrophoresis run
.
TOP
Visualizing E-Gel® with SYBR Safe™ Agarose Gels
Introduction
Bound to nucleic acids, SYBR Safe™ DNA gel stain has fluorescence excitation maxima at 280 and 502 nm, and an emission maximum at 530 nm. Use a blue light or UV light transilluminator to view the gel; a filter is required to photograph the gel (your standard ethidium bromide filter may not be appropriate; see below).
Viewing E-Gel® with SYBR Safe™
View E-Gel ® with SYBR Safe™ using these instruments:
  • Blue-light transilluminator. The Safe Imager™ Blue Light Transilluminator (available from Invitrogen, S37102) is designed specifically for use with SYBR Safe™ stained DNA gels. Refer to the next section for instructions on using the Safe Imager™ Blue Light Transilluminator. Blue-light transilluminators available from other manufacturers are also compatible for use with E-Gel® with SYBR Safe™.
  • Standard 300 nm UV transilluminator
  • Imaging systems such as laser based scanners equipped with an excitation source in the UV range or between 470-530 nm
Note:   For optimizing cloning efficiency or minimizing UV exposure, we recommend using a blue light transilluminator
Imaging E-Gel® with SYBR Safe™
Photograph E-Gel ® with SYBR Safe™ using Polaroid ® 667 black-and-white print film, or image using a CCD camera or a laser-based scanner.
Required Filters
For photographing gels, use a filter such as SYBR Safe™ photographic filter (S37100), Invitrogen's SYPRO ® photographic filter (S6656) or Kodak ® Wratten #9 filter. A long pass ethidium bromide filter that transmits all light above 500 nm also works.
Do not use ethidium bromide filters that block light above 500 nm for photographing E-Gel® with SYBR Safe™.
Exposure Time and Gain Setting
While yielding similar sensitivities to ethidium bromide, SYBR Safe™ is somewhat dimmer yet with a lower background than ethidium bromide. As a result a slightly longer exposure time, or higher gain setting may be necessary.
TOP
Opening an E-Gel® with SYBR Safe™ Cassette
Introduction
The E-Gel® Opener is an easy-to-use device specifically designed to open any E-Gel® single comb or double comb gels for excision of DNA fragments or for blotting.
Do not use the E-Gel® Opener to open the E-Gel® 48 or 96 cassettes. The E-Gel® 48 or 96 cassette cannot be opened.
Description of Opener
The E-Gel® Opener consists of an anodized aluminum platform housing two recessed steel blades, one which is stationary and one which is movable. The blades are brought into contact with the E-Gel®cassette by turning the large knob clockwise (see Figure 1).
Before using the E-Gel® Opener for the first time, we recommend that you practice opening a few used E-Gels® to familiarize yourself with the process. Practice on E-Gels®that will not be used further for preparative purposes.
The blades on the E-Gel® Opener are extremely sharp. Do not insert your fingers into the area housing the blades! Pick up the E-Gel® Opener by holding the large knob only (see Figure 1 above). Exercise caution when handling and cleaning the E-Gel®Opener. Dispose of blades in a needle disposal container or a Sharps disposal box.
  • Electrophoresis must be complete before opening the E-Gel®. We recommend that you place the E-Gel® on the transilluminator and photograph the gel before proceeding further. If you plan to isolate DNA from the E-Gel®, we recommend that you open the gel and excise the gel fragment immediately after electrophoresis as bands will diffuse within 20 minutes. If you plan to blot the gel, keep your blotting apparatus ready before opening the gel. 
Procedure
The following section provides instructions to open an E-Gel®cassette. Before beginning, you should wear safety goggles and gloves.
  1. Place the E-Gel® Opener on a flat surface, with the knob extending off the edge of the laboratory bench and facing the user. Set the E-Gel® Opener to its widest open position by turning the knob counterclockwise.
  2. Insert the E-Gel® into the E-Gel® Opener so that two opposing sides of the gel cassette are aligned with the blades (see Figure 1). Position the E-Gel® such that the two sides fit into the grooves housing the blades.
  3. Turn the knob steadily clockwise to bring the blades in contact with the E-Gel® cassette. As the knob is tightened, you will hear a series of pops. Continue to turn the knob until the resistance increases. Stop turning the knob as soon as the E-Gel® cassette begins to lift off the surface of the platform. Two sides of the E-Gel®will now be unsealed. Note: Once you observe the E-Gel® cassette begins to lift off the surface of the platform, do not continue to tighten the knob, as you will damage the E-Gel®.
  4. Unscrew the knob and remove the E-Gel®. The E-Gel® cassette fits snugly in the recessed groove, and you may have to carefully work the cassette from the housing. Turn the E-Gel® 90° and re-insert the gel cassette into the Opener so that the two remaining sealed sides can be opened.
  5. Repeat Step 2 to open the remaining two sides of the E-Gel®. Stop turning the knob when you see the top of the E-Gel® cassette begins to lift off the gel.
  6. Unscrew the knob and carefully remove the E-Gel® cassette. The 4 sides of the cassette should be unsealed. If not, repeat Steps 2-5 as necessary. Remove the E-Gel® and set the opened cassette on your bench.
  7. If you plan to blot the gel, do not pick up the gel from the cassette. Lift off the top of the gel cassette. Place the blotting membrane on the gel and pick up the cassette with the gel and membrane. Flip the gel and membrane out of the cassette onto your gloved hand and then flip the gel and the membrane directly onto your wet blotting paper.
If you plan to purify DNA from the gel, lift off the top of the gel cassette and excise the gel fragment. Transfer the gel slice to a microcentrifuge tube.
Cleaning and Storage
After use, clean the E-Gel® Opener with mild detergent and water to remove excess agarose and plastic from the platform. Use a squirt bottle and wipe the platform dry with a clean tissue. Do not insert your fingers into the area housing the blades, and do not immerse the E-Gel® Opener in water as the blades may rust. Store the E-Gel® Opener at room temperature.
Disposal of E-Gel® with SYBR Safe™
SYBR Safe™ DNA gel stain shows no or very low mutagenic activity when tested by an independent, licensed testing laboratory, and this stain is not classified as hazardous waste under U.S. Federal regulations. As disposal regulations vary, please contact your safety office or local municipality for appropriate SYBR Safe disposal in your community.
TOP
Results Using E-Gel® with SYBR Safe™ Agarose Gels
E-Gel® 1.2% with SYBR Safe™
An example of DNA samples run on an E-Gel® 1.2% with SYBR Safe™ is shown below. Samples were loaded in a total volume of 20 µl and visualized on a standard 312 nm UV transilluminator. Photographs were taken using the MiniBis photo documentation system from DNR, and the SYBR Safe™ photographic filter using an exposure time of 1.8 sec.
Lane Sample
1 High DNA Mass Ladder (130 ng)
2 E-Gel® High Range DNA Marker (200 ng)
3 1Kb PCR product (100 ng)
4 3Kb PCR product (200 ng)
5 9Kb PCR product (200 ng)
6 1Kb plus DNA Ladder (500 ng)
7 1Kb plus DNA Ladder (500 ng)
8 pBR322 EcoR1 cut (100 ng)
9 pBR322 uncut (100 ng)
10 pUC19 EcoR1 cut (50ng)
11 E-Gel® High Range DNA Marker (200 ng)
12 High DNA Mass Ladder (130 ng)
E-Gel® 2% with SYBR Safe™
An example of DNA samples run on an E-Gel® 2% with SYBR Safe™ is shown below. Samples were loaded in a total volume of 20 µl and visualized on a standard 312 nm UV transilluminator. Photographs were taken using the MiniBis photo documentation system from DNR, and the SYBR Safe™ photographic filter using an exposure time of 1.8 sec.
TOP
Lane Sample
1 Low DNA Mass Ladder (470 ng)
2 E-Gel® Low Range DNA Marker (350 ng)
3 240bp PCR product (500 ng)
4 317bp PCR product (700 ng)
5 1Kb PCR product (100 ng)
6 100bp DNA Ladder (900 ng)
7 50bp DNA Ladder (700 ng)
8 100bp DNA Ladder (900 ng)
9 pUC19 EcoR1 cut (50 ng)
10 pUC19 uncut (50 ng)
11 E-Gel® Low Range DNA Marker (350 ng)
12 Low DNA Mass Ladder (470 ng)
Examples Using Different Imaging Methods
DNA samples run on an E-Gel® 2% with SYBR Safe™ are shown below, recorded using different imaging methods. Samples were loaded in a total volume of 20 µl and visualized using the indicated transilluminator and filter. Photographs were taken using the MiniBis photo documentation system from DNR.
Standard 312 nm UV transilluminator
Long pass ethidium bromide filter
E-Gel® with SYBR Safe™
Standard 312 nm UV transilluminator
SYBR Safe™ photographic filter
E-Gel® with SYBR Safe™
Blue-light transilluminator
SYBR Safe™ photographic filter
E-Gel® with SYBR™ Safe
Lane Sample
1 Low DNA Mass Ladder (470 ng)
2 E-Gel® Low Range DNA Marker (350 ng)
3 240bp PCR product (500 ng)
4 317bp PCR product (700 ng)
5 1Kb PCR product (100 ng)
6 100bp DNA Ladder (900 ng)
7 50bp DNA Ladder (700 ng)
8 100bp DNA Ladder (900 ng)
9 pUC19 EcoR1 cut (50 ng)
10 pUC19 uncut (50 ng)
11 E-Gel® Low Range DNA Marker (350 ng)
12 Low DNA Mass Ladder (470 ng)
TOP
Troubleshooting
The table below provides solutions to some problems that you may encounter with E-Gel® with SYBR Safe™ agarose gels.
Problem
Cause
Solution
No current
Copper contacts in the base are damaged due to improper use
Make sure the copper contacts in the base are intact.
Expired or defective gel cassette
Use fresh gel cassette. Use properly stored gels before the specified expiration date.
E-Gel ® with SYBR Safe cassette is not inserted properly into a base
Remove cassette and reinsert; a steady red light illuminates on the base when the cassette is correctly inserted and power is on.
Incorrect adaptor used
Use only UL Listed Class 2 Direct Plug-in Adaptor included with the E-Gel ® PowerBase .
Poor resolution or smearing of bands
Sample is overloaded
Do not load more than 200 ng of sample DNA per band.
High salt concentration
Dilute your high-salt samples as described .
Aberrant pre-run step
Be sure to pre-run the gel but do not exceed 2 minutes.
Very low volume of sample loaded or sample was not loaded properly
Avoid introducing bubbles while loading the samples. Bubbles will cause band distortion.
    Load the recommended sample volume based on the gel type and loading method.
    For proper band separation, we recommend keeping sample volumes uniform. Load deionized water or TE into any empty wells
Gel was not electrophoresed immediately after sample loading
For best results, run the gel within 15 minutes of sample loading. If you cannot run the gel immediately after sample loading, use the Two-Step Loading method
Expired gel used
Use properly stored gels before the expiration date.
Longer electrophoresis run time or high current during the run
Longer run times cause an increase in the current, resulting in poor band migration or a melted gel. Do not run the gel longer than recommended time for each gel type.
Melted gel
Increased current due to longer run times
Do not run the gel longer than 40 minutes.
Sample leaking from the wells
Sample is overloaded
Load the recommended sample volume per well.
Use the Two-Step Loading method.
Wells damaged during comb removal
Remove the comb gently without damaging the wells.
Failure Mode indicated by a steady red light and continuous rapid beeping
Defective cassette
Disconnect the base and replace gel  cassette with a fresh gel cassette.
Press and release the power button to return to Ready Mode.
Cold cassette or improper operating conditions
Use a cassette stored at room temperature. Avoid storing gel cassettes at 4°C. Use E-Gel® PowerBase™ and E-Gel® Base at room temperature (20-25°C).
Speckles visible
Dust fluorescing in same wavelength as SYBR-Safe™
Make sure gel is clean before imaging.
High background, suboptimal, or no image
No filters or wrong filter set
Contact the instrument manufacturer for advice.
Photographic settings not optimal
Optimize settings of your system for E-Gel® with SYBR Safe™ empirically. You may need to increase the exposure time or gain setting.
Stripes visible on image
No IR coating on camera when using an UV system
Use IR blocking filter or emission filter with IR coating.
TOP
Filter Selection Guide for E-Gel® with SYBR Safe™
Filter Selection Guide
Use the filter recommended with your instrument below to photograph E-Gel® with SYBR Safe™. We have shown the most popular instruments; other instruments with an excitation source in the UV range or between 470–530 nm may also be used with the proper filter. Contact your instrument manufacturer for advice.
Instrument (Manufacturer)
Excitation Source
Emission Filter
AlphaImager (Alpha Innotech)
302 nm
SYB-500
AlphaImager HP (Alpha Innotech)
302 nm
SYB-500
AlphaDigiDoc RT (Alpha Innotech)
UV transilluminator
Shroud, Camera Stand (Alpha Innotech)
UV transilluminator
SYB-100
DE500 or DE400 light cabinet 2.17” diam.(Alpha Innotech)
UV transilluminator
SYB-500
DE500 or DE400 light cabinet 2” diam. (Alpha Innotech)
UV transilluminator
SYB-400
VersaDoc Imaging Systems (Bio-Rad)
Broadband UV
520LP
Molecular Imager FX Systems (Bio-Rad)
488 nm
530 nm BP
Gel Doc Systems (Bio-Rad)
302 nm
520DF30 (#170-8074)
Typhoon 9400/9410 (GE Healthcare)
488 nm
520 BP 40
Typhoon 9200/9210/8600/8610 (GE Healthcare)
488 nm
526 SP
FluorImager (GE Healthcare)
488 nm
530 DF 30
Storm (GE Healthcare)
Blue (fluorescence mode)
VDS-CL (GE Healthcare)
Transmission
UV Low
Ultracam/Gel Imager (Ultra-Lum)
UV
Yellow Filter (#990-0804-07)
Omega Systems (Ultra-Lum)
UV
520 nm
Polaroid Camera (Polaroid)
UV
SYBR Safe Photographic Filter (S27100)
FOTO/Analyst Express/Investigator/Plus/Luminary (FOTODYNE)
UV
Fluorescent Green (#60-2034)
FOTO/Analyst Minivisionary (FOTODYNE)
UV
Fluorescent Green (#62-4289)
FOTO/Analyst Apprentice (FOTODYNE)
UV
Fluorescent Green (#62-2535)
FOTO/Analyst Luminary (FOTODYNE)
UV
Fluorescent Green (#60-2056)
FCR-10 (Polaroid)
UV
#3-4218
FUJI FLA-3000 (FUJI Film)
473 nm
520LP
BioDocIt/AC1/EC3/BioSpectrum (UVP)
302 nm
SYBR ® Green (#38-0219-01) or SYBR ® Gold (#38-0221-01)
Gel Logic (Kodak)
UV
535 nm WB50
Syngene Instruments (Syngene)
UV
500–600 nm Shortpass filter
TOP
LT074