Quicklinks
Related Product Information
Share |
Introduction
Enrich untouched Dendritic Cells (DCs) by depleting T cells, B cells, monocytes/ macrophages, NK cells, erythrocytes and most granulocytes from blood mononuclear cells with this product. The DC enriched population is bead- and antibody-free and intended for further isolation of any DC subpopulation by flow sorting. This kit provides high recovery of lineage specific markers (Lin-) CD4 + cells and is therefore suitable for further isolation of any DC subpopulation, such as myeloid and plasmacytoid DCs.
Principle of Isolation
Add a mixture of biotinylated monoclonal antibodies against non-DC cells to your starting sample. Add Depletion MyOne™ SA Dynabeads® and allow them to bind to the non-DCs during a short incubation. Separate the beadbound cells with a magnet. Discard the bead-bound cells and use the remaining untouched, enriched cell population for further flow sorting into the DC subpopulation(s) of interest
Description of Materials
Materials Supplied
  • 20 ml Depletion MyOne SA Dynabeads®
Depletion MyOne SA Dynabeads® are uniform, superparamagnetic polystyrene beads (1.0 μm diameter) coated
with streptavidin (SA). Supplied at 10 mg beads per ml in phosphate buffered saline (PBS), pH 7.4, containing 0.1% bovine serum albumin (BSA) and 0.02% sodium azide.
  • 4 ml Antibody Mix for DC kit
The Antibody Mix contains an optimized mixture of biotinylated monoclonal antibodies against CD3, CD14, CD16, CD19, CD56 and Glycophorin A supplied in phosphate buffered saline (PBS), pH 7.4, containing 0.02% sodium azide. The kit will process 2 x 109 mononuclear cells.
Additional Materials Required
  • Magnet: (Dynal® MPC™), MPC-L for 1–5 ml samples, MPC-15 for 1–15 ml samples and MPC-50 for 15–50 ml samples.
  • Mixer allowing both tilting and rotation.
  • Buffer 1: PBS (without Ca2+ and Mg2+) w/0.1% BSA and 2 mM EDTA, pH 7.4.
  • Buffer 2: PBS pH 7.4 (without Ca2+ and Mg2+).
  • Lymphoprep®
Important Notes:
  • It is critical to keep the buffers cold (2–8˚C) during the enrichment steps.
  • PBS containing Ca2+ or Mg2+ is not recommended.
  • Do not use buffers or additives (i.e. FCS) containing biotin since this may reduce efficiency of depletion.
  • Use a mixer that provides tilting and rotation of the tubes to ensure Dynabeads® do not settle at the bottom of the tube.
  • Follow the magnet recommendations to ensure a successful isolation.
TOP
Protocol
Dynabeads® Washing Procedure
Dynabeads® should be washed before use.
  1. Resuspend the Dynabeads® in the vial.
  2. Transfer the desired volume of Dynabeads® to a tube.
  3. Add the same volume of Buffer 1, or at least 1 ml, and mix.
  4. Place the tube in a magnet for 3 min and discard the supernatant.
  5. Remove the tube from the magnet and resuspend the washed Dynabeads® in the same volume of Buffer 1 as the initial volume transferred from the vial in step 2.
Sample Preparation
Preparation of MNC from Buffy Coat to Obtain Low Platelet Numbers
  1.    Dilute 10 – 18 ml buffy coat with Buffer 2 to a total volume of 35 ml at room temperature.
  2.    Add the diluted buffy coat on top of 15 ml of Lymphoprep.
  3.    Centrifuge at 160 x g for 20 minutes at 20°C. Allow to decellerate without brakes.
  4.    Remove 20 ml of supernatant to eliminate platelets.
  5.    Centrifuge at 350 x g for 20 minutes at 20°C. Allow to decellerate without brakes.
  6.    Recover MNC from the plasma/Lymphoprep interface and transfer the cells to a 50 ml tube.
  7.    Wash MNC once with Buffer 1 by centrifugation at 400 x g for 8 minutes at 2–8°C.
  8.    Wash MNC twice with Buffer 1 by centrifugation at 225 x g for 8 minutes at 2–8°C and resuspend the MNC at 1 x 10 8 MNC per ml in Buffer 1.
Enrichment of DCs
  • This protocol is based on enrichment from 1 x 107 leucocytes (MNC). It is scalable from 1 x 107–2 x 109 cells, (see table 1).
  • Keep the cells and buffers cold (2–8°C) during the whole process.
  1. Transfer 100 μl (1 x 107) leucocytes in Buffer 1 to a tube.
  2. Add 20 μl of Antibody Mix.
  3. Mix well and incubate for 20 min at 2–8°C.
  4. Wash the cells by adding 2 ml Buffer 1. Mix well by tilting the tube several times and centrifuge at 300 x g for 10 min at 2–8°C. Discard the supernatant.
  5. Resuspend the cells in 900 μl Buffer 1.
  6. Add 100 μl pre-washed Depletion MyOne SA Dynabeads®.
  7. Incubate for 15 min at 2–8°C with gentle tilting and rotation.
  8. Resuspend the bead-bound cells by thorough pipetting or vortex for 5 secs.
  9. Add 1 ml Buffer 1.
  10. Place the tube in the magnet for 3 min.
  11. Transfer the supernatant to a new tube.
The supernatant contains the DC enriched cell population.
Table 1. Volume requirements for DC enrichment.
  Working volumes Working volumes per 1 x 107 leucocytes Working volumes per 5 x 108 leucocytes
Cell volume (step 1) 100 μl 5 ml
Antibody Mix (step 2) 20 μl 1 ml
Washing (step 4) 2 ml 35 ml
Resuspension (step 5) 900 μl 10 ml
Depletion MyOne SA Dynabeads® (step 6) 100 μl 5 ml
Volume added before magnet (step 9 1 ml 15 ml
Dynal® MPC recommended MPC-L/MPC-15 MPC-50
When working with other cell numbers, scale up all reagents and volumes accordingly.
Downstream Applications
The DC enriched cell population can be used for further isolation of DC subpopulations to high purity using flow sorting.
TOP
General Information
Invitrogen Dynal® AS complies with the Quality System Standards ISO 9001:2000 and ISO 13485:2003.
Storage/Stability
This product is stable until the expiry date stated on the label when stored unopened at 2-8°C. Store opened vials at 2-8°C and avoid bacterial contamination. Keep Dynabeads® in liquid suspension during storage and all handling steps, as drying will result in reduced performance. Resuspend well before use.
Warnings And Limitations
This product is for research use only. Not intended for any animal or human therapeutic or diagnostic use unless otherwise stated. Follow appropriate laboratory guidelines. This product contains 0.02% sodium azide as a preservative, which is cytotoxic.
Avoid pipetting by mouth!
Sodium azide may react with lead and copper plumbing to form highly explosive metal azides. When disposing through plumbing drains, flush with large volumes of water to prevent azide build up. Certificate of Analysis (CoA) is available upon request. Material Safety Data Sheet (MSDS) is available at .
TOP
References
  1.    Wu L et al. (2004) Development of dendritic cell system. Cell Mol Immunol. 1(2):112-8.
  2.    Quah BJ et al. (2005) Maturation of function in dendritic cells for tolerance and immunity. J Cell Mol Med. 9(3):643-54.
  3.    Mellor A. et al. (2005) Indoleamine 2,3 dioxygenase and regulation of T cell immunity. Biochem Biophys Res Commun. 9;338(1):20-4.
  4.    Sheng KC et al. (2005) Dendritic cells: activation and maturation-applications for cancer immunotherapy. Curr Med Chem. 12(15):1783- 800.
  5.    Barchet W et al. (2005) Plasmacytoid dendritic cells-virus experts of innate immunity. Semin Immunol. 17(4):253-61.
  6.    Mahnke K et al. (2005) Dendritic cells: key cells for the induction of regulatory T cells? Curr Top Microbiol Immunol. 293:133-50.
  7.    Palucka AK et al. (2005) Immunotherapy via dendritic cells. Adv Exp Med Biol. 560:105-14.
  8.    Mowat AM.(2005) Dendritic cells and immune responses to orally administered antigens. Vaccine. 7;23(15):1797-9.
  9.    Wu L et al. (2005) Heterogeneity of thymic dendritic cells. Semin Immunol. 2005 Aug;17(4):304-12.
10.    Proietto AI et al. (2004) Differential production of inflammatory chemokines by murine dendritic cell subsets. Immunobiology 209(1-2):163- 72.
11.   Shortman K et al. (2002) Mouse and human dendritic cell subtypes. Nat Rev Immunol. 2(3):151-61.
TOP
113.08D.indd   Rev 001     5-May-2006