Find valuable information.
Optimize your animal health disease detection experiments to get the best results. We’ve compiled a detailed knowledge base of the top tips and tricks to meet your research needs.
View the relevant questions below:
Having problems with your experiment?  Visit our
Troubleshooting page
General
Please see our list of animal health products broken down by application.
Please contact Technical Support at animalhealthandfoodsafety@lifetech.com for further information on how to reach your local sales representative.
ROX is a passive reference dye that is included in many qPCR master mixes. For more details on what it is and its use in qPCR, please check out our short video
Check out this short video to understand the different phases of the PCR reaction and why they are important.
Please view this short video which explains some best practices for replicates and plate setup.
In most cases your instrument software can automatically set the proper threshold for your data. Check out our short video for more information on how to set them.
We recommend running the Xeno Internal Positive Control with our animal health qPCR reagents as a positive control and to help determine whether or not a sample has inhibitors. Many animal health samples will not have a high level or any disease target, so running the Xeno control allows you to be confident that your qPCR reaction worked and that the absence of target signal is due to the true absence of the target instead of a reaction failure. We have Xeno Control Assays labeled with either VIC or LIZ.
We recommend one of the following assays:
  • VetMAX Xeno Internal Positive Control—VIC Assay (Cat. No. A29765)
  • VetMAX Xeno Internal Positive Control—LIZ Assay (Cat. No. A29766).
The VetMAX Xeno Internal Positive Control is derived from a plasmid source.
The quencher used in these assays is an Eclipse Q quencher. Within the qPCR instrument’s software, choose “none”.
We recommend validating this kit by using the VetMAX-Plus One-Step RT-PCR Kit protocol
Generally, we recommend using this master mix with standard cycling conditions, but depending on the assay being used, you could use fast cycling conditions with optimization.