TaqMan® Genotyper Software Automatically Determines Sample Genotypes and Displays Data
TaqMan® Genotyper Software is a great resource for fast and accurate genotype calling. It is a free SNP genotyping data analysis tool for use with TaqMan® SNP Genotyping Assays performed in 48-, 96-, or 384-well microtiter plates or on OpenArray® Plates. It has a state-of-the-art genotype-calling algorithm, an intuitive user interface, and enhanced study-based analysis features. The software enables multi-plate data analysis for high-throughput workflows and improved accuracy in genotype calling; versatile export features and comprehensive quality-control features facilitate streamlining of the entire workflow. TaqMan® Genotyper Software can be downloaded at www.appliedbiosystems.com/taqmangenotyper.