TaqMan® SNAP Assays
Applied Biosystems TaqMan 5'-nuclease assay chemistry provides a fast and simple way to get single nucleotide polymorphism (SNP) genotyping results. Each predesigned TaqMan SNP Genotyping Assay in our massive collection includes two allele-specific TaqMan MGB probes containing distinct fluorescent dyes and a PCR primer pair to detect specific SNP targets. These TaqMan probe and primer sets (assays) uniquely align with the genome to provide unmatched specificity for the allele of interest. With more than 6.2 million predesigned TaqMan SNP Genotyping Assays available for use with 48-, 96-, 384-well and OpenArray real-time PCR instrument formats, you can select the workflow that best fits your genotyping needs.
TaqMan SNP Genotyping User Guide ›
Assay search and ordering tool
TaqMan SNP Genotyping assay formats
View specifications and documentation for TaqMan SNP Genotyping Assays.
Order your TaqMan SNP Genotyping Assays preplated into Fast or standard 96- or 384-well plates with the TaqMan Custom Plating Service. Perfect for analyzing a large number of samples for a few SNPs or evaluating a few samples for a large number of SNPs.
Custom TaqMan Assays for performing genotyping studies with any possible SNP, in any organism.
TaqMan OpenArray Genotyping Plates enable the highest sample throughput for mid-density genotyping: >110,000 genotypes in a single day with the QuantStudio 12K Flex Real-Time PCR System.
It's never been easier to search the more than 4.5 million TaqMan SNP Genotyping Assays with our new Assay Search Tool. Watch this short video to see how easy it is to find the right TaqMan SNP Genotyping Assay.
Resources
Brochure: TaqMan Assays for genetic variation research
Quick reference card: TaqMan Assays for genetic variation
Product bulletin: TaqMan SNP genotyping Assays
User Guide: TaqMan SNP Genotyping Assays
Quality value in TaqMan Genotyper Software
Increasing the call rate in TaqMan Genotyper Software
Support
QuantStudio 12K Flex Real-Time PCR System