Many of the PerkinElmer plate readers including the Envision, ViewLux and VICTOR™ are compatible with Life Technologies assay technologies for TR-FRET, FRET, Fluorescence Intensity (kinetic), and Fluorescence Polarization.
Before using LanthaScreen Terbium and Europium assays
Test your plate reader set-up
These technical notes provide a method for verifying that a fluorescent plate reader is able to detect a change in time-resolved fluorescence energy transfer (TR-FRET) signal, confirming proper instrument set-up and a suitable response. The method is independent of any biological reaction or equilibrium and uses reagents that are on-hand for the LanthaScreen assay.
Download Terbium assay application note
Download Europium assay application note
EnVision Multilabel Reader
Instrument Configuration:
Assay Technology  Adapta &
LanthaScreen
Europium
GeneBLAzer LanthaScreen
Terbium
Predictor™ Z'-LYTE
Modality TR-FRET FRET (bottom) TR-FRET FP FRET
Setup Guide      
Application Note - - - - -
Poster - - Lab Auto 08 - -
Excitation Filter (15 mm) 340/30
(PV00215)
400/25 340/30
(PV00215)
531/25 FP 400/25
Emission 1 Filter (15 mm) 615/8.5 460/25 495/10
(PV00315)
579/25 FP P-pol 460/25
Emission 2 Filter (15 mm) 665/7.5 535/25 520/25
(PV00315)
579 FP S-pol 535/25
Dichroic Mirror D400/630 TRF Dual or D400 TRF D425/495 BLA Dual or D425 BLA; 50/50 General Dual or 50/50 General D400/630 TRF Dual or D400 TRF; D400/505 LanthaScreen Dual D555fp/D595 Dual or D555fp D425/495 BLA Dual or D425 BLA; 50/50 General Dual or 50/50 General
VICTOR™ Multilabel Plate Readers
Instrument Configuration:
Assay Technology  Adapta &
LanthaScreen
Europium
GeneBLAzer LanthaScreen
Terbium
Predictor™ Z'-LYTE
Modality TR-FRET FRET (bottom) TR-FRET FP FRET
Setup Guide Reader compatible but settings not validated Reader not
compatible
  Reader not
compatible
Reader not
tested
Application Note - - - - -
Poster - - - - -
Excitation Filter (15 mm) 340/60
(built-in)
- 340/60
(built-in)
- NA
Emission 1 Filter (25 mm) 616/8.5 - 495/10
(PV00325)
- NA
Emission 2 Filter (25 mm) 665/7.5 - 520/25
(PV00325)
- NA
Instrument Configuration:
Assay Technology  Adapta &
LanthaScreen
Europium
GeneBLAzer LanthaScreen
Terbium
Predictor™ Z'-LYTE
Modality TR-FRET FRET (bottom) TR-FRET N/A N/A
Setup Guide Reader compatible but settings not validated Reader not
tested
  Reader not
tested
Reader not
tested
Application Note - - - - -
Poster - - Lab Auto 08 - -
Excitation Filter (60 mm) 340/60 NA 340/60 NA NA
Emission 1 Filter (60 mm) 616/8 NA 495/10
(Chroma)
NA NA
Emission 2 Filter (60 mm) 671/7.8 NA 520/20 NA NA
Dichroic Mirror 400 NA 400 NA NA
* Filters (diameter) shown as center wavelength/bandwidth (nm) & dichroic mirrors shown as cutoff wavelength (nm); Contact PerkinElmer for information on ordering filters & dichroic mirrors suitable with the microplate readers listed above
** Some compatible filter sets are available for purchase from Life Technologies.  Click on selected filter sets for more information.
Envision and ViewLux are registered trademarks and Victor™ is a trademark of PerkinElmer.