AgeI nucleotide recognition sequence
Figure color legend
Name derivation: Agrobacterium gelatinovorum
Recognition and Cutting site: A^CCGG T
T GGCC^A
Resulting DNA Ends: 5′ protruding ends
Prototype: AgeI
Isoschizomers: BshTI, AsiGI, CspAI, PinAI
Sensitivity to methylation:  CpG blocked
Neoschizomers None
Anza RE generating compatible ends Anza™ 60 Kpn2I, Anza™ 85 MreI, Anza™ 90 Eco88I* (*if recognized sequence is 5'-CCCGGG-3')
References