Water-Jacketed CO2 Incubator
Enhanced temperature stability
Thermo Scientific water jacketed CO2 incubators deliver dependable thermal protection from swings in ambient temperature and power variations. This traditional CO2 incubator is the benchmark for reliability and uniformity.
Contact us
Quick links
Choose a water jacketed CO2 incubator
Unique triple wall construction provides outstanding temperature stability supplied by dual layers of water and high-quality insulation.
  Series 2 Water Jacketed Series 3 Water Jacketed
  Forma series 2 co2 incubator Forma series 3 co2 incubator
Description Proven benchmark for reliability Smart simplicity added to proven reliability
Capacity 184L (6.5 cu. ft.) 184L (6.5 cu. ft.)
Chamber type Water jacketed Water jacketed
Door swing  Left hinged  Left hinged
Interior configuration choices Stainless steel › Stainless steel ›
HEPA filtration ISO Class 5 in 5 min. ISO Class 5 in 5 min.
Airflow design Fan assisted Fan assisted
CO2 sensors TC or IR TC or IR
O2 control Optional 1–21%
Optional 1–21%
Humidity Removable water pan Removable water pan
User interface Enviro-Scan iCAN touchscreen
Removable water pan
Top
Performance after unexpected power loss
The triple-wall construction of our water jacketed CO2 incubators offers outstanding temperature stability during a power disruption. Sixty minutes after power loss, the Forma Water Jacketed CO2 Incubator dropped only 1°C, maintaining the chamber’s specified growing environment.
Key features to help you achieve your next discovery
Based on 60 years of engineering and design breakthroughs, our expertise is represented in key features that span our portfolio. Together these enhancements provide optimal cell growth, valuable contamination control technologies, and advanced design for highly critical applications, plus simplicity which allows you to spend more time pursuing your goals.
Connectivity, including the important benefits of remote monitoring, are enhancing workflows and increasing efficiency and productivity within the lab. Thermo Scientific™ DeviceLink HUB™, our easy-to-install solution, works with the Thermo Fisher Cloud to monitor conditions inside your CO2 incubators.
  • DeviceLink HUB monitors chamber temperature, gas levels, alarms, and alerts from the incubator data output connection.
  • Alarms and notifications are also sent to the user by email and mobile push notification.
  • DeviceLink is an optional accessory for the following models with the iCAN touchscreen interface:
    • Heracell 150i and 240i CO2 incubators
    • Forma Series 3 Water Jacket CO2 incubators
    • Heracell VIOS 160i and 250i CO2 incubators
    • Forma Steri-cycle i160 and i250 CO2 incubators
  • Device Link is available in the following regions:
    • United States and Canada
    • 28 European Union countries
    • Iceland, Liechtenstein, Norway, and Switzerland
HEPA air filtration designed for air purity in-chamber.
  • HEPA continuously filters the entire chamber every 60 seconds
  • Protects cultures against airborne contaminants
  • Achieves cleanroom quality ISO Class 5 (Class 100) air within 5 minutes of routine door opening
Thermo Scientific CO2 incubators feature fan-assisted airflow circulation for fast recovery and tight uniformity. Our airflow patterns are specifically designed for homogeneous distribution of critical environmental conditions (temperature, gas exchange and humidity). The in-chamber fan gently distributes the conditioned, humidified air throughout the chamber, ensuring all cells—regardless of location—experience the same conditions without risk of desiccation.1
  • Temperature uniformity specifications are validated according to the DIN 12880 standard, which requires 27 individual measurements at specific locations in the chamber.
  • Sensors and probes are positioned directly in the incubator chamber to ensure they measure the exact environment experienced by your cells.
  • A secondary temperature probe acts as an alarm to the primary sensor, providing peace of mind for you and extra security for your cells.
See THRIVE active airflow in action
Thermo Scientific THRIVE active airflow, included on our Heracell VIOS 160i and 250i and Forma Steri-Cycle i160 and i250 CO2 incubators, provides total recovery of critical growth parameters in 10 minutes or less following a 30-second door opening.1
  • The variable-speed fan precisely controls the airflow to provide leading-edge recovery.
  • The air stream first travels over the direct-heated covered, integrated water reservoir, which results in 50% faster humidity recovery than a standard water pan design.
  • The in-line HEPA filter cleans the airstream of all sizes of particles and microorganisms, protecting cultures from contamination.
  • The air then passes over the in-chamber sensors, which react and condition the air before gently circulating it among the cultures.
1 Based on internal standards for 30-second door opening, recovery calculated to 98% of starting value for temperature and CO2 , and 95% of starting value of humidity.
Because the goal is to mimic in vivo conditions, all incubator sensors are positioned directly in the incubation chamber to ensure they measure the exact environment experienced by your cells. This design requires more robust sensor construction but means the sensors are designed to react immediately to correct temperature and atmosphere variations as they occur. We offer two different CO2 sensor technologies.
Thermal conductivity (TC) CO2 sensor
  • Our most commonly requested CO2 sensor due to its reliability and longevity of operation
  • Measures electrical resistance according to thermal conductivity of gas passing over the sensor
  • Low maintenance, stable, and long life because it has no consuming element such as an incandescent bulb
  • High temperature resistance—does not need to be removed during sterilization cycles
  • NEW TC180 CO2 sensor incorporates humidity compensation for enhanced stability and recovery (available in the Heracell VIOS and Forma Steri-Cycle CO2 incubators)
Infrared (IR) CO2 sensor
  • Ideal for sensitive applications and remote monitoring or when frequent access to incubator is required
  • Determines the CO2 concentration based on its IR light absorption at the characteristic wavelength of 4.3 µm
  • Operates independent of humidity and temperature
  • NEW IR180Si replaces the traditional incandescent IR light source with silicone MEMS emitter technology for improved stability and service life (available in the Heracell VIOS and Forma Steri-Cycle CO2 incubators)
Added culturing flexibility with variable oxygen control (tri-gas)
Increase biological relevance by simulating physiological conditions, resulting in cell behaviors that are more predictive of an in vivo environment.
  • Choice of  1–21% and 5–90% O2 ranges
  • Simulate hypoxic (1–21%) environments for primary cell, stem cell, and embryo research applications
  • Achieve better simulation of tumor microenviroments with low oxygen culturing
  • Simulate hyperoxic (5–90%) conditions for research in lung, retina, and other sensitive tissues
Cells cultured in low oxygen (hypoxia) will generally grow faster, live longer, and show lower stress.
Featured resource
Five reasons to enhance your cell culture using a Thermo Scientific CO2 incubator with variable oxygen control
Learn more
A selection of humidity reservoir designs are available to enhance performance and workflows. Our unique integrated solutions are directly heated to facilitate high relative humidity (rH) levels and faster recovery. An external humidity reservoir simplifies maintenance by allowing water refill without disrupting culturing. Full active rH parameter control is ideal for applications requiring flexibility and precise monitoring of humidity levels.
  Water pan Integrated water reservoir Covered integrated water reservoir External humidity reservoir
Description Traditional removable humidity pan Large directly heated panless reservoir provides high rH levels and fast recovery Protected, directly heated integrated water pan provides high rH levels with leading recovery and easy filling and draining Active humidity control with an external reservoir for easy replenishment of water
Volume 3L 3L (4.5L for large capacity models) 3L 3L
Water level monitoring   Yes, low level alarm Yes, visual monitoring and low level alarm Yes, visual monitoring and low level alarm
Popular models Forma Direct Heat and Steri-Cycle; Forma Series 2 and Series 3 Water Jacketed; Midi 40
Heracell 150i and 240i
Heracell VIOS 160i and 250i; Forma Steri-cycle i160 and i250
Forma Steri-cult; BBD 6220
iCAN touchscreen interface
The intelligent Thermo Scientific iCAN touchscreen interface is designed to simplify your interactions with the CO2 incubator. The bright LED screen provides at-a-glance monitoring even from across the lab. Enjoy complete data visibility and performance trend graphing, in multiple languages. Store data and usage logs onboard or download.
  • Door mounted position for easy access
  • Configurable settings and options
  • Audio and visual alarms
  • On-screen menu prompts
  • Error and usage logs
  • Data logging and performance trend graphing
  • Multiple language selection
Enviro-Scan interface
The Thermo Scientific Enviro-Scan LED membrane display lets you easily monitor temperature, CO2, and rH (optional) operating parameters via the bright display. The alphanumeric message center provides process and alarm status.
  • Configurable settings and options
  • Audio and visual alarms
  • On-screen menu prompts
Top
CO2 incubator selector tool
With so many choices, which Thermo Scientific CO2 incubator should you choose? Based on your cell culture applications, needs, or preferences, our selector can help you decide on the best incubator for your lab.
Go to selector tool
CO2 incubator family brochure
See the full portfolio of Thermo Scientific CO2 incubators and discover why more scientists trust us with their valuable cultures.
Download brochure
Top
Free webinar: cell culture contamination
Topics covered:
  • Varied types of contamination and the extent of the problem
  • Reducing contamination in your laboratory
  • How to evaluate CO2 incubator design innovations for contamination prevention
Listen to our on-demand webinar ›
Check out our Cell Culture Café webinar series, subscribe to informative publications, blogs and more, and search our on-demand webinars library.
Visit Cell Culture Café
Top
Related product categories
CO2 Resistant Shakers
Biological Safety Cabinets
General Purpose Centrifuges
Cell Culture Flasks
Top
Resources
Brochures
Technical notes
Smart notes
Support