Flammable Material Freezers
Maximum safety for flammable materials
Our flammable material freezers meet NFPA and OSHA standards to help ensure maximum safety for storing flammable and volatile materials. All our flammable-material, UL-listed freezers feature CFC-free, foamed-in-place cabinet insulation, hermetically-sealed refrigeration compressors, and adjustable temperature controls.
Popular flammable material freezers
Featured flammable material freezer categories
Reduce risk of accidental explosion and associated sample damage with Thermo Scientific flammable materials storage laboratory freezers. These freezers are built with no internal electrical components that could trigger an explosion and are ideal for storing potentially flammable material in general laboratory settings.
Thermo Scientific high-performance flammable materials freezers meet stringent U.S. Pharmacopeia standards for drug storage and NFPA safety provisions for storing flammable liquids. Store flammable materials, solvents, and intermediates within a tight temperature range.