Minus 40 Lab Freezers
Receive uncompromised sample protection for -10°C to -40°C applications with our -40°C lab freezers. Rugged design for daily use and dependability, available in both chest and upright models.
  • Rugged construction for everyday use
  • Microprocessor control
  • Optional chart recorder
  • Easy-to-remove washable filter
Use our ULT selection guide, browse through our portfolio below, or ask one of our experts to help tailor a solution specific to your needs.
ULT selection guide
Quick links
Minus 40 chest freezer
Versatile and reliable sample storage for applications requiring a temperature range from -10°C to -40°C, these standard chest freezers come in a variety of sizes and offer a full range of racking solutions. View chest freezers in a 2-inch box capacity from 63 to 396 boxes to fit your needs.
Minus 40 upright freezer
Multiple sizes accommodate your space, and they feature an eye level, easy-to-use microprocessor control panel. Triple-point door gasket provides longer holding time with door openings, and four inner doors reduce cold air loss while improving temperature recovery. Choose a 2-inch box capacity from 240 to 400 boxes.
-40°C chest freezers 63 to 396 box capacity
  63 Boxes* 252 Boxes 348 Boxes 396 Boxes
  63 box Forma -40 freezer 396 box Forma -40 freezer
Request a Quote
Capacity 3 cu. ft. (85L) 12.7 cu. ft. (360L) 17 cu. ft. (481L) 20 cu. ft. (566L)
2mL Vial Capacity 6,300 25,200 34,800 39,600
Interior (DxWxH) 18.5 x 18.5 x 16.5 in.
47.0 x 47.0 x 41.9cm
42.4 x 18.5 x 27.8 in.
107.7 x 47.0 x 70.6cm
58.5 x 18.5 x 27.8 in.
147.3 x 47.0 x 70.6cm
66.4 x 18.5 x 27.8 in.
168.7 x 47.0 x 70.6cm
Exterior (LxWxH) 32.3 x 28.7 x 43.8 in.
72.9 x 82.0 x 111.3cm
72.0 x 33.4 x 40.6 in.
182.9 x 84.8 x 103.1cm
87.6 x 33.8 x 40.5 in.
222.5 x 85.9 x 102.9cm
90.0 x 33.4 x 40.6 in.
243.8 x 84.8 x 103.1cm
Shipping Weight 395 lbs. (179kg) 659 lbs. (299kg) 764 lbs. (347kg) 776 lbs. (352kg)
  63 Boxes* 252 Boxes 348 Boxes 396 Boxes
  63 box Revco ULT freezer 252 box Revco ULT freezer 348 box Revco ULT freezer 396 box Revco ULT freezer
Request a Quote
Capacity 3 cu. ft. (85L) 12.7 cu. ft. (360L) 17 cu. ft. (481L) 20 cu. ft. (566L)
2mL Vial Capacity 6,300 25,200 34,800 39,600
Interior (DxWxH) 18.5 x 18.5 x 16.5 in.
47.0 x 47.0 x 41.9cm
42.4 x 18.5 x 27.8 in.
107.7 x 47.0 x 70.6cm
58.5 x 18.5 x 27.8 in.
147.3 x 47.0 x 70.6cm
66.4 x 18.5 x 27.8 in.
168.7 x 47.0 x 70.6cm
Exterior (LxWxH) 32.3 x 28.7 x 43.8 in.
72.9 x 82.0 x 111.3cm
72.0 x 33.4 x 40.6 in.
182.9 x 84.8 x 103.1cm
87.6 x 33.8 x 40.5 in.
222.5 x 85.9 x 102.9cm
90.0 x 33.4 x 40.6 in.
243.8 x 84.8 x 103.1cm
Shipping Weight 395 lbs. (179kg) 659 lbs. (299kg) 764 lbs. (347kg) 776 lbs. (352kg)
  63 Boxes* 252 Boxes 348 Boxes 396 Boxes
  63 box TS ULT freezer 252 box TS ULT freezer 348 box TS ULT freezer 396 box TS ULT freezer
Request a Quote
Capacity 3 cu. ft. (85L) 12.7 cu. ft. (360L) 17 cu. ft. (481L) 20 cu. ft. (566L)
2mL Vial Capacity 6,300 25,200 34,800 39,600
Interior (DxWxH) 18.5 x 18.5 x 16.5 in.
47.0 x 47.0 x 41.9cm
42.4 x 18.5 x 27.8 in.
107.7 x 47.0 x 70.6cm
58.5 x 18.5 x 27.8 in.
147.3 x 47.0 x 70.6cm
66.4 x 18.5 x 27.8 in.
168.7 x 47.0 x 70.6cm
Exterior (LxWxH) 32.3 x 28.7 x 43.8 in.
72.9 x 82.0 x 111.3cm
72.0 x 33.4 x 40.6 in.
182.9 x 84.8 x 103.1cm
87.6 x 33.8 x 40.5 in.
222.5 x 85.9 x 102.9cm
90.0 x 33.4 x 40.6 in.
243.8 x 84.8 x 103.1cm
Shipping Weight 395 lbs. (179kg) 659 lbs. (299kg) 764 lbs. (347kg) 776 lbs. (352kg)
  63 Boxes* 252 Boxes 348 Boxes 396 Boxes
  63 box HERAfreeze ULT freezer 252 box HERAfreeze ULT freezer 348 box HERAfreeze ULT freezer 396 box HERAfreeze ULT freezer
Request a Quote
Capacity 3 cu. ft. (85L) 12.7 cu. ft. (360L) 17 cu. ft. (481L) 20 cu. ft. (566L)
2mL Vial Capacity 6,300 25,200 34,800 39,600
Interior (DxWxH) 18.5 x 18.5 x 16.5 in.
47.0 x 47.0 x 41.9cm
42.4 x 18.5 x 27.8 in.
107.7 x 47.0 x 70.6cm
58.5 x 18.5 x 27.8 in.
147.3 x 47.0 x 70.6cm
66.4 x 18.5 x 27.8 in.
168.7 x 47.0 x 70.6cm
Exterior (LxWxH) 32.3 x 28.7 x 43.8 in.
72.9 x 82.0 x 111.3cm
72.0 x 33.4 x 40.6 in.
182.9 x 84.8 x 103.1cm
87.6 x 33.8 x 40.5 in.
222.5 x 85.9 x 102.9cm
90.0 x 33.4 x 40.6 in.
243.8 x 84.8 x 103.1cm
Shipping Weight 395 lbs. (179kg) 659 lbs. (299kg) 764 lbs. (347kg) 776 lbs. (352kg)
  63 Boxes* 252 Boxes 348 Boxes 396 Boxes
  63 box Forma -40 freezer 396 box Forma -40 freezer
Request a Quote
Capacity 3 cu. ft. (85L) 12.7 cu. ft. (360L) 17 cu. ft. (481L) 20 cu. ft. (566L)
2mL Vial Capacity 6,300 25,200 34,800 39,600
Interior (DxWxH) 18.5 x 18.5 x 16.5 in.
47.0 x 47.0 x 41.9cm
42.4 x 18.5 x 27.8 in.
107.7 x 47.0 x 70.6cm
58.5 x 18.5 x 27.8 in.
147.3 x 47.0 x 70.6cm
66.4 x 18.5 x 27.8 in.
168.7 x 47.0 x 70.6cm
Exterior (LxWxH) 32.3 x 28.7 x 43.8 in.
72.9 x 82.0 x 111.3cm
72.0 x 33.4 x 40.6 in.
182.9 x 84.8 x 103.1cm
87.6 x 33.8 x 40.5 in.
222.5 x 85.9 x 102.9cm
90.0 x 33.4 x 40.6 in.
243.8 x 84.8 x 103.1cm
Shipping Weight 395 lbs. (179kg) 659 lbs. (299kg) 764 lbs. (347kg) 776 lbs. (352kg)
  63 Boxes* 252 Boxes 348 Boxes 396 Boxes
  63 box Revco ULT freezer 252 box Revco ULT freezer 348 box Revco ULT freezer 396 box Revco ULT freezer
Request a Quote
Capacity 3 cu. ft. (85L) 12.7 cu. ft. (360L) 17 cu. ft. (481L) 20 cu. ft. (566L)
2mL Vial Capacity 6,300 25,200 34,800 39,600
Interior (DxWxH) 18.5 x 18.5 x 16.5 in.
47.0 x 47.0 x 41.9cm
42.4 x 18.5 x 27.8 in.
107.7 x 47.0 x 70.6cm
58.5 x 18.5 x 27.8 in.
147.3 x 47.0 x 70.6cm
66.4 x 18.5 x 27.8 in.
168.7 x 47.0 x 70.6cm
Exterior (LxWxH) 32.3 x 28.7 x 43.8 in.
72.9 x 82.0 x 111.3cm
72.0 x 33.4 x 40.6 in.
182.9 x 84.8 x 103.1cm
87.6 x 33.8 x 40.5 in.
222.5 x 85.9 x 102.9cm
90.0 x 33.4 x 40.6 in.
243.8 x 84.8 x 103.1cm
Shipping Weight 395 lbs. (179kg) 659 lbs. (299kg) 764 lbs. (347kg) 776 lbs. (352kg)
  63 Boxes* 252 Boxes 348 Boxes 396 Boxes
  63 box TS ULT freezer 252 box TS ULT freezer 348 box TS ULT freezer 396 box TS ULT freezer
Request a Quote
Capacity 3 cu. ft. (85L) 12.7 cu. ft. (360L) 17 cu. ft. (481L) 20 cu. ft. (566L)
2mL Vial Capacity 6,300 25,200 34,800 39,600
Interior (DxWxH) 18.5 x 18.5 x 16.5 in.
47.0 x 47.0 x 41.9cm
42.4 x 18.5 x 27.8 in.
107.7 x 47.0 x 70.6cm
58.5 x 18.5 x 27.8 in.
147.3 x 47.0 x 70.6cm
66.4 x 18.5 x 27.8 in.
168.7 x 47.0 x 70.6cm
Exterior (LxWxH) 32.3 x 28.7 x 43.8 in.
72.9 x 82.0 x 111.3cm
72.0 x 33.4 x 40.6 in.
182.9 x 84.8 x 103.1cm
87.6 x 33.8 x 40.5 in.
222.5 x 85.9 x 102.9cm
90.0 x 33.4 x 40.6 in.
243.8 x 84.8 x 103.1cm
Shipping Weight 395 lbs. (179kg) 659 lbs. (299kg) 764 lbs. (347kg) 776 lbs. (352kg)
  63 Boxes* 252 Boxes 348 Boxes 396 Boxes
  63 box HERAfreeze ULT freezer 252 box HERAfreeze ULT freezer 348 box HERAfreeze ULT freezer 396 box HERAfreeze ULT freezer
Request a Quote
Capacity 3 cu. ft. (85L) 12.7 cu. ft. (360L) 17 cu. ft. (481L) 20 cu. ft. (566L)
2mL Vial Capacity 6,300 25,200 34,800 39,600
Interior (DxWxH) 18.5 x 18.5 x 16.5 in.
47.0 x 47.0 x 41.9cm
42.4 x 18.5 x 27.8 in.
107.7 x 47.0 x 70.6cm
58.5 x 18.5 x 27.8 in.
147.3 x 47.0 x 70.6cm
66.4 x 18.5 x 27.8 in.
168.7 x 47.0 x 70.6cm
Exterior (LxWxH) 32.3 x 28.7 x 43.8 in.
72.9 x 82.0 x 111.3cm
72.0 x 33.4 x 40.6 in.
182.9 x 84.8 x 103.1cm
87.6 x 33.8 x 40.5 in.
222.5 x 85.9 x 102.9cm
90.0 x 33.4 x 40.6 in.
243.8 x 84.8 x 103.1cm
Shipping Weight 395 lbs. (179kg) 659 lbs. (299kg) 764 lbs. (347kg) 776 lbs. (352kg)
*Box capacity is in terms of 2-inch (5cm) high boxes.
Top
-40°C upright freezers with 240 to 400 box capacity
  240 Boxes* 320 Boxes 400 Boxes
  240 box Forma -40 freezer 320 box Forma -40 freezer 400 box Forma -40 freezer
Request a Quote
Capacity 13 cu. ft. (368L) 17.3 cu. ft. (490L) 23 cu. ft. (651L)
2mL Vial Capacity 21,600 32,000 40,000
Interior (DxWxH) 22.5 x 19.4 x 51.5 in.
(57.2 x 49.3 x 130.8cm)
22.5 x 25.4 x 51.5 in.
(57.2 x 64.5 x 130.8cm)
30.0 x 25.4 x 51.5 in.
(76.2 x 64.5 x 130.8cm)
Exterior (LxWxH) 33.3 x 31.6 x 77.9 in.
(84.6 x 80.3 x 197.9cm)
33.3 x 37.5 x 77.9 in.
(84.6 x 95.3 x 197.9cm)
40.8 x 37.5 x 77.9 in.
(103.6 x 95.3 x 197.9cm)
Shipping Weight 696 lbs. (316kg) 795 lbs. (361kg) 839 lbs. (381kg)
  240 Boxes* 320 Boxes 400 Boxes
  240 box Revco -40 freezer 320 box Revco -40 freezer 400 box Revco -40 freezer
Request a Quote
Capacity 13 cu. ft. (368L) 17.3 cu. ft. (490L) 23 cu. ft. (651L)
2mL Vial Capacity 21,600 32,000 40,000
Interior (DxWxH) 22.5 x 19.4 x 51.5 in.
(57.2 x 49.3 x 130.8cm)
22.5 x 25.4 x 51.5 in.
(57.2 x 64.5 x 130.8cm)
30.0 x 25.4 x 51.5 in.
(76.2 x 64.5 x 130.8cm)
Exterior (LxWxH) 33.3 x 31.6 x 77.9 in.
(84.6 x 80.3 x 197.9cm)
33.3 x 37.5 x 77.9 in.
(84.6 x 95.3 x 197.9cm)
40.8 x 37.5 x 77.9 in.
(103.6 x 95.3 x 197.9cm)
Shipping Weight 696 lbs. (316kg) 795 lbs. (361kg) 839 lbs. (381kg)
  240 Boxes* 320 Boxes 400 Boxes
  240 box TS -40 freezer 320 box TS -40 freezer 400 box TS -40 freezer
Request a Quote
Capacity 13 cu. ft. (368L) 17.3 cu. ft. (490L) 23 cu. ft. (651L)
2mL Vial Capacity 21,600 32,000 40,000
Interior (DxWxH) 22.5 x 19.4 x 51.5 in.
(57.2 x 49.3 x 130.8cm)
22.5 x 25.4 x 51.5 in.
(57.2 x 64.5 x 130.8cm)
30.0 x 25.4 x 51.5 in.
(76.2 x 64.5 x 130.8cm)
Exterior (LxWxH) 33.3 x 31.6 x 77.9 in.
(84.6 x 80.3 x 197.9cm)
33.3 x 37.5 x 77.9 in.
(84.6 x 95.3 x 197.9cm)
40.8 x 37.5 x 77.9 in.
(103.6 x 95.3 x 197.9cm)
Shipping Weight 696 lbs. (316kg) 795 lbs. (361kg) 839 lbs. (381kg)
  240 Boxes* 320 Boxes 400 Boxes
  240 box HERAfreeze -40 freezer 320 box HERAfreeze -40 freezer 400 box HERAfreeze -40 freezer
Request a Quote
Capacity 13 cu. ft. (368L) 17.3 cu. ft. (490L) 23 cu. ft. (651L)
2mL Vial Capacity 21,600 32,000 40,000
Interior (DxWxH) 22.5 x 19.4 x 51.5 in.
(57.2 x 49.3 x 130.8cm)
22.5 x 25.4 x 51.5 in.
(57.2 x 64.5 x 130.8cm)
30.0 x 25.4 x 51.5 in.
(76.2 x 64.5 x 130.8cm)
Exterior (LxWxH) 33.3 x 31.6 x 77.9 in.
(84.6 x 80.3 x 197.9cm)
33.3 x 37.5 x 77.9 in.
(84.6 x 95.3 x 197.9cm)
40.8 x 37.5 x 77.9 in.
(103.6 x 95.3 x 197.9cm)
Shipping Weight 696 lbs. (316kg) 795 lbs. (361kg) 839 lbs. (381kg)
  240 Boxes* 320 Boxes 400 Boxes
  240 box Forma -40 freezer 320 box Forma -40 freezer 400 box Forma -40 freezer
Request a Quote
Capacity 13 cu. ft. (368L) 17.3 cu. ft. (490L) 23 cu. ft. (651L)
2mL Vial Capacity 21,600 32,000 40,000
Interior (DxWxH) 22.5 x 19.4 x 51.5 in.
(57.2 x 49.3 x 130.8cm)
22.5 x 25.4 x 51.5 in.
(57.2 x 64.5 x 130.8cm)
30.0 x 25.4 x 51.5 in.
(76.2 x 64.5 x 130.8cm)
Exterior (LxWxH) 33.3 x 31.6 x 77.9 in.
(84.6 x 80.3 x 197.9cm)
33.3 x 37.5 x 77.9 in.
(84.6 x 95.3 x 197.9cm)
40.8 x 37.5 x 77.9 in.
(103.6 x 95.3 x 197.9cm)
Shipping Weight 696 lbs. (316kg) 795 lbs. (361kg) 839 lbs. (381kg)
  240 Boxes* 320 Boxes 400 Boxes
  240 box Revco -40 freezer 320 box Revco -40 freezer 400 box Revco -40 freezer
Request a Quote
Capacity 13 cu. ft. (368L) 17.3 cu. ft. (490L) 23 cu. ft. (651L)
2mL Vial Capacity 21,600 32,000 40,000
Interior (DxWxH) 22.5 x 19.4 x 51.5 in.
(57.2 x 49.3 x 130.8cm)
22.5 x 25.4 x 51.5 in.
(57.2 x 64.5 x 130.8cm)
30.0 x 25.4 x 51.5 in.
(76.2 x 64.5 x 130.8cm)
Exterior (LxWxH) 33.3 x 31.6 x 77.9 in.
(84.6 x 80.3 x 197.9cm)
33.3 x 37.5 x 77.9 in.
(84.6 x 95.3 x 197.9cm)
40.8 x 37.5 x 77.9 in.
(103.6 x 95.3 x 197.9cm)
Shipping Weight 696 lbs. (316kg) 795 lbs. (361kg) 839 lbs. (381kg)
  240 Boxes* 320 Boxes 400 Boxes
  240 box TS -40 freezer 320 box TS -40 freezer 400 box TS -40 freezer
Request a Quote
Capacity 13 cu. ft. (368L) 17.3 cu. ft. (490L) 23 cu. ft. (651L)
2mL Vial Capacity 21,600 32,000 40,000
Interior (DxWxH) 22.5 x 19.4 x 51.5 in.
(57.2 x 49.3 x 130.8cm)
22.5 x 25.4 x 51.5 in.
(57.2 x 64.5 x 130.8cm)
30.0 x 25.4 x 51.5 in.
(76.2 x 64.5 x 130.8cm)
Exterior (LxWxH) 33.3 x 31.6 x 77.9 in.
(84.6 x 80.3 x 197.9cm)
33.3 x 37.5 x 77.9 in.
(84.6 x 95.3 x 197.9cm)
40.8 x 37.5 x 77.9 in.
(103.6 x 95.3 x 197.9cm)
Shipping Weight 696 lbs. (316kg) 795 lbs. (361kg) 839 lbs. (381kg)
  240 Boxes* 320 Boxes 400 Boxes
  240 box HERAfreeze -40 freezer 320 box HERAfreeze -40 freezer 400 box HERAfreeze -40 freezer
Request a Quote
Capacity 13 cu. ft. (368L) 17.3 cu. ft. (490L) 23 cu. ft. (651L)
2mL Vial Capacity 21,600 32,000 40,000
Interior (DxWxH) 22.5 x 19.4 x 51.5 in.
(57.2 x 49.3 x 130.8cm)
22.5 x 25.4 x 51.5 in.
(57.2 x 64.5 x 130.8cm)
30.0 x 25.4 x 51.5 in.
(76.2 x 64.5 x 130.8cm)
Exterior (LxWxH) 33.3 x 31.6 x 77.9 in.
(84.6 x 80.3 x 197.9cm)
33.3 x 37.5 x 77.9 in.
(84.6 x 95.3 x 197.9cm)
40.8 x 37.5 x 77.9 in.
(103.6 x 95.3 x 197.9cm)
Shipping Weight 696 lbs. (316kg) 795 lbs. (361kg) 839 lbs. (381kg)
*Box capacity is in terms of 2-inch (5cm) high boxes.
Top
Which plug type do I need?
  Standard Plug Type Voltage Hz
NEMA 5-15 115–120 60
NEMA 5-20 115–120 60
NEMA 6-15 208–230 60
Continental EU
230 50
Top
Monitor your sample storage conditions 24/7
Smart-vue-monitor-mobile-application-278x195
In the event of a power or mechanical issue, you need to be notified right away. Our wireless monitoring systems are designed for 24/7 monitoring of your ultra-low freezer's performance along with reporting, remote alarm notification and continuous data collection.
View our wireless monitoring solutions ›
Top
Cryogenic storage
Matrix storage (barcoded tubes)
Cryo extra sample prep
Standard storage tubes
Racks, boxes and accessories
Freezer racks and boxes
How-to freezer videos
Top