vacuum-concentrators-kits-showcase-image-230x195-Recovered
Work with speed and precision in pursuit of new discoveries
Samples are precious and so is time. Our vacuum concentrators use state-of-the-art centrifugation, vacuum, and heat technologies for removing solvents and concentrating samples while maintaining sample integrity.
Selector guide for vacuum concentrators
There are several things to consider when choosing your SpeedVac system. Selecting the correct system is crucial for optimum performance and reliability.
Find your system
Popular vacuum concentrators and kits
Savant™ SpeedVac™ High Capacity Concentrators
Savant SPD1010 and SPD2010 SpeedVac Kits
Savant Refrigerated Vapor Traps
Featured vacuum concentrators and kits categories
Providing solutions for a wide variety of solvent applications, Thermo Scientific™ SpeedVac systems use state-of-the-art centrifugation, vacuum, and heat technologies to remove solvents and concentrated samples. From traditional drying down of DNA preparations in water and methanol—to the more complex and aggressive applications presented by the combinatorical chemistry and drug discovery sectors of the pharmaceutical industry—our centrifugal vacuum concentrators remain a market leader.
Each one of our vacuum concentrator kits consists of four components: concentrator, trap, rotor, and pump. In addition, all kits have all the necessary components and accessories under one part number. Interchangeable accessories include flask supports, vials, ampules, trays, and microplates.
We offer a complete range of vacuum concentrator accessories to suit a wide variety of applications and sample formats. Concentrator rotors, vacuum oil pumps, digital vacuum gauges, and universal vacuum systems will help complete your vacuum concentrator system.
Resources
Support
Unity Lab Services: Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.