Pluripotent Stem Cell (PSC) Culture Media
PSC research requires careful attention to culture conditions. From KnockOut media to Essential 8 media and cell therapy research media, Gibco media deliver culture with confidence. Find the right solution for your stem cell culture needs.
Request information   Register for workshop
Which pluripotent stem cell culture medium is right for you?
Product
KnockOut Serum Replacement – MultiSpecies
CTS KnockOut SR XenoFree Medium
StemFlex Medium
Essential 8 Medium
CTS Essential 8 Medium
Ideal for
Feeder-based human and mouse PSC culture, reprogramming, gene editing, and differentiation
Translational or clinical research applications
Robust maintenance of PSC culture, difficult cell lines, and superior performance in single cell passaging and gene editing
Consistent PSC culture and superior reprogramming
Translational or clinical research applications
Defined
Animal origin components (BSA)
Xeno-free; no animal derived components, human derived
Animal origin components
(BSA)
Xeno-free; no animal derived components, human derived
Animal origin free (AOF)
Recommended cell types
Mammalian PSCs
Human PSCs
Human PSCs
Human PSCs Human PSCs
Weekend-free feeding schedule
No
No
Yes
Yes
Essential 8 Flex Medium
No
Genome editing
Fair
Fair
Best
Fair
Fair
Robustness
Good
Good
Best
Fair
Fair
Recommended
matrix
Gibco Mouse Embryonic Fibroblasts and Attachment Factor (for human and mouse) Geltrex Matrix
Geltrex Matrix
rhLaminin-521
Vitronectin (VTN-N) Recombinant Protein
rhLaminin-521
CTS Vitronectin (VTN-N) Recombinant Protein
rhLaminin-521
Recommended level of dissociation and passaging reagent
Clump
Gibco Collagenase IV
Versene Solution
Versene Solution
Versene Solution
Versene Solution
Small cluster
N/A
N/A
StemPro Accutase Cell Dissociation Reagent, 
RevitaCell Supplement helpful but not required
StemPro Accutase Cell Dissociation Reagent, optional RevitaCell Supplement added to medium during first 18-24 hours post-passage for improved recovery
StemPro Accutase Cell Dissociation Reagent, optional RevitaCell Supplement added to medium during first 18-24
hours post-passage for improved recovery
Single cell
TrypLE Select  Enzyme (for mouse)
Gibco CTS TrypLE Select Enzyme**
TrypLE Select or Tryple Express Enzyme, RevitaCell Supplement helpful but not required during recovery if using rhLaminin-521
TrypLE Select or Tryple Express Enzyme with RevitaCell Supplement added to medium during first 18-24 hours post-passage
CTS TrypLE Select Enzyme with RevitaCell Supplement added to medium during first 18-24 hours post-passage
Unless otherwise indicated, all products are For Research Use Only. Not for use in diagnostic procedures
*For human ex vivo tissue and cell culture processing applications. CAUTION: When used as a medical device, Federal Law restricts this device to sale by or on the order of a physician.
**For Research Use or Noncommercial Manufacturing of Cell-Based Products for Clinical Research. CAUTION: Not for direct administration into humans or animals.
Resources