The all-in-one qPCR instrument
Combining flexible throughput capabilities with a streamlined workflow, the Applied Biosystems™ QuantStudio™ 12K Flex system takes you from targeted discovery through confirmation and screening, all on a single platform.
Request a quote
QuantStudio 12K Flex
Maximum Throughput
Maximize throughput with minimal resources.
Increase your throughput by as much as 32-fold by running 4 Applied Biosystems™ OpenArray™ plates in a single run on the Applied Biosystems™ QuantStudio™ 12K Flex system. It's the equivalent of 32 traditional 384-well qPCR plates or 12,000 data points.
Explore OpenArray Technology ›
Push your throughput even more.
When equipped with the OpenArray block, a single 9700 thermal cycler and the QuantStudio 12K Flex Applied Biosystems™ OpenArray™ AccuFill™ System, the QuantStudio 12K Flex system can produce up to 110,000 data points or more in an eight-hour day. 
Reduce start up time and hands on time.
Start a 12,000 data point experiment typically in 20 minutes or less.
Scalability
Execute today and plan for tomorrow.
As you scale your experiments and move up in throughput, be assured that Applied Biosystems™ TaqMan™ Assay sensitivity, specificity, and wide dynamic range are optimized across all QuantStudio 12K Flex system block formats.
Even if you just have a few samples/targets to analyze, there is a TaqMan Assay format to fit your research needs.
Flexibility
Scientist using QuantStudio
Five interchangeable blocks.
Choose and load the thermal-cycling block you need for the experiment at hand (OpenArray plate, TaqMan Array Card, 384-well, Fast 96-well, and 96-well blocks).
Provides a seamless switch from qPCR to digital PCR.
Increase the precision and sensitivity of your nucleic acid experiment by switching from qPCR to digital PCR mode using QuantStudio Digital PCR kits and DigitalSuite Software with the QuantStudio OpenArray block.
Maximum multiplexing & chemistry options.
Proven fluorescence detection with Enhanced OptiFlex System (e.g. white light LED and 21 filter combinations for extremely accurate and sensitive data collection.
Integrated analysis and data quality.
Comprehensive software analysis tools for gene expression, genotyping, and digital PCR are available at your fingertips. Laboratory Information Management Systems (LIMS) and features that assist with 21CFR Part 11 compliance are enabled for high-throughput, validated environments.
Applied Biosystems Service Plans:
The QuantStudio™ 12K Flex Real-Time PCR System is supported by:
  • Professional installation
  • Premium training package conducted by Field Scientists
  • Worldwide application, technical and field-training support
  AB® Complete AB® Assurance AB® Maintenance
Repair response time (business days) Guaranteed next day:
on-site repairs
Guaranteed 2-day:
on-site repairs
Target 2 day:
remedial repairs
Remote instrument monitoring and diagnostics + +
Priority phone and email access to instrument
support
+ + +
Instrument operating software upgrades +
Instrument qualification services +
Planned maintenance + + +
Parts, labor, and travel Included Included
Download Brochure: qPCR for who you are–QuantStudio real-time PCR and digital PCR systems
QuantStudio 12K Flex
Maximum Throughput
Maximize throughput with minimal resources.
Increase your throughput by as much as 32-fold by running 4 Applied Biosystems™ OpenArray™ plates in a single run on the Applied Biosystems™ QuantStudio™ 12K Flex system. It's the equivalent of 32 traditional 384-well qPCR plates or 12,000 data points.
Explore OpenArray Technology ›
Push your throughput even more.
When equipped with the OpenArray block, a single 9700 thermal cycler and the QuantStudio 12K Flex Applied Biosystems™ OpenArray™ AccuFill™ System, the QuantStudio 12K Flex system can produce up to 110,000 data points or more in an eight-hour day. 
Reduce start up time and hands on time.
Start a 12,000 data point experiment typically in 20 minutes or less.
Scalability
Execute today and plan for tomorrow.
As you scale your experiments and move up in throughput, be assured that Applied Biosystems™ TaqMan™ Assay sensitivity, specificity, and wide dynamic range are optimized across all QuantStudio 12K Flex system block formats.
Even if you just have a few samples/targets to analyze, there is a TaqMan Assay format to fit your research needs.
Flexibility
Scientist using QuantStudio
Five interchangeable blocks.
Choose and load the thermal-cycling block you need for the experiment at hand (OpenArray plate, TaqMan Array Card, 384-well, Fast 96-well, and 96-well blocks).
Provides a seamless switch from qPCR to digital PCR.
Increase the precision and sensitivity of your nucleic acid experiment by switching from qPCR to digital PCR mode using QuantStudio Digital PCR kits and DigitalSuite Software with the QuantStudio OpenArray block.
Maximum multiplexing & chemistry options.
Proven fluorescence detection with Enhanced OptiFlex System (e.g. white light LED and 21 filter combinations for extremely accurate and sensitive data collection.
Integrated analysis and data quality.
Comprehensive software analysis tools for gene expression, genotyping, and digital PCR are available at your fingertips. Laboratory Information Management Systems (LIMS) and features that assist with 21CFR Part 11 compliance are enabled for high-throughput, validated environments.
Applied Biosystems Service Plans:
The QuantStudio™ 12K Flex Real-Time PCR System is supported by:
  • Professional installation
  • Premium training package conducted by Field Scientists
  • Worldwide application, technical and field-training support
  AB® Complete AB® Assurance AB® Maintenance
Repair response time (business days) Guaranteed next day:
on-site repairs
Guaranteed 2-day:
on-site repairs
Target 2 day:
remedial repairs
Remote instrument monitoring and diagnostics + +
Priority phone and email access to instrument
support
+ + +
Instrument operating software upgrades +
Instrument qualification services +
Planned maintenance + + +
Parts, labor, and travel Included Included
Download Brochure: qPCR for who you are–QuantStudio real-time PCR and digital PCR systems
Powerful qPCR analysis tools
Applied Biosystems™ analysis modules make it  easier and faster to view, analyze, remotely access, store, and share qPCR data. Discover Standard Curve, Genotyping, and Relative Quantification analysis apps now.
To access qPCR apps, download Cloud Connect Utility ›