The Pathatrix® Auto System  has obtained AFNOR certification
The Pathatrix® Auto Instrument is an automated large volume re-circulating immuno-magnetic separation sample processing device used for rapid detection of specific pathogens from enriched pooled food samples. The Pathatrix® Auto System has obtained AFNOR certification for the ‘Pathatrix® Auto Salmonella spp. Protocol’. Validation data has proven equivalent with the reference method ISO 6579 (2002).
Request a demo
Food categories:
  • Meat products: including raw beef, ready-to-reheat meals and meat products
  • Dairy products: including heat-treated milk and UHT, canned or pasteurised dairy products
Salmonella detection workflows:
  • After enrichment, 10 portions of enriched meat or dairy samples are pooled over the Pathatrix® Auto System followed by detection using the MicroSEQ® Salmonella spp. real-time PCR detection kit or by detection on selective agar plates according to ISO 6579
  • Positive pooled samples are re-tested individually by reprocessing 10 ml samples of stored individual enriched cultures on the Pathatrix® Auto System, continuing through the chosen detection procedure
  • Independent confirmation testing is performed only on positive individual samples, according to the detection method chosen
Two protocols have been validated:
  • Pathatrix® Auto Salmonella spp. linked to MicroSEQ® Salmonella spp. real-time PCR Detection Kit
  • Pathatrix® Auto Salmonella spp. linked to Selective Agar Detection
Both protocols allow the screening of the negative samples within 2 days, while 3 days are required for the positive samples; compared to 4 days for culture negatives following ISO.
The Pathatrix® Auto Instrument is a recirculation immunomagnetic separation system. Using magnetic beads coated with antibodies, the Pathatrix® Auto Instrument quickly concentrates the target organisms such as Salmonella from the enriched samples, and the on-line wash step removes non-target organisms and any PCR inhibitors. This reduces the time to results and enables increased pathogen testing at lower costs.
Applying the Pathatrix® Auto System full workflows in your food testing lab means:
  • more testing at lower costs
  • faster release of your meat and dairy products
  • less false positives due the removal of matrix effects
Learn more
For testing of Food and Environmental samples only.
© 2013 Life Technologies Corporation. All rights reserved. The trademarks mentioned herein are the property of Life Technologies Corporation and/or its affiliate(s) or their respective owners.