Thermo Fisher Scientific Home  |  Careers  |  Locations  |  Site Map
« Return to Analytica Media Room
Thermo Fisher Scientific Offers Integrated Centrifugation Solution for Biological Applications
MUNICH, Germany (April 1, 2008) - Thermo Fisher Scientific Inc., the world leader in serving science, introduced its NALGENE 2L Bio Bottle, a two-liter centrifuge bottle for high-volume sample processing, at Analytica 2008. Designed with workflow in mind, NALGENE 2L Bio Bottles create a complete solution when combined with the Thermo Scientific Sorvall RC12BP centrifuge and H-12000 swinging-bucket rotor, enabling scientists to process 12 liters of sample in a single run. The NALGENE Bio Bottle will be showcased at the Thermo Scientific booth 105/204, Hall B1 at Analytica 2008, being held in Munich, Germany, from April 1-4.
Developed for gross separation of samples, including bacterial, yeast and tissue isolations performed in compliance with GMP (Good Manufactured Practice) regulations, NALGENE 2L Bio Bottles are highly suitable for biopharmaceutical applications.
The NALGENE 2L Bio Bottle and sealing cap are leakproof up to 7,333 x G and have an operating temperature range of 4°C to 22°C, with storage to 0°C. Constructed of chemical-resistant polypropylene copolymer, the bottles meet USP Class VI and are certified non-cytotoxic and non-hemolytic. A wide mouth opening simplifies decanting, re-suspensions and pellet removal, and the smooth inner walls are less likely to shear cells. Loading, recovery and sample storage are facilitated by its ergonomic design, which includes graduations for precise measurement. Furthermore, the bottles can also be used in an autoclave.
NALGENE 2L Bio Bottles are designed to fit the Sorvall H-12000 swinging-bucket rotor, an anodized aluminum rotor with a unique conical shape that improves pelleting. The system greatly increases process efficiency when used with the Sorvall RC12BP centrifuge, a dependable workhorse for large volume separations. This combination further illustrates the commitment of Thermo Fisher to provide high quality consumables and equipment that can be integrated to meet specific application-based requirements.
For additional information on NALGENE 2L Bio Bottles, please visit the Thermo Scientific booth 105/204, Hall B1 at Analytica 2008. Alternatively, please call +1-800-625-4327 or visit www.nalgene.com/centrifuge for further details. For information on the Thermo Scientific Sorvall RC12BP centrifuge and the Sorvall H 12000 swinging-bucket rotor, please visit www.thermo.com/ultimate.
NALGENE is part of Thermo Fisher Scientific, the world leader in serving science.
About Thermo Fisher Scientific
Thermo Fisher Scientific Inc. (NYSE: TMO) is the world leader in serving science, enabling our customers to make the world healthier, cleaner and safer. With annual revenues of $10 billion, we have more than 30,000 employees and serve over 350,000 customers within pharmaceutical and biotech companies, hospitals and clinical diagnostic labs, universities, research institutions and government agencies, as well as environmental and industrial process control settings. Serving customers through two premier brands, Thermo Scientific and Fisher Scientific, we help solve analytical challenges from routine testing to complex research and discovery. Thermo Scientific offers customers a complete range of high-end analytical instruments as well as laboratory equipment, software, services, consumables and reagents to enable integrated laboratory workflow solutions. Fisher Scientific provides a complete portfolio of laboratory equipment, chemicals, supplies and services used in healthcare, scientific research, safety and education. Together, we offer the most convenient purchasing options to customers and continuously advance our technologies to accelerate the pace of scientific discovery, enhance value for customers and fuel growth for shareholders and employees alike. Visit www.thermofisher.com.
« Return to Analytica Media Room
Contact Us
Lori Gorski
lori.gorski@thermofisher.com
+1 781-622-1242
Thermo Scientific NALGENE® 2L Bio Bottle
Thermo Scientific NALGENE® 2L Bio Bottle
Download Hi-Res version
215 KB, JPG