AvrII nucleotide recognition sequence
Figure color legend
Name derivation: Anabaena variabilis uw
Recognition and Cutting site: C^CTAG G
G GATC^C
Resulting DNA Ends: 5′ protruding ends
Prototype: AvrII
Isoschizomers: XmaJI, AspA2I, BlnI
Sensitivity to methylation:  None
Neoschizomers None
Anza RE generating compatible ends Anza™ 3 BcuI, Anza™ 6 NheI, Anza™ 12 XbaI
References