Name derivation: Haemophilus parainfluenzae
Recognition and Cutting site: GTT^AAC
CAA^TTG
Resulting DNA Ends: blunt
Prototype: HpaI
Isoschizomers: KspAI
Sensitivity to methylation:  CpG impaired if overlapped, EcoKI blocked if overlapped
Neoschizomers None
Anza RE generating compatible ends Any enzyme generating blunt ends
References