PvuI nucleotide recognition sequence
Figure color legend
Name derivation: Proteus vulgaris
Recognition and Cutting site: CG AT^CG
GC^AT GC
Resulting DNA Ends: 3′ protruding ends
Prototype: PvuI
Isoschizomers: Ple19I
Sensitivity to methylation:  CpG blocked
Neoschizomers None
Anza RE generating compatible ends Anza™ 18 PacI, Anza™ 40 SfaAI
References