HistoGrip™, Concentrate

Catalog number: 008050

 Related applications: Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

An easy-to-use product for adhering tissue to glass slides. Treating the glass with HistoGrip results in a strong bond between the tissue and slide. Normal enzyme digestions, antigen recovery procedures, and even the harsh processes of in situ hybridization will not remove tissues adhered with HistoGrip. Superior to poly-L-lysine. Sufficient for approximately 500-1000 slides.
For In Vitro Diagnostic Use.

Specifications

Reagent Type: Cell & Tissue Preparation Reagent
Product Line: HistoGrip™
Product Size: 10 mL
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols