Ki-67 Mouse Monoclonal Antibody, 2ndGen Predilute

Catalog number: 081156

 Related applications: Anatomical Pathology

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This monoclonal antibody reacts with Ki-67, a cell proliferation marker for all active stages of the cell cycle (G1, S, M, and G2) but not the G0 stage. Can be used to identify the proliferative rate of cell growth. Immunogen source: Recombinant peptide. Positive tissue: Breast carcinoma.

Immunogen: Recombinant peptide corresponding to a 1002 bp Human Ki-67 cDNA sequence.

Specificity: Ki-67

Reactivity: Human

Applications: Immunohistochemistry (FFPE)
For In Vitro Diagnostic Use.

Specifications

Reactivity: Human
Validated Application: Immunohistochemistry (FFPE)
Host Species: Mouse
Target Gene: Ki-67
Gene ID: 4288
Label or Dye: Unconjugated
Dilution: 2ndGen Predilute Antibody
Product Size: 6 mL
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols