Bcl-2 Mouse mAb (clone Bcl-2-100), 2ndGen predilute

Catalog number: 081193

 Related applications: Anatomical Pathology

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This monoclonal antibody reacts with Bcl-2 proto-oncogene protein. It is detectable in neoplastic cells of human lymphoma in which the 14:18 chromosonal translocation is present. Lymphoid medullary T Cells and lymphomas lacking detectable (14:18) translocation also express Bcl-2 protein. Bcl-2-100 is useful in the differentiation of reactive follicular lymphoid hyperplasia from follicular lymphoma. Positive tissue: Breast or prostate carcinoma/follicular lymphoma

Clone/PAD: Bcl-2-100

Immunogen: Synthetic peptide of human bcl-2 protein

Specificity: Bcl-2

Reactivity: Human

Applications: Immunohistochemistry (FFPE)

Pretreatment: Epitope Retrieval: Required (Citrate, pH 6.0), Enzyme Digestion: Not required

Expected Staining Pattern: Cytoplasmic
For In Vitro Diagnostic Use.

Specifications

Reactivity: Human
Validated Application: Immunohistochemistry (FFPE)
Host Species: Mouse
Target Gene: Bcl-2
Gene ID: 596
Label or Dye: Unconjugated
Dilution: 2ndGen Predilute Antibody
Product Size: 6 mL
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols