PBS, pH 7.4

Catalog number: 10010031

Gibco™  Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

PBS (phosphate buffered saline) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation. We offer a variety of PBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.

This PBS is manufactured as follows:
Without
• Calcium
• Magnesium
• Phenol Red

The complete formulation is available.

cGMP manufacturing and quality system
PBS is manufactured at a cGMP-compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical PBS product made in our Grand Island facility (20012-050). This facility is also registered with the FDA as a medical device manufacturer and certified to the ISO 13485 standards.

“This reagent is classified as a medical device in several countries. If it is not regulated as a medical device in specific region, it should be used for laboratory use only.”

Specifications

Concentrated: 1 X
pH: pH 7.4
Inorganic Salts: No Calcium, No Magnesium
Form: Liquid
pH Range: 7.3 - 7.5
Osmolality: 280 - 315 mOsm/kg
Product Size: 1,000 mL
Classification: Animal Origin-Free
Phenol Red Indicator: No Phenol Red
Sodium Pyruvate Additive: No Sodium Pyruvate
Shipping Condition: Room Temperature

Contents & storage

Storage conditions: 15 – 30°C
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture

Media formulations

Documents

Manuals & protocols

Recommended products