Interested in a bulk or custom version of this product? Request pricing.

EIF4EBP1 Recombinant Human Protein, N-His Tag

Catalog number: 10022H07E250

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Eukaryotic Translation Initiation Factor 4E Binding Protein 1 (EIF4EBP1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the human 4EBP1 (NP_004086.1) (Ser 2-Ile 118) fused to a polyhistidine tag at the N-terminus.

Activity: This recombinant protein has not been tested.

Formulation: 20 mM Tris, 500 mM NaCl, 10% glycerol, pH 8.0.

Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2 mg/mL or lower. It is recommended that the protein be aliquoted and used as soon as possible. Store aliquots under sterile conditions at -5 to -30°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Frozen
Purity: ≥ 85% by SDS-PAGE
Gene ID: 1978
Species: Human
Expression: E. coli
Gene Symbol: EIF4EBP1
Gene Aliases: 4E-BP1, EIF4EBP1, BP-1, MGC4316, PHAS-I
Product Size: 5 x 50 µg
Protein Form: Full Length, Recombinant
Purification: PAGE
Protein Family: Other Proteins
Molecular Weight: 13.4 kDa
Research Category: Signal Transduction
Protein Tag or Fusion: N-His
Validated Application: Assay Standard

Contents & storage

Includes 5 vials of protein in suspension buffer. Store in freezer (-5 to -30°C).

Documents

Manuals & protocols