This product has been Gibco FBS Fingerprinted. We developed a technology in partnership with Oritain, a global leader in verifying product origins. Based on geographical differences, we can create "fingerprints" of our FBS that connect them to their origin. Find out more ›

Fetal Bovine Serum, qualified, New Zealand origin

Catalog number: 10091155

Gibco™  Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Sera Category: Secure
Origin: New Zealand
Endotoxin level: <=10 EU/ml.
Hemoglobin level: <=30 mg/dl (levels routinely <=25 mg/dl).
For In Vitro Diagnostic Use.

Specifications

Serum Treatment: Standard (Sterile-Filtered)
Country of Origin: New Zealand
Format: Bottle(s)
Age: Fetal
Species: Cattle⁄Bovine
Product Size: 1,000 mL
Endotoxin Level: ≤ 10 EU/ml
Shipping Condition: Dry Ice

Contents & storage

Storage conditions: -5 to -20°C
Shipping conditions: Frozen

Documents

Manuals & protocols

Recommended products