Interested in a bulk or custom version of this product? Request pricing.

IL8 Recombinant Human Protein, hIgG1-Fc Tag

Catalog number: 10098H01H2U25

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Interleukin 8 (IL8), recombinant human protein is supplied as a lyophilized powder.

This recombinant protein was expressed from a DNA sequence encoding the 72 amino acid residue form (Ser 28-Ser 99) of mature human IL8 (NP_000575.1) fused to the Fc region of human IgG1 at the N-terminus.

Activity: This recombinant protein has not been tested.

Formulation: 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by SINO Biological.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Lyophilized
Purity: ≥ 85% by SDS-PAGE
Gene ID: 3576
Species: Human
Expression: Mammalian
Gene Symbol: IL8
Gene Aliases: CXCL8, GCP-1, GCP1, LECT, LUCT, LYNAP, MDNCF, MONAP, NAF, NAP-1, NAP1
Product Size: 5 x 5 µg
Protein Form: Truncated, Recombinant
Purification: PAGE
Tag Position: C-Terminal
Protein Family: Chemokines & Receptors
Endotoxin Level: < 1 EU/µg
Molecular Weight: 35 kDa
Research Category: Immunology, Signal Transduction
Protein Tag or Fusion: IgG1 Fc Tag
Validated Application: Assay Standard

Contents & storage

Includes 5 vials of lyophilized protein. Store at -20 °C.

Documents

Manuals & protocols