Interested in a bulk or custom version of this product? Request pricing.

IL1RL1 Recombinant Human Protein, hIgG1-Fc Tag

Catalog number: 10105H02H25

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Interleukin 1 receptor-like 1 (IL1RL1), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the human IL1RL1, isoform 2 (NP_003847.2), residues 1 to 328, fused to the Fc region of human IgG1 at the C-terminus.

Activity: Measured by its binding ability in a functional ELISA . Immobilized human IL33 at 10 µg/ml(100 µl/well) can bind human IL1RL1-Fc with a linear range of 0.31-5 ng/ml.

Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Lyophilized
Purity: ≥ 90% by SDS-PAGE
Gene ID: 9173
Species: Human
Expression: Mammalian
Gene Symbol: IL1RL1
Gene Aliases: IL-1RL1, T1, ST2, DER4, ST2L, ST2V, FIT-1, MGC32623
Product Size: 5 x 5 µg
Protein Form: Recombinant
Protein Family: Cytokines & Receptors
Endotoxin Level: < 1 EU/µg
Protein Subtype: Interleukins, Cytokine Receptors
Molecular Weight: 67.1 kDa
Research Category: Immunology, Cardiovascular Research, Inflammation, Biomarker-Related Pathway(s), Signal Transduction
Protein Tag or Fusion: IgG1 Fc Tag
Validated Application: Assay Standard
Shipping Condition: Room Temperature

Contents & storage

Store at -20 to -70 °C

Documents

Manuals & protocols

Recommended products