Interested in a bulk or custom version of this product? Request pricing.

MCAM (CD146) Recombinant Human Protein, hIgG1-Fc Tag

Catalog number: 10115H02H250

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Melanoma cell adhesion molecule (MCAM/CD146), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in cell biology research applications. It can also be used as a protein standard.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gly 559) of human MCAM precursor (NP_006491.2) fused to the Fc region of human IgG1 at the C-terminus.

Activity: Measured by the ability of the immobilized protein to support the adhesion of the HUVEC human umbilical vein endothelial cell line . When 3 x 104 cells/well are added to human CD146 coated plates (0.8 µg/ml, 100 µl/well), approximately >30 % will adhere after one hour at 37°C.

Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Angiogenesis, Cardiovascular Research, Development, Extracellular Matrix & Cell Adhesion, Immunology, Oncology, Signal Transduction
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: MCAM, CD146, MUC18
Gene ID: 4162
Gene Symbol: MCAM
Molecular Weight: 86.4 kDa
Product Line: Sino Biological®
Product Size: 5 x 50 µg
Protein Family: CD Proteins
Protein Form: Recombinant, Truncated
Protein Name: melanoma cell adhesion molecule
Protein Subtype: Other Proteins
Protein Tag or Fusion: IgG1 Fc Tag
Purity: ≥ 90% by SDS-PAGE
Species: Human
Validated Application: Assay Standard
Shipping Condition: Room Temperature

Contents & storage

Store at -20 to -70 °C

Documents

Manuals & protocols